Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27814188-27814698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2AJ
Entrez Gene ID
8331
Gene Name
histone cluster 1, H2aj
Gene Aliases
H2A/E, H2AFE, dJ160A22.4
UniGene
Hs.406691
Gene Chromosome Location
Chr.6: 27814302-27814740 on Build GRCh38
Transcripts
NM_021066.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779773 c.301G>A p.V101I Chr.6: 27814440-27814440 on Build GRCh38
5962427 c.133G>A p.G45S Chr.6: 27814608-27814608 on Build GRCh38
1734122 c.151T>C p.Y51H Chr.6: 27814590-27814590 on Build GRCh38
6197202 c.204C>A p.G68G Chr.6: 27814537-27814537 on Build GRCh38
304787 c.58T>C p.S20P Chr.6: 27814683-27814683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764536652 Chr.6: 27814657-27814657 on Build GRCh38
rs909679228 Chr.6: 27814231-27814231 on Build GRCh38
rs553123298 Chr.6: 27814287-27814287 on Build GRCh38
rs771751755 Chr.6: 27814344-27814344 on Build GRCh38
rs760144793 Chr.6: 27814303-27814303 on Build GRCh38
rs796425066 Chr.6: 27814377-27814378 on Build GRCh38
rs765140362 Chr.6: 27814453-27814453 on Build GRCh38
rs537052297 Chr.6: 27814256-27814256 on Build GRCh38
rs45558641 Chr.6: 27814538-27814538 on Build GRCh38
rs1007745778 Chr.6: 27814447-27814447 on Build GRCh38
rs997137287 Chr.6: 27814250-27814250 on Build GRCh38
rs772388802 Chr.6: 27814324-27814326 on Build GRCh38
rs748335987 Chr.6: 27814666-27814667 on Build GRCh38
rs757484204 Chr.6: 27814377-27814377 on Build GRCh38
rs1041238333 Chr.6: 27814239-27814239 on Build GRCh38
rs758194782 Chr.6: 27814361-27814361 on Build GRCh38
rs200310270 Chr.6: 27814381-27814381 on Build GRCh38
rs759449450 Chr.6: 27814357-27814357 on Build GRCh38
rs201406770 Chr.6: 27814558-27814558 on Build GRCh38
rs769268872 Chr.6: 27814607-27814607 on Build GRCh38
rs767679934 Chr.6: 27814551-27814551 on Build GRCh38
rs767844961 Chr.6: 27814486-27814486 on Build GRCh38
rs560353752 Chr.6: 27814432-27814432 on Build GRCh38
rs745464227 Chr.6: 27814511-27814511 on Build GRCh38
rs766495794 Chr.6: 27814577-27814577 on Build GRCh38
rs781104813 Chr.6: 27814450-27814450 on Build GRCh38
rs770611224 Chr.6: 27814510-27814510 on Build GRCh38
rs536832765 Chr.6: 27814283-27814283 on Build GRCh38
rs765254443 Chr.6: 27814647-27814647 on Build GRCh38
rs148765429 Chr.6: 27814402-27814402 on Build GRCh38
rs202142647 Chr.6: 27814378-27814378 on Build GRCh38
rs756195484 Chr.6: 27814492-27814492 on Build GRCh38
rs200522974 Chr.6: 27814598-27814598 on Build GRCh38
rs553061767 Chr.6: 27814648-27814648 on Build GRCh38
rs762181990 Chr.6: 27814485-27814485 on Build GRCh38
rs150955249 Chr.6: 27814627-27814627 on Build GRCh38
rs774352500 Chr.6: 27814543-27814543 on Build GRCh38
rs773751331 Chr.6: 27814348-27814348 on Build GRCh38
rs759590946 Chr.6: 27814641-27814641 on Build GRCh38
rs752242859 Chr.6: 27814359-27814360 on Build GRCh38
rs915926569 Chr.6: 27814215-27814215 on Build GRCh38
rs888670807 Chr.6: 27814259-27814259 on Build GRCh38
rs761666821 Chr.6: 27814546-27814546 on Build GRCh38
rs779224891 Chr.6: 27814630-27814631 on Build GRCh38
rs751865437 Chr.6: 27814369-27814369 on Build GRCh38
rs769380786 Chr.6: 27814519-27814519 on Build GRCh38
rs772827641 Chr.6: 27814346-27814346 on Build GRCh38
rs776064316 Chr.6: 27814349-27814349 on Build GRCh38
rs778114510 Chr.6: 27814317-27814317 on Build GRCh38
rs372650213 Chr.6: 27814498-27814498 on Build GRCh38
rs566748497 Chr.6: 27814238-27814238 on Build GRCh38
rs761016061 Chr.6: 27814624-27814624 on Build GRCh38
rs575196700 Chr.6: 27814379-27814379 on Build GRCh38
rs9368532 Chr.6: 27814232-27814232 on Build GRCh38
rs1034849543 Chr.6: 27814295-27814295 on Build GRCh38
rs762300071 Chr.6: 27814377-27814378 on Build GRCh38
rs749398014 Chr.6: 27814570-27814598 on Build GRCh38
rs200560117 Chr.6: 27814417-27814417 on Build GRCh38
rs749661944 Chr.6: 27814398-27814398 on Build GRCh38
rs1005229686 Chr.6: 27814266-27814266 on Build GRCh38
rs755548266 Chr.6: 27814579-27814579 on Build GRCh38
rs762946659 Chr.6: 27814465-27814465 on Build GRCh38
rs552453097 Chr.6: 27814474-27814474 on Build GRCh38
rs778525630 Chr.6: 27814672-27814672 on Build GRCh38
rs750436061 Chr.6: 27814456-27814460 on Build GRCh38
rs755420196 Chr.6: 27814502-27814502 on Build GRCh38
rs573056214 Chr.6: 27814669-27814669 on Build GRCh38
rs762262435 Chr.6: 27814315-27814316 on Build GRCh38
rs747137197 Chr.6: 27814328-27814328 on Build GRCh38
rs748509780 Chr.6: 27814508-27814508 on Build GRCh38
rs779307470 Chr.6: 27814507-27814507 on Build GRCh38
rs774407719 Chr.6: 27814613-27814613 on Build GRCh38
rs766351369 Chr.6: 27814306-27814306 on Build GRCh38
rs199658559 Chr.6: 27814318-27814318 on Build GRCh38
rs756115733 Chr.6: 27814462-27814464 on Build GRCh38
rs892562408 Chr.6: 27814257-27814257 on Build GRCh38
rs758768724 Chr.6: 27814650-27814650 on Build GRCh38
rs775772955 Chr.6: 27814337-27814337 on Build GRCh38
rs766505750 Chr.6: 27814626-27814626 on Build GRCh38
rs201764606 Chr.6: 27814376-27814376 on Build GRCh38
rs749658664 Chr.6: 27814338-27814338 on Build GRCh38
rs774633066 Chr.6: 27814416-27814416 on Build GRCh38
rs752479134 Chr.6: 27814360-27814360 on Build GRCh38
rs1017837899 Chr.6: 27814287-27814288 on Build GRCh38
rs980642894 Chr.6: 27814545-27814545 on Build GRCh38
rs760271262 Chr.6: 27814348-27814348 on Build GRCh38
rs34452669 Chr.6: 27814231-27814232 on Build GRCh38
rs764789471 Chr.6: 27814357-27814358 on Build GRCh38
rs377069608 Chr.6: 27814501-27814501 on Build GRCh38
rs753848923 Chr.6: 27814578-27814578 on Build GRCh38
rs926643691 Chr.6: 27814240-27814240 on Build GRCh38
rs771932401 Chr.6: 27814619-27814619 on Build GRCh38
rs990689274 Chr.6: 27814214-27814214 on Build GRCh38
rs201234682 Chr.6: 27814235-27814235 on Build GRCh38
rs746680206 Chr.6: 27814420-27814420 on Build GRCh38
rs748854027 Chr.6: 27814668-27814668 on Build GRCh38
rs781378620 Chr.6: 27814329-27814329 on Build GRCh38
rs997875527 Chr.6: 27814294-27814294 on Build GRCh38
rs758787555 Chr.6: 27814594-27814594 on Build GRCh38
rs751853187 Chr.6: 27814658-27814658 on Build GRCh38
rs142430355 Chr.6: 27814438-27814438 on Build GRCh38
rs188809245 Chr.6: 27814293-27814293 on Build GRCh38
rs779539257 Chr.6: 27814662-27814662 on Build GRCh38
rs1043776003 Chr.6: 27814399-27814399 on Build GRCh38
rs757151249 Chr.6: 27814380-27814381 on Build GRCh38
rs558904372 Chr.6: 27814244-27814244 on Build GRCh38
rs763982404 Chr.6: 27814468-27814468 on Build GRCh38
rs201651725 Chr.6: 27814675-27814675 on Build GRCh38
rs200127614 Chr.6: 27814230-27814231 on Build GRCh38
rs770272995 Chr.6: 27814336-27814336 on Build GRCh38
rs746363860 Chr.6: 27814335-27814335 on Build GRCh38
rs532355124 Chr.6: 27814450-27814450 on Build GRCh38
rs761791554 Chr.6: 27814618-27814618 on Build GRCh38
rs768751850 Chr.6: 27814611-27814611 on Build GRCh38
rs574090628 Chr.6: 27814280-27814280 on Build GRCh38
rs772815758 Chr.6: 27814622-27814622 on Build GRCh38
rs1039585296 Chr.6: 27814456-27814456 on Build GRCh38
rs745543290 Chr.6: 27814603-27814603 on Build GRCh38
rs758619153 Chr.6: 27814315-27814315 on Build GRCh38
rs1056398811 Chr.6: 27814350-27814350 on Build GRCh38
rs9380019 Chr.6: 27814235-27814235 on Build GRCh38
rs199735306 Chr.6: 27814630-27814630 on Build GRCh38
rs201386125 Chr.6: 27814387-27814387 on Build GRCh38
rs773126600 Chr.6: 27814679-27814679 on Build GRCh38
rs1022231418 Chr.6: 27814334-27814334 on Build GRCh38
rs750508104 Chr.6: 27814489-27814489 on Build GRCh38
rs779668241 Chr.6: 27814609-27814609 on Build GRCh38
rs960659586 Chr.6: 27814564-27814564 on Build GRCh38
rs761152407 Chr.6: 27814354-27814357 on Build GRCh38
rs201801053 Chr.6: 27814393-27814393 on Build GRCh38
rs992034808 Chr.6: 27814596-27814596 on Build GRCh38
rs774799647 Chr.6: 27814527-27814527 on Build GRCh38
rs200393289 Chr.6: 27814537-27814537 on Build GRCh38
rs763935373 Chr.6: 27814367-27814367 on Build GRCh38
rs775523081 Chr.6: 27814461-27814461 on Build GRCh38
rs904023285 Chr.6: 27814278-27814278 on Build GRCh38
rs753112022 Chr.6: 27814589-27814589 on Build GRCh38
rs190854526 Chr.6: 27814241-27814241 on Build GRCh38
rs779788905 Chr.6: 27814517-27814519 on Build GRCh38
rs755858444 Chr.6: 27814659-27814659 on Build GRCh38
rs779362597 Chr.6: 27814582-27814582 on Build GRCh38
rs183907264 Chr.6: 27814270-27814270 on Build GRCh38
rs760768500 Chr.6: 27814563-27814563 on Build GRCh38
rs369040595 Chr.6: 27814314-27814314 on Build GRCh38
rs144406971 Chr.6: 27814229-27814229 on Build GRCh38
rs376510829 Chr.6: 27814309-27814309 on Build GRCh38
rs768567701 Chr.6: 27814624-27814625 on Build GRCh38
rs751125305 Chr.6: 27814380-27814380 on Build GRCh38
rs545258692 Chr.6: 27814310-27814310 on Build GRCh38
rs370155 Chr.6: 27814253-27814253 on Build GRCh38

View Full Product Details