Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36916367-36916883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf89
Entrez Gene ID
221477
Gene Name
chromosome 6 open reading frame 89
Gene Aliases
BRAP
UniGene
Hs.433381
Gene Chromosome Location
Chr.6: 36834886-36928964 on Build GRCh38
Transcripts
NM_001286635.1 NM_001286636.1 NM_001286637.1 NM_152734.3 XM_011514377.2 XM_017010432.1 XM_017010433.1 XM_017010434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752882 c.846+1G>T p.? Chr.6: 36916575-36916575 on Build GRCh38
742169 c.779C>T p.P260L Chr.6: 36916507-36916507 on Build GRCh38
4667955 c.749G>A p.R250H Chr.6: 36916477-36916477 on Build GRCh38
3626296 c.839G>T p.G280V Chr.6: 36916567-36916567 on Build GRCh38
159653 c.806A>T p.K269M Chr.6: 36916534-36916534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748113353 Chr.6: 36916553-36916553 on Build GRCh38
rs570245444 Chr.6: 36916576-36916576 on Build GRCh38
rs1041239750 Chr.6: 36916797-36916797 on Build GRCh38
rs758281292 Chr.6: 36916555-36916555 on Build GRCh38
rs80341332 Chr.6: 36916427-36916427 on Build GRCh38
rs139497629 Chr.6: 36916794-36916794 on Build GRCh38
rs907764716 Chr.6: 36916676-36916676 on Build GRCh38
rs765938617 Chr.6: 36916515-36916515 on Build GRCh38
rs144834171 Chr.6: 36916477-36916477 on Build GRCh38
rs563186853 Chr.6: 36916787-36916787 on Build GRCh38
rs1021185692 Chr.6: 36916670-36916670 on Build GRCh38
rs553066293 Chr.6: 36916742-36916742 on Build GRCh38
rs901429611 Chr.6: 36916827-36916827 on Build GRCh38
rs777665393 Chr.6: 36916568-36916568 on Build GRCh38
rs1054843984 Chr.6: 36916405-36916405 on Build GRCh38
rs762462243 Chr.6: 36916625-36916625 on Build GRCh38
rs1020313878 Chr.6: 36916774-36916774 on Build GRCh38
rs374930621 Chr.6: 36916617-36916617 on Build GRCh38
rs545543809 Chr.6: 36916804-36916804 on Build GRCh38
rs989606778 Chr.6: 36916654-36916654 on Build GRCh38
rs764215385 Chr.6: 36916497-36916497 on Build GRCh38
rs536515496 Chr.6: 36916503-36916503 on Build GRCh38
rs35567953 Chr.6: 36916599-36916599 on Build GRCh38
rs373515629 Chr.6: 36916680-36916680 on Build GRCh38
rs564543641 Chr.6: 36916793-36916793 on Build GRCh38
rs762372195 Chr.6: 36916499-36916499 on Build GRCh38
rs140691016 Chr.6: 36916467-36916467 on Build GRCh38
rs778581694 Chr.6: 36916518-36916518 on Build GRCh38
rs987170405 Chr.6: 36916671-36916671 on Build GRCh38
rs1034438109 Chr.6: 36916406-36916406 on Build GRCh38
rs148571454 Chr.6: 36916540-36916540 on Build GRCh38
rs763113395 Chr.6: 36916491-36916491 on Build GRCh38
rs774397220 Chr.6: 36916652-36916652 on Build GRCh38
rs375478179 Chr.6: 36916521-36916521 on Build GRCh38
rs974424198 Chr.6: 36916575-36916575 on Build GRCh38
rs141618907 Chr.6: 36916442-36916442 on Build GRCh38
rs937952270 Chr.6: 36916703-36916703 on Build GRCh38
rs778327673 Chr.6: 36916551-36916551 on Build GRCh38
rs748339621 Chr.6: 36916538-36916540 on Build GRCh38
rs755828718 Chr.6: 36916513-36916513 on Build GRCh38
rs143348146 Chr.6: 36916476-36916476 on Build GRCh38
rs770885611 Chr.6: 36916641-36916641 on Build GRCh38
rs772840762 Chr.6: 36916465-36916465 on Build GRCh38
rs903426375 Chr.6: 36916784-36916784 on Build GRCh38
rs1033225860 Chr.6: 36916788-36916788 on Build GRCh38
rs775589299 Chr.6: 36916592-36916592 on Build GRCh38
rs771857654 Chr.6: 36916462-36916462 on Build GRCh38
rs2293394 Chr.6: 36916714-36916714 on Build GRCh38
rs1028673612 Chr.6: 36916853-36916853 on Build GRCh38
rs1004848482 Chr.6: 36916505-36916505 on Build GRCh38
rs748714460 Chr.6: 36916436-36916436 on Build GRCh38
rs755676166 Chr.6: 36916422-36916422 on Build GRCh38
rs749458274 Chr.6: 36916597-36916597 on Build GRCh38
rs921693118 Chr.6: 36916635-36916635 on Build GRCh38
rs774526409 Chr.6: 36916619-36916619 on Build GRCh38
rs767949783 Chr.6: 36916501-36916501 on Build GRCh38
rs200678588 Chr.6: 36916588-36916588 on Build GRCh38
rs754715762 Chr.6: 36916425-36916426 on Build GRCh38
rs750124003 Chr.6: 36916417-36916417 on Build GRCh38
rs768643969 Chr.6: 36916441-36916441 on Build GRCh38
rs996804299 Chr.6: 36916590-36916590 on Build GRCh38
rs1019836640 Chr.6: 36916535-36916535 on Build GRCh38
rs147237659 Chr.6: 36916455-36916455 on Build GRCh38
rs927871002 Chr.6: 36916694-36916694 on Build GRCh38
rs760332988 Chr.6: 36916765-36916765 on Build GRCh38
rs748021103 Chr.6: 36916460-36916460 on Build GRCh38
rs566867077 Chr.6: 36916805-36916805 on Build GRCh38
rs541744444 Chr.6: 36916760-36916760 on Build GRCh38
rs898961831 Chr.6: 36916722-36916722 on Build GRCh38
rs73420354 Chr.6: 36916751-36916751 on Build GRCh38
rs150515659 Chr.6: 36916729-36916729 on Build GRCh38
rs559090939 Chr.6: 36916636-36916636 on Build GRCh38

View Full Product Details