Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44140936-44141442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM63B
Entrez Gene ID
55362
Gene Name
transmembrane protein 63B
Gene Aliases
C6orf110
UniGene
Hs.414473
Gene Chromosome Location
Chr.6: 44126628-44155519 on Build GRCh38
Transcripts
NM_001318792.1 NM_018426.2 XM_005249211.3 XM_005249213.4 XM_005249214.4 XM_005249215.4 XM_005249217.1 XM_006715135.1 XM_017010999.1 XM_017011000.1 XM_017011001.1 XM_017011002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5815656 c.749A>T p.Y250F Chr.6: 44141065-44141065 on Build GRCh38
4799162 c.766A>T p.I256F Chr.6: 44141082-44141082 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534792638 Chr.6: 44141237-44141237 on Build GRCh38
rs900718680 Chr.6: 44141238-44141238 on Build GRCh38
rs774066080 Chr.6: 44141142-44141142 on Build GRCh38
rs1003537342 Chr.6: 44141151-44141151 on Build GRCh38
rs973872839 Chr.6: 44141223-44141223 on Build GRCh38
rs893557911 Chr.6: 44141353-44141353 on Build GRCh38
rs891915425 Chr.6: 44141273-44141273 on Build GRCh38
rs372108714 Chr.6: 44140988-44140988 on Build GRCh38
rs912129782 Chr.6: 44141265-44141265 on Build GRCh38
rs911526975 Chr.6: 44141193-44141193 on Build GRCh38
rs142579579 Chr.6: 44141066-44141066 on Build GRCh38
rs751580304 Chr.6: 44141042-44141042 on Build GRCh38
rs747660423 Chr.6: 44141149-44141149 on Build GRCh38
rs1012491244 Chr.6: 44141370-44141370 on Build GRCh38
rs560734182 Chr.6: 44140961-44140961 on Build GRCh38
rs773935604 Chr.6: 44140994-44140994 on Build GRCh38
rs12660230 Chr.6: 44141121-44141121 on Build GRCh38
rs369979589 Chr.6: 44141138-44141138 on Build GRCh38
rs1000386577 Chr.6: 44141246-44141246 on Build GRCh38
rs759790223 Chr.6: 44141012-44141012 on Build GRCh38
rs1000334238 Chr.6: 44141010-44141010 on Build GRCh38
rs4628095 Chr.6: 44141200-44141200 on Build GRCh38
rs111988028 Chr.6: 44141413-44141413 on Build GRCh38
rs752885932 Chr.6: 44141018-44141018 on Build GRCh38
rs1053556409 Chr.6: 44140991-44140991 on Build GRCh38
rs765545700 Chr.6: 44141014-44141014 on Build GRCh38
rs12528844 Chr.6: 44141111-44141111 on Build GRCh38
rs944986391 Chr.6: 44141271-44141271 on Build GRCh38
rs1044939632 Chr.6: 44141404-44141404 on Build GRCh38
rs929028023 Chr.6: 44141179-44141179 on Build GRCh38
rs761280774 Chr.6: 44140995-44140995 on Build GRCh38
rs1053912370 Chr.6: 44141291-44141291 on Build GRCh38
rs375770219 Chr.6: 44141268-44141268 on Build GRCh38
rs1010256254 Chr.6: 44141090-44141090 on Build GRCh38
rs755839712 Chr.6: 44141112-44141112 on Build GRCh38
rs748920933 Chr.6: 44141132-44141132 on Build GRCh38
rs982435496 Chr.6: 44141116-44141116 on Build GRCh38
rs1040718621 Chr.6: 44140971-44140971 on Build GRCh38
rs12661573 Chr.6: 44141122-44141122 on Build GRCh38
rs764321448 Chr.6: 44141115-44141116 on Build GRCh38
rs777122799 Chr.6: 44141005-44141005 on Build GRCh38
rs775512339 Chr.6: 44141389-44141389 on Build GRCh38
rs766982137 Chr.6: 44141004-44141004 on Build GRCh38
rs140159200 Chr.6: 44141030-44141030 on Build GRCh38
rs971796664 Chr.6: 44141392-44141392 on Build GRCh38
rs1054601318 Chr.6: 44141266-44141266 on Build GRCh38
rs779845952 Chr.6: 44141128-44141128 on Build GRCh38
rs920362845 Chr.6: 44141148-44141148 on Build GRCh38
rs12529211 Chr.6: 44141275-44141275 on Build GRCh38
rs532214388 Chr.6: 44141176-44141176 on Build GRCh38
rs749435382 Chr.6: 44141186-44141186 on Build GRCh38
rs1014962872 Chr.6: 44141175-44141175 on Build GRCh38
rs1033100006 Chr.6: 44141011-44141011 on Build GRCh38
rs781100569 Chr.6: 44141110-44141110 on Build GRCh38
rs553624412 Chr.6: 44141341-44141341 on Build GRCh38
rs752082189 Chr.6: 44141145-44141146 on Build GRCh38
rs986320873 Chr.6: 44141248-44141248 on Build GRCh38
rs758579128 Chr.6: 44141025-44141025 on Build GRCh38
rs772048881 Chr.6: 44141136-44141136 on Build GRCh38
rs1001509667 Chr.6: 44141408-44141408 on Build GRCh38
rs774305747 Chr.6: 44141035-44141035 on Build GRCh38
rs926511742 Chr.6: 44141119-44141119 on Build GRCh38
rs187171059 Chr.6: 44140987-44140987 on Build GRCh38
rs1039224498 Chr.6: 44140997-44140997 on Build GRCh38
rs768298946 Chr.6: 44140992-44140992 on Build GRCh38
rs1047417336 Chr.6: 44141185-44141185 on Build GRCh38

View Full Product Details