Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46859306-46859803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF5
Entrez Gene ID
221395
Gene Name
adhesion G protein-coupled receptor F5
Gene Aliases
GPR116, KPG_001
UniGene
Hs.362806
Gene Chromosome Location
Chr.6: 46852505-46954954 on Build GRCh38
Transcripts
NM_001098518.1 NM_015234.4 XM_005248892.2 XM_005248893.3 XM_005248894.3 XM_005248895.3 XM_011514353.2 XM_011514354.2 XM_017010418.1 XM_017010419.1 XM_017010420.1 XM_017010421.1 XM_017010422.1 XM_017010423.1 XM_017010424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4817320 c.2527C>A p.P843T Chr.6: 46859376-46859376 on Build GRCh38
5870829 c.2498C>T p.S833F Chr.6: 46859405-46859405 on Build GRCh38
3874560 c.2438T>G p.V813G Chr.6: 46859465-46859465 on Build GRCh38
5830493 c.854T>C p.M285T Chr.6: 46859336-46859336 on Build GRCh38
6231819 c.877G>A p.E293K Chr.6: 46859313-46859313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763150735 Chr.6: 46859570-46859570 on Build GRCh38
rs960868596 Chr.6: 46859393-46859393 on Build GRCh38
rs781391045 Chr.6: 46859549-46859549 on Build GRCh38
rs756296075 Chr.6: 46859457-46859457 on Build GRCh38
rs150267672 Chr.6: 46859357-46859357 on Build GRCh38
rs533986341 Chr.6: 46859466-46859466 on Build GRCh38
rs779781103 Chr.6: 46859536-46859536 on Build GRCh38
rs200056704 Chr.6: 46859500-46859500 on Build GRCh38
rs750867567 Chr.6: 46859344-46859344 on Build GRCh38
rs527287317 Chr.6: 46859728-46859728 on Build GRCh38
rs758833940 Chr.6: 46859371-46859372 on Build GRCh38
rs369658688 Chr.6: 46859686-46859686 on Build GRCh38
rs776909869 Chr.6: 46859467-46859467 on Build GRCh38
rs9395218 Chr.6: 46859502-46859502 on Build GRCh38
rs560181166 Chr.6: 46859704-46859704 on Build GRCh38
rs375810557 Chr.6: 46859554-46859554 on Build GRCh38
rs374021268 Chr.6: 46859445-46859445 on Build GRCh38
rs1022616734 Chr.6: 46859383-46859383 on Build GRCh38
rs1002446969 Chr.6: 46859390-46859390 on Build GRCh38
rs376295199 Chr.6: 46859332-46859332 on Build GRCh38
rs776695518 Chr.6: 46859579-46859579 on Build GRCh38
rs200248669 Chr.6: 46859358-46859358 on Build GRCh38
rs758149594 Chr.6: 46859527-46859527 on Build GRCh38
rs610856 Chr.6: 46859389-46859389 on Build GRCh38
rs923188020 Chr.6: 46859585-46859587 on Build GRCh38
rs762104523 Chr.6: 46859482-46859482 on Build GRCh38
rs143997423 Chr.6: 46859598-46859598 on Build GRCh38
rs746954473 Chr.6: 46859542-46859542 on Build GRCh38
rs755583854 Chr.6: 46859367-46859367 on Build GRCh38
rs771789130 Chr.6: 46859438-46859438 on Build GRCh38
rs756795131 Chr.6: 46859515-46859515 on Build GRCh38
rs1044199664 Chr.6: 46859739-46859739 on Build GRCh38
rs201123540 Chr.6: 46859349-46859349 on Build GRCh38
rs115182431 Chr.6: 46859737-46859737 on Build GRCh38
rs12374645 Chr.6: 46859397-46859397 on Build GRCh38
rs12374606 Chr.6: 46859491-46859491 on Build GRCh38
rs776848598 Chr.6: 46859339-46859339 on Build GRCh38
rs929986239 Chr.6: 46859361-46859361 on Build GRCh38
rs185180507 Chr.6: 46859562-46859562 on Build GRCh38
rs753382377 Chr.6: 46859510-46859510 on Build GRCh38
rs762277718 Chr.6: 46859363-46859363 on Build GRCh38
rs1053381190 Chr.6: 46859723-46859723 on Build GRCh38
rs147076717 Chr.6: 46859489-46859489 on Build GRCh38
rs141776519 Chr.6: 46859391-46859391 on Build GRCh38
rs770008329 Chr.6: 46859561-46859561 on Build GRCh38
rs746928110 Chr.6: 46859453-46859453 on Build GRCh38
rs200270176 Chr.6: 46859520-46859520 on Build GRCh38
rs190284325 Chr.6: 46859747-46859747 on Build GRCh38
rs12374647 Chr.6: 46859642-46859642 on Build GRCh38
rs564186831 Chr.6: 46859769-46859769 on Build GRCh38
rs582185 Chr.6: 46859773-46859773 on Build GRCh38
rs12374646 Chr.6: 46859557-46859557 on Build GRCh38
rs531430006 Chr.6: 46859335-46859335 on Build GRCh38
rs753583166 Chr.6: 46859496-46859496 on Build GRCh38
rs558626782 Chr.6: 46859484-46859484 on Build GRCh38
rs778142912 Chr.6: 46859537-46859543 on Build GRCh38
rs771310860 Chr.6: 46859572-46859572 on Build GRCh38
rs751996260 Chr.6: 46859542-46859542 on Build GRCh38
rs180960579 Chr.6: 46859398-46859398 on Build GRCh38
rs768524840 Chr.6: 46859545-46859545 on Build GRCh38
rs763633467 Chr.6: 46859509-46859509 on Build GRCh38
rs547499 Chr.6: 46859336-46859336 on Build GRCh38
rs780370039 Chr.6: 46859365-46859365 on Build GRCh38
rs765684377 Chr.6: 46859342-46859342 on Build GRCh38
rs1002540223 Chr.6: 46859764-46859764 on Build GRCh38
rs370634213 Chr.6: 46859411-46859411 on Build GRCh38
rs745536179 Chr.6: 46859521-46859521 on Build GRCh38
rs892993742 Chr.6: 46859724-46859724 on Build GRCh38
rs984864762 Chr.6: 46859513-46859513 on Build GRCh38
rs554543643 Chr.6: 46859395-46859395 on Build GRCh38

View Full Product Details