Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 85610014-85610515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNCRIP
Entrez Gene ID
10492
Gene Name
synaptotagmin binding cytoplasmic RNA interacting protein
Gene Aliases
GRY-RBP, GRYRBP, HNRNPQ, HNRPQ1, NSAP1, PP68, hnRNP-Q
UniGene
Hs.571177
Gene Chromosome Location
Chr.6: 85607784-85643862 on Build GRCh38
Transcripts
NM_001159673.1 NM_001159674.1 NM_001159676.1 NM_001159677.1 NM_001253771.1 XM_005248635.4 XM_017010176.1 XM_017010177.1 XM_017010178.1 XM_017010179.1

SNP Detail

SNP ID Chromosome Location
rs1029347177 Chr.6: 85610081-85610081 on Build GRCh38
rs938385126 Chr.6: 85610086-85610086 on Build GRCh38
rs546034658 Chr.6: 85610196-85610196 on Build GRCh38
rs1050697318 Chr.6: 85610475-85610476 on Build GRCh38
rs953794229 Chr.6: 85610122-85610122 on Build GRCh38
rs997792315 Chr.6: 85610416-85610416 on Build GRCh38
rs1028347637 Chr.6: 85610434-85610434 on Build GRCh38
rs1015135348 Chr.6: 85610056-85610056 on Build GRCh38
rs117793750 Chr.6: 85610380-85610380 on Build GRCh38
rs758636792 Chr.6: 85610080-85610081 on Build GRCh38
rs904397208 Chr.6: 85610227-85610227 on Build GRCh38
rs1049615197 Chr.6: 85610287-85610287 on Build GRCh38
rs939316667 Chr.6: 85610201-85610201 on Build GRCh38
rs548975250 Chr.6: 85610236-85610237 on Build GRCh38
rs1021295793 Chr.6: 85610195-85610195 on Build GRCh38
rs999090112 Chr.6: 85610231-85610231 on Build GRCh38
rs902126329 Chr.6: 85610407-85610411 on Build GRCh38
rs965659288 Chr.6: 85610371-85610371 on Build GRCh38
rs575559244 Chr.6: 85610116-85610116 on Build GRCh38
rs896816260 Chr.6: 85610069-85610069 on Build GRCh38
rs767115669 Chr.6: 85610401-85610404 on Build GRCh38
rs894120646 Chr.6: 85610175-85610175 on Build GRCh38
rs953990856 Chr.6: 85610449-85610449 on Build GRCh38
rs572764009 Chr.6: 85610452-85610452 on Build GRCh38
rs985634650 Chr.6: 85610161-85610161 on Build GRCh38
rs1012622947 Chr.6: 85610193-85610193 on Build GRCh38
rs997844821 Chr.6: 85610075-85610075 on Build GRCh38
rs1057214147 Chr.6: 85610173-85610173 on Build GRCh38
rs948137677 Chr.6: 85610228-85610228 on Build GRCh38
rs887917982 Chr.6: 85610290-85610295 on Build GRCh38
rs1031609427 Chr.6: 85610293-85610293 on Build GRCh38
rs532995720 Chr.6: 85610171-85610172 on Build GRCh38
rs992189884 Chr.6: 85610220-85610221 on Build GRCh38
rs369156334 Chr.6: 85610064-85610064 on Build GRCh38
rs539871262 Chr.6: 85610467-85610467 on Build GRCh38
rs781191390 Chr.6: 85610323-85610328 on Build GRCh38
rs752540115 Chr.6: 85610374-85610374 on Build GRCh38
rs1036567697 Chr.6: 85610404-85610406 on Build GRCh38

View Full Product Details