Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 113943215-113943716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC2
Entrez Gene ID
3066
Gene Name
histone deacetylase 2
Gene Aliases
HD2, RPD3, YAF1
UniGene
Hs.3352
Gene Chromosome Location
Chr.6: 113936156-113971195 on Build GRCh38
Transcripts
NM_001527.3 NR_033441.1 NR_073443.1 XM_011535788.1 XM_017010799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947911 c.1252G>A p.D418N Chr.6: 113943477-113943477 on Build GRCh38
1439942 c.1511C>T p.A504V Chr.6: 113943500-113943500 on Build GRCh38
3994562 c.1530T>G p.A510A Chr.6: 113943481-113943481 on Build GRCh38
392468 c.1375_1376insA p.T459fs*4 Chr.6: 113943353-113943354 on Build GRCh38
5558089 c.1652A>G p.K551R Chr.6: 113943359-113943359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898477215 Chr.6: 113943409-113943409 on Build GRCh38
rs759195320 Chr.6: 113943356-113943356 on Build GRCh38
rs145972581 Chr.6: 113943650-113943650 on Build GRCh38
rs936424975 Chr.6: 113943656-113943656 on Build GRCh38
rs55796700 Chr.6: 113943526-113943526 on Build GRCh38
rs772729099 Chr.6: 113943452-113943452 on Build GRCh38
rs139036465 Chr.6: 113943567-113943567 on Build GRCh38
rs748408899 Chr.6: 113943475-113943475 on Build GRCh38
rs995437670 Chr.6: 113943423-113943423 on Build GRCh38
rs142302027 Chr.6: 113943612-113943612 on Build GRCh38
rs746922822 Chr.6: 113943659-113943659 on Build GRCh38
rs191208195 Chr.6: 113943620-113943620 on Build GRCh38
rs778911906 Chr.6: 113943301-113943301 on Build GRCh38
rs991596514 Chr.6: 113943257-113943257 on Build GRCh38
rs756145331 Chr.6: 113943503-113943503 on Build GRCh38
rs374684204 Chr.6: 113943652-113943655 on Build GRCh38
rs942797919 Chr.6: 113943673-113943673 on Build GRCh38
rs774745631 Chr.6: 113943355-113943355 on Build GRCh38
rs781214400 Chr.6: 113943463-113943463 on Build GRCh38
rs369559085 Chr.6: 113943554-113943554 on Build GRCh38
rs748128191 Chr.6: 113943333-113943333 on Build GRCh38
rs1006151970 Chr.6: 113943441-113943441 on Build GRCh38
rs182415495 Chr.6: 113943249-113943249 on Build GRCh38
rs1039762787 Chr.6: 113943691-113943691 on Build GRCh38
rs562250416 Chr.6: 113943665-113943665 on Build GRCh38
rs557774086 Chr.6: 113943613-113943613 on Build GRCh38
rs556662064 Chr.6: 113943563-113943563 on Build GRCh38
rs774870177 Chr.6: 113943537-113943537 on Build GRCh38
rs914226839 Chr.6: 113943273-113943273 on Build GRCh38
rs1021263127 Chr.6: 113943570-113943570 on Build GRCh38
rs773644732 Chr.6: 113943337-113943337 on Build GRCh38
rs749857030 Chr.6: 113943347-113943347 on Build GRCh38
rs769362495 Chr.6: 113943379-113943379 on Build GRCh38
rs568166905 Chr.6: 113943359-113943359 on Build GRCh38
rs771271108 Chr.6: 113943353-113943353 on Build GRCh38
rs186701762 Chr.6: 113943317-113943317 on Build GRCh38
rs267600769 Chr.6: 113943504-113943504 on Build GRCh38
rs371501541 Chr.6: 113943434-113943434 on Build GRCh38
rs373338476 Chr.6: 113943243-113943243 on Build GRCh38
rs535146771 Chr.6: 113943532-113943532 on Build GRCh38
rs776578588 Chr.6: 113943293-113943293 on Build GRCh38
rs762338944 Chr.6: 113943453-113943453 on Build GRCh38
rs377062530 Chr.6: 113943389-113943389 on Build GRCh38
rs757316260 Chr.6: 113943385-113943385 on Build GRCh38
rs958726337 Chr.6: 113943533-113943533 on Build GRCh38
rs777886649 Chr.6: 113943522-113943522 on Build GRCh38
rs546369889 Chr.6: 113943367-113943367 on Build GRCh38
rs761489162 Chr.6: 113943294-113943294 on Build GRCh38
rs200492163 Chr.6: 113943274-113943274 on Build GRCh38
rs1053418705 Chr.6: 113943661-113943661 on Build GRCh38
rs913568308 Chr.6: 113943685-113943685 on Build GRCh38
rs936351653 Chr.6: 113943287-113943287 on Build GRCh38
rs373953184 Chr.6: 113943382-113943382 on Build GRCh38
rs781508197 Chr.6: 113943321-113943321 on Build GRCh38
rs747214999 Chr.6: 113943310-113943310 on Build GRCh38
rs775964258 Chr.6: 113943602-113943602 on Build GRCh38
rs912712923 Chr.6: 113943654-113943654 on Build GRCh38
rs775730268 Chr.6: 113943335-113943335 on Build GRCh38
rs958793647 Chr.6: 113943431-113943431 on Build GRCh38
rs769931575 Chr.6: 113943335-113943335 on Build GRCh38
rs968325459 Chr.6: 113943593-113943593 on Build GRCh38

View Full Product Details