Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 34811689-34812199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPSR1
Entrez Gene ID
387129
Gene Name
neuropeptide S receptor 1
Gene Aliases
ASRT2, GPR154, GPRA, NPSR, PGR14, VRR1
UniGene
Hs.652373
Gene Chromosome Location
Chr.7: 34658239-34878332 on Build GRCh38
Transcripts
NM_001300933.1 NM_001300934.1 NM_001300935.1 NM_207172.1 NM_207173.1 NR_015356.2

Gene Symbol
NPSR1-AS1
Entrez Gene ID
404744
Gene Name
NPSR1 antisense RNA 1
Gene Aliases
AAA1
UniGene
Hs.487951
Gene Chromosome Location
Chr.7: 34346512-34834331 on Build GRCh38
Transcripts
NM_001300933.1 NM_001300934.1 NM_001300935.1 NM_207172.1 NM_207173.1 NR_015356.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3638304 c.442C>T p.H148Y Chr.7: 34811827-34811827 on Build GRCh38
1089320 c.399C>T p.Y133Y Chr.7: 34811784-34811784 on Build GRCh38
228094 c.433G>A p.D145N Chr.7: 34811818-34811818 on Build GRCh38
3638315 c.459C>T p.P153P Chr.7: 34811844-34811844 on Build GRCh38
3638298 c.401C>T p.A134V Chr.7: 34811786-34811786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940913631 Chr.7: 34812133-34812133 on Build GRCh38
rs956057448 Chr.7: 34811764-34811764 on Build GRCh38
rs958604064 Chr.7: 34811940-34811940 on Build GRCh38
rs376690361 Chr.7: 34811797-34811797 on Build GRCh38
rs114775765 Chr.7: 34811785-34811785 on Build GRCh38
rs143552245 Chr.7: 34811784-34811784 on Build GRCh38
rs149649480 Chr.7: 34812139-34812139 on Build GRCh38
rs561539553 Chr.7: 34812094-34812094 on Build GRCh38
rs747437542 Chr.7: 34811873-34811873 on Build GRCh38
rs370033286 Chr.7: 34811901-34811901 on Build GRCh38
rs372055049 Chr.7: 34811759-34811759 on Build GRCh38
rs751345398 Chr.7: 34811935-34811935 on Build GRCh38
rs778278230 Chr.7: 34811872-34811872 on Build GRCh38
rs116733850 Chr.7: 34811835-34811835 on Build GRCh38
rs191425628 Chr.7: 34811903-34811903 on Build GRCh38
rs756751266 Chr.7: 34811843-34811843 on Build GRCh38
rs878989171 Chr.7: 34811856-34811856 on Build GRCh38
rs545032596 Chr.7: 34811964-34811964 on Build GRCh38
rs773833668 Chr.7: 34811971-34811971 on Build GRCh38
rs775440956 Chr.7: 34811820-34811820 on Build GRCh38
rs1046359228 Chr.7: 34811993-34811993 on Build GRCh38
rs767254511 Chr.7: 34811842-34811842 on Build GRCh38
rs1028773844 Chr.7: 34811850-34811850 on Build GRCh38
rs9655357 Chr.7: 34811781-34811781 on Build GRCh38
rs868156143 Chr.7: 34811857-34811857 on Build GRCh38
rs1044346013 Chr.7: 34811808-34811808 on Build GRCh38
rs200993440 Chr.7: 34811863-34811863 on Build GRCh38
rs751238663 Chr.7: 34811730-34811730 on Build GRCh38
rs930638272 Chr.7: 34812082-34812082 on Build GRCh38
rs1031823236 Chr.7: 34811966-34811966 on Build GRCh38
rs955837310 Chr.7: 34811989-34811989 on Build GRCh38
rs762796972 Chr.7: 34811907-34811910 on Build GRCh38
rs747202773 Chr.7: 34811789-34811789 on Build GRCh38
rs34052081 Chr.7: 34812171-34812171 on Build GRCh38
rs769943478 Chr.7: 34811895-34811895 on Build GRCh38
rs767009460 Chr.7: 34811908-34811908 on Build GRCh38
rs991304163 Chr.7: 34811951-34811951 on Build GRCh38
rs917101926 Chr.7: 34812011-34812011 on Build GRCh38
rs983501214 Chr.7: 34812132-34812132 on Build GRCh38
rs780533230 Chr.7: 34811739-34811739 on Build GRCh38
rs754690002 Chr.7: 34812029-34812029 on Build GRCh38
rs753949346 Chr.7: 34812156-34812156 on Build GRCh38
rs907896840 Chr.7: 34812038-34812038 on Build GRCh38
rs138267463 Chr.7: 34811950-34811950 on Build GRCh38
rs949396782 Chr.7: 34811978-34811978 on Build GRCh38
rs542725241 Chr.7: 34811899-34811899 on Build GRCh38
rs969913813 Chr.7: 34812027-34812027 on Build GRCh38
rs763858785 Chr.7: 34811830-34811830 on Build GRCh38
rs745447749 Chr.7: 34811742-34811742 on Build GRCh38
rs544431178 Chr.7: 34812080-34812080 on Build GRCh38
rs531256356 Chr.7: 34812075-34812075 on Build GRCh38
rs565402643 Chr.7: 34812046-34812046 on Build GRCh38
rs372990147 Chr.7: 34811796-34811796 on Build GRCh38
rs778062937 Chr.7: 34812061-34812061 on Build GRCh38
rs373169107 Chr.7: 34811868-34811868 on Build GRCh38
rs745898629 Chr.7: 34811891-34811891 on Build GRCh38
rs764007180 Chr.7: 34811922-34811922 on Build GRCh38
rs993045600 Chr.7: 34812001-34812001 on Build GRCh38
rs747871201 Chr.7: 34812123-34812123 on Build GRCh38
rs186939149 Chr.7: 34811885-34811885 on Build GRCh38
rs775600288 Chr.7: 34811897-34811897 on Build GRCh38
rs78508704 Chr.7: 34811770-34811770 on Build GRCh38
rs373527092 Chr.7: 34811870-34811870 on Build GRCh38
rs181364571 Chr.7: 34811844-34811844 on Build GRCh38
rs755652890 Chr.7: 34811854-34811854 on Build GRCh38
rs756851967 Chr.7: 34811836-34811836 on Build GRCh38
rs996385519 Chr.7: 34812174-34812174 on Build GRCh38
rs143746221 Chr.7: 34811823-34811823 on Build GRCh38
rs771829048 Chr.7: 34812135-34812135 on Build GRCh38
rs325465 Chr.7: 34811812-34811812 on Build GRCh38
rs138240804 Chr.7: 34811778-34811778 on Build GRCh38
rs756948450 Chr.7: 34811736-34811736 on Build GRCh38
rs759677549 Chr.7: 34811807-34811807 on Build GRCh38
rs376640561 Chr.7: 34811779-34811779 on Build GRCh38

View Full Product Details