Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158089476-158089969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIRREL
Entrez Gene ID
55243
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
NEPH1
UniGene
Hs.272234
Gene Chromosome Location
Chr.1: 157993273-158100262 on Build GRCh38
Transcripts
NM_001286349.1 NM_018240.6 XM_005245305.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463195 c.1219+2T>G p.? Chr.1: 158089630-158089630 on Build GRCh38
6201956 c.1096G>A p.A366T Chr.1: 158089553-158089553 on Build GRCh38
5727879 c.1260C>T p.P420P Chr.1: 158089806-158089806 on Build GRCh38
897632 c.1284G>T p.E428D Chr.1: 158089782-158089782 on Build GRCh38
4925259 c.1138G>A p.V380M Chr.1: 158089595-158089595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010826810 Chr.1: 158089940-158089940 on Build GRCh38
rs533402402 Chr.1: 158089936-158089936 on Build GRCh38
rs775683863 Chr.1: 158089654-158089654 on Build GRCh38
rs756185853 Chr.1: 158089761-158089761 on Build GRCh38
rs781087658 Chr.1: 158089618-158089618 on Build GRCh38
rs377074799 Chr.1: 158089800-158089800 on Build GRCh38
rs369810345 Chr.1: 158089552-158089552 on Build GRCh38
rs989786404 Chr.1: 158089646-158089646 on Build GRCh38
rs780340737 Chr.1: 158089777-158089777 on Build GRCh38
rs373920124 Chr.1: 158089523-158089523 on Build GRCh38
rs780358653 Chr.1: 158089840-158089840 on Build GRCh38
rs187001204 Chr.1: 158089839-158089839 on Build GRCh38
rs113267679 Chr.1: 158089906-158089906 on Build GRCh38
rs779988044 Chr.1: 158089609-158089609 on Build GRCh38
rs766001386 Chr.1: 158089826-158089826 on Build GRCh38
rs921973633 Chr.1: 158089804-158089804 on Build GRCh38
rs757563195 Chr.1: 158089624-158089624 on Build GRCh38
rs867471177 Chr.1: 158089579-158089579 on Build GRCh38
rs756831592 Chr.1: 158089507-158089507 on Build GRCh38
rs367592663 Chr.1: 158089831-158089831 on Build GRCh38
rs768392065 Chr.1: 158089536-158089536 on Build GRCh38
rs1007372875 Chr.1: 158089558-158089558 on Build GRCh38
rs139347921 Chr.1: 158089768-158089768 on Build GRCh38
rs958085852 Chr.1: 158089923-158089923 on Build GRCh38
rs745970000 Chr.1: 158089640-158089640 on Build GRCh38
rs761157828 Chr.1: 158089586-158089586 on Build GRCh38
rs1031178522 Chr.1: 158089605-158089605 on Build GRCh38
rs762018192 Chr.1: 158089667-158089667 on Build GRCh38
rs1011559746 Chr.1: 158089924-158089924 on Build GRCh38
rs767499528 Chr.1: 158089830-158089830 on Build GRCh38
rs979827102 Chr.1: 158089704-158089704 on Build GRCh38
rs774545635 Chr.1: 158089555-158089555 on Build GRCh38
rs747880117 Chr.1: 158089620-158089620 on Build GRCh38
rs764794274 Chr.1: 158089598-158089598 on Build GRCh38
rs770014051 Chr.1: 158089813-158089813 on Build GRCh38
rs761100201 Chr.1: 158089689-158089689 on Build GRCh38
rs762885869 Chr.1: 158089587-158089587 on Build GRCh38
rs754964453 Chr.1: 158089699-158089699 on Build GRCh38
rs185656135 Chr.1: 158089560-158089560 on Build GRCh38
rs779214667 Chr.1: 158089731-158089731 on Build GRCh38
rs6683714 Chr.1: 158089627-158089627 on Build GRCh38
rs772243443 Chr.1: 158089643-158089643 on Build GRCh38
rs749954360 Chr.1: 158089604-158089604 on Build GRCh38
rs372659797 Chr.1: 158089580-158089580 on Build GRCh38
rs748573118 Chr.1: 158089513-158089513 on Build GRCh38
rs370686077 Chr.1: 158089760-158089760 on Build GRCh38
rs567539621 Chr.1: 158089505-158089505 on Build GRCh38
rs761586318 Chr.1: 158089592-158089592 on Build GRCh38
rs762641862 Chr.1: 158089822-158089822 on Build GRCh38
rs1020483916 Chr.1: 158089944-158089944 on Build GRCh38
rs778537144 Chr.1: 158089509-158089509 on Build GRCh38
rs772707999 Chr.1: 158089567-158089567 on Build GRCh38
rs541325906 Chr.1: 158089655-158089655 on Build GRCh38
rs776945930 Chr.1: 158089796-158089796 on Build GRCh38
rs905552425 Chr.1: 158089870-158089870 on Build GRCh38
rs567911220 Chr.1: 158089665-158089665 on Build GRCh38
rs766434496 Chr.1: 158089827-158089827 on Build GRCh38
rs372759941 Chr.1: 158089676-158089676 on Build GRCh38
rs372126342 Chr.1: 158089835-158089835 on Build GRCh38
rs889651469 Chr.1: 158089498-158089498 on Build GRCh38
rs755802847 Chr.1: 158089712-158089712 on Build GRCh38
rs901272490 Chr.1: 158089846-158089846 on Build GRCh38
rs751044495 Chr.1: 158089693-158089693 on Build GRCh38
rs752538899 Chr.1: 158089711-158089711 on Build GRCh38
rs543383113 Chr.1: 158089767-158089767 on Build GRCh38
rs931206602 Chr.1: 158089817-158089817 on Build GRCh38
rs779900251 Chr.1: 158089521-158089521 on Build GRCh38
rs145693910 Chr.1: 158089815-158089815 on Build GRCh38
rs767348850 Chr.1: 158089703-158089703 on Build GRCh38
rs769212897 Chr.1: 158089794-158089794 on Build GRCh38
rs368503496 Chr.1: 158089856-158089856 on Build GRCh38
rs369737138 Chr.1: 158089862-158089862 on Build GRCh38
rs182854509 Chr.1: 158089615-158089615 on Build GRCh38
rs763848085 Chr.1: 158089832-158089832 on Build GRCh38
rs201805473 Chr.1: 158089537-158089537 on Build GRCh38
rs758430194 Chr.1: 158089519-158089519 on Build GRCh38
rs373086254 Chr.1: 158089851-158089851 on Build GRCh38
rs112477747 Chr.1: 158089671-158089671 on Build GRCh38
rs374977006 Chr.1: 158089828-158089828 on Build GRCh38
rs776604086 Chr.1: 158089902-158089902 on Build GRCh38
rs765913419 Chr.1: 158089680-158089680 on Build GRCh38
rs369327721 Chr.1: 158089664-158089664 on Build GRCh38
rs146308217 Chr.1: 158089591-158089591 on Build GRCh38
rs375141355 Chr.1: 158089790-158089790 on Build GRCh38
rs576363418 Chr.1: 158089677-158089677 on Build GRCh38
rs764956502 Chr.1: 158089673-158089673 on Build GRCh38

View Full Product Details