Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161525528-161526019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA6
Entrez Gene ID
3310
Gene Name
heat shock protein family A (Hsp70) member 6
Gene Aliases
HSP70B'
UniGene
Hs.654614
Gene Chromosome Location
Chr.1: 161524540-161526897 on Build GRCh38
Transcripts
NM_002155.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
395784 c.1315G>T p.G439W Chr.1: 161525973-161525973 on Build GRCh38
6037221 c.1006G>T p.V336F Chr.1: 161525664-161525664 on Build GRCh38
4599798 c.1078G>T p.E360* Chr.1: 161525736-161525736 on Build GRCh38
7138981 c.1320C>G p.V440V Chr.1: 161525978-161525978 on Build GRCh38
4599417 c.1123G>T p.A375S Chr.1: 161525781-161525781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760299991 Chr.1: 161525610-161525610 on Build GRCh38
rs150927976 Chr.1: 161525909-161525909 on Build GRCh38
rs559953007 Chr.1: 161525853-161525853 on Build GRCh38
rs781208419 Chr.1: 161525938-161525938 on Build GRCh38
rs777776533 Chr.1: 161525949-161525949 on Build GRCh38
rs375473426 Chr.1: 161525925-161525925 on Build GRCh38
rs770559320 Chr.1: 161525859-161525859 on Build GRCh38
rs747731136 Chr.1: 161525756-161525756 on Build GRCh38
rs747835514 Chr.1: 161525944-161525944 on Build GRCh38
rs892634899 Chr.1: 161525699-161525699 on Build GRCh38
rs745780265 Chr.1: 161525665-161525665 on Build GRCh38
rs142859332 Chr.1: 161525673-161525673 on Build GRCh38
rs772349234 Chr.1: 161525770-161525771 on Build GRCh38
rs948651316 Chr.1: 161525623-161525623 on Build GRCh38
rs781731364 Chr.1: 161525571-161525571 on Build GRCh38
rs772930470 Chr.1: 161525850-161525850 on Build GRCh38
rs764355185 Chr.1: 161525785-161525785 on Build GRCh38
rs41297720 Chr.1: 161525945-161525945 on Build GRCh38
rs746948130 Chr.1: 161525811-161525811 on Build GRCh38
rs778837292 Chr.1: 161525712-161525712 on Build GRCh38
rs756559306 Chr.1: 161525698-161525698 on Build GRCh38
rs417707 Chr.1: 161525664-161525664 on Build GRCh38
rs757559207 Chr.1: 161525708-161525708 on Build GRCh38
rs908331997 Chr.1: 161525846-161525846 on Build GRCh38
rs375948126 Chr.1: 161525990-161525990 on Build GRCh38
rs915726576 Chr.1: 161525924-161525924 on Build GRCh38
rs776820851 Chr.1: 161525902-161525902 on Build GRCh38
rs768447503 Chr.1: 161525749-161525749 on Build GRCh38
rs750651630 Chr.1: 161525912-161525912 on Build GRCh38
rs777983283 Chr.1: 161525703-161525703 on Build GRCh38
rs748365478 Chr.1: 161525574-161525574 on Build GRCh38
rs55898103 Chr.1: 161525582-161525582 on Build GRCh38
rs748002346 Chr.1: 161525829-161525829 on Build GRCh38
rs781671425 Chr.1: 161525644-161525644 on Build GRCh38
rs11577457 Chr.1: 161525976-161525976 on Build GRCh38
rs755563815 Chr.1: 161525640-161525640 on Build GRCh38
rs369670009 Chr.1: 161525922-161525922 on Build GRCh38
rs749357670 Chr.1: 161525592-161525592 on Build GRCh38
rs765529044 Chr.1: 161525792-161525792 on Build GRCh38
rs140320557 Chr.1: 161525967-161525967 on Build GRCh38
rs773628292 Chr.1: 161525974-161525974 on Build GRCh38
rs143204924 Chr.1: 161525678-161525678 on Build GRCh38
rs9659608 Chr.1: 161525781-161525781 on Build GRCh38
rs775324314 Chr.1: 161525603-161525603 on Build GRCh38
rs867989346 Chr.1: 161525635-161525635 on Build GRCh38
rs780461359 Chr.1: 161525563-161525563 on Build GRCh38
rs373877131 Chr.1: 161525936-161525936 on Build GRCh38
rs776559394 Chr.1: 161525753-161525753 on Build GRCh38
rs745403310 Chr.1: 161525961-161525961 on Build GRCh38
rs375064286 Chr.1: 161525790-161525790 on Build GRCh38
rs1042924129 Chr.1: 161525966-161525966 on Build GRCh38
rs751586268 Chr.1: 161525988-161525988 on Build GRCh38
rs548751388 Chr.1: 161525674-161525674 on Build GRCh38
rs768996694 Chr.1: 161525963-161525963 on Build GRCh38
rs753061469 Chr.1: 161525694-161525694 on Build GRCh38
rs779902210 Chr.1: 161525805-161525805 on Build GRCh38
rs759307548 Chr.1: 161525597-161525597 on Build GRCh38
rs758911776 Chr.1: 161525559-161525559 on Build GRCh38
rs758490304 Chr.1: 161525799-161525799 on Build GRCh38
rs760084677 Chr.1: 161525688-161525688 on Build GRCh38
rs375914735 Chr.1: 161525663-161525663 on Build GRCh38
rs778163585 Chr.1: 161525659-161525659 on Build GRCh38
rs774167328 Chr.1: 161525595-161525595 on Build GRCh38
rs766436986 Chr.1: 161525933-161525933 on Build GRCh38
rs754954136 Chr.1: 161525813-161525813 on Build GRCh38
rs769278723 Chr.1: 161525848-161525848 on Build GRCh38
rs200985857 Chr.1: 161525730-161525730 on Build GRCh38
rs754828859 Chr.1: 161525935-161525935 on Build GRCh38
rs772554235 Chr.1: 161525759-161525759 on Build GRCh38
rs763776911 Chr.1: 161525614-161525614 on Build GRCh38
rs368785490 Chr.1: 161525806-161525806 on Build GRCh38
rs17853454 Chr.1: 161525687-161525687 on Build GRCh38
rs770193043 Chr.1: 161525970-161525970 on Build GRCh38
rs768165808 Chr.1: 161525689-161525689 on Build GRCh38
rs745644751 Chr.1: 161525761-161525761 on Build GRCh38
rs758689258 Chr.1: 161525930-161525930 on Build GRCh38
rs778537530 Chr.1: 161525960-161525960 on Build GRCh38
rs752015144 Chr.1: 161525639-161525639 on Build GRCh38
rs200111192 Chr.1: 161525959-161525959 on Build GRCh38
rs751196854 Chr.1: 161525650-161525650 on Build GRCh38
rs749331338 Chr.1: 161525661-161525661 on Build GRCh38
rs753149053 Chr.1: 161525655-161525655 on Build GRCh38
rs775538197 Chr.1: 161525838-161525840 on Build GRCh38
rs145456895 Chr.1: 161525992-161525992 on Build GRCh38
rs993097933 Chr.1: 161525870-161525870 on Build GRCh38
rs144853604 Chr.1: 161525707-161525707 on Build GRCh38
rs997085398 Chr.1: 161525737-161525737 on Build GRCh38
rs12119062 Chr.1: 161525677-161525677 on Build GRCh38
rs141771248 Chr.1: 161525679-161525679 on Build GRCh38
rs1026665677 Chr.1: 161525763-161525763 on Build GRCh38
rs780817032 Chr.1: 161525828-161525828 on Build GRCh38
rs766617427 Chr.1: 161525982-161525982 on Build GRCh38
rs371491425 Chr.1: 161525600-161525600 on Build GRCh38
rs774061722 Chr.1: 161525860-161525860 on Build GRCh38
rs56366425 Chr.1: 161525998-161525998 on Build GRCh38
rs768078077 Chr.1: 161525778-161525778 on Build GRCh38
rs754280500 Chr.1: 161525788-161525788 on Build GRCh38
rs772047847 Chr.1: 161525666-161525666 on Build GRCh38
rs952324501 Chr.1: 161525795-161525795 on Build GRCh38
rs755314257 Chr.1: 161525570-161525570 on Build GRCh38
rs778744518 Chr.1: 161525674-161525675 on Build GRCh38
rs138329594 Chr.1: 161525680-161525680 on Build GRCh38
rs748933785 Chr.1: 161525957-161525957 on Build GRCh38
rs41297718 Chr.1: 161525548-161525548 on Build GRCh38
rs776145445 Chr.1: 161525780-161525780 on Build GRCh38
rs529246286 Chr.1: 161525586-161525586 on Build GRCh38
rs775234265 Chr.1: 161525671-161525671 on Build GRCh38
rs371404994 Chr.1: 161525907-161525907 on Build GRCh38
rs750458058 Chr.1: 161525796-161525796 on Build GRCh38
rs151297860 Chr.1: 161525577-161525577 on Build GRCh38
rs761255637 Chr.1: 161525630-161525630 on Build GRCh38
rs766855467 Chr.1: 161525631-161525631 on Build GRCh38
rs762570646 Chr.1: 161525777-161525777 on Build GRCh38
rs761243662 Chr.1: 161525897-161525897 on Build GRCh38
rs148797832 Chr.1: 161525557-161525557 on Build GRCh38
rs746752089 Chr.1: 161525741-161525741 on Build GRCh38
rs747352949 Chr.1: 161525566-161525566 on Build GRCh38
rs779993877 Chr.1: 161525738-161525738 on Build GRCh38
rs368481392 Chr.1: 161525843-161525843 on Build GRCh38
rs753352679 Chr.1: 161525624-161525624 on Build GRCh38
rs770851366 Chr.1: 161525594-161525594 on Build GRCh38
rs756360777 Chr.1: 161525656-161525656 on Build GRCh38
rs528780088 Chr.1: 161525901-161525901 on Build GRCh38

View Full Product Details