Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171636128-171636636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOC
Entrez Gene ID
4653
Gene Name
myocilin
Gene Aliases
GLC1A, GPOA, JOAG, JOAG1, TIGR
UniGene
Hs.436037
Gene Chromosome Location
Chr.1: 171635417-171652633 on Build GRCh38
Transcripts
NM_000261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6720752 c.905A>G p.D302G Chr.1: 171636535-171636535 on Build GRCh38
5795866 c.1109C>T p.P370L Chr.1: 171636331-171636331 on Build GRCh38
4186462 c.1082C>G p.P361R Chr.1: 171636358-171636358 on Build GRCh38
1662977 c.1130C>T p.T377M Chr.1: 171636310-171636310 on Build GRCh38
5844496 c.1099G>A p.G367R Chr.1: 171636341-171636341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751319338 Chr.1: 171636563-171636563 on Build GRCh38
rs761755712 Chr.1: 171636573-171636573 on Build GRCh38
rs756397695 Chr.1: 171636525-171636525 on Build GRCh38
rs780460955 Chr.1: 171636527-171636527 on Build GRCh38
rs963929660 Chr.1: 171636362-171636362 on Build GRCh38
rs201573718 Chr.1: 171636175-171636175 on Build GRCh38
rs1056051290 Chr.1: 171636229-171636229 on Build GRCh38
rs998968146 Chr.1: 171636261-171636261 on Build GRCh38
rs150755204 Chr.1: 171636312-171636312 on Build GRCh38
rs762876843 Chr.1: 171636452-171636453 on Build GRCh38
rs764942545 Chr.1: 171636416-171636416 on Build GRCh38
rs867417217 Chr.1: 171636582-171636582 on Build GRCh38
rs767627671 Chr.1: 171636575-171636575 on Build GRCh38
rs776956998 Chr.1: 171636368-171636368 on Build GRCh38
rs61730974 Chr.1: 171636399-171636399 on Build GRCh38
rs780876895 Chr.1: 171636188-171636188 on Build GRCh38
rs369169139 Chr.1: 171636561-171636561 on Build GRCh38
rs769011297 Chr.1: 171636453-171636453 on Build GRCh38
rs745961303 Chr.1: 171636513-171636513 on Build GRCh38
rs748201066 Chr.1: 171636231-171636231 on Build GRCh38
rs369157032 Chr.1: 171636257-171636257 on Build GRCh38
rs759863280 Chr.1: 171636603-171636603 on Build GRCh38
rs752488318 Chr.1: 171636488-171636488 on Build GRCh38
rs779176698 Chr.1: 171636540-171636540 on Build GRCh38
rs61730976 Chr.1: 171636465-171636465 on Build GRCh38
rs1003013455 Chr.1: 171636332-171636332 on Build GRCh38
rs536180276 Chr.1: 171636213-171636213 on Build GRCh38
rs760271913 Chr.1: 171636388-171636388 on Build GRCh38
rs772087054 Chr.1: 171636234-171636234 on Build GRCh38
rs886045565 Chr.1: 171636569-171636569 on Build GRCh38
rs760065904 Chr.1: 171636168-171636168 on Build GRCh38
rs765926032 Chr.1: 171636389-171636389 on Build GRCh38
rs144469479 Chr.1: 171636533-171636533 on Build GRCh38
rs772580255 Chr.1: 171636550-171636550 on Build GRCh38
rs534806873 Chr.1: 171636420-171636420 on Build GRCh38
rs768574920 Chr.1: 171636203-171636203 on Build GRCh38
rs770236294 Chr.1: 171636371-171636371 on Build GRCh38
rs749960559 Chr.1: 171636280-171636280 on Build GRCh38
rs757407270 Chr.1: 171636438-171636438 on Build GRCh38
rs121909193 Chr.1: 171636349-171636349 on Build GRCh38
rs202003673 Chr.1: 171636387-171636387 on Build GRCh38
rs374805593 Chr.1: 171636501-171636501 on Build GRCh38
rs145229172 Chr.1: 171636580-171636580 on Build GRCh38
rs888606739 Chr.1: 171636169-171636169 on Build GRCh38
rs754237376 Chr.1: 171636161-171636161 on Build GRCh38
rs766899237 Chr.1: 171636264-171636264 on Build GRCh38
rs144386835 Chr.1: 171636342-171636342 on Build GRCh38
rs547608702 Chr.1: 171636447-171636447 on Build GRCh38
rs774268708 Chr.1: 171636596-171636596 on Build GRCh38
rs139804296 Chr.1: 171636189-171636189 on Build GRCh38
rs566289099 Chr.1: 171636310-171636310 on Build GRCh38
rs750946621 Chr.1: 171636426-171636426 on Build GRCh38
rs778699435 Chr.1: 171636318-171636318 on Build GRCh38
rs772565902 Chr.1: 171636599-171636599 on Build GRCh38
rs74315336 Chr.1: 171636173-171636173 on Build GRCh38
rs907466277 Chr.1: 171636549-171636549 on Build GRCh38
rs769406646 Chr.1: 171636556-171636556 on Build GRCh38
rs755159007 Chr.1: 171636587-171636587 on Build GRCh38
rs779654151 Chr.1: 171636193-171636193 on Build GRCh38
rs150438494 Chr.1: 171636182-171636182 on Build GRCh38
rs74315334 Chr.1: 171636341-171636341 on Build GRCh38
rs771057339 Chr.1: 171636350-171636350 on Build GRCh38
rs763577837 Chr.1: 171636258-171636258 on Build GRCh38
rs754051534 Chr.1: 171636586-171636586 on Build GRCh38
rs1041675997 Chr.1: 171636543-171636543 on Build GRCh38
rs878854408 Chr.1: 171636329-171636329 on Build GRCh38
rs74315335 Chr.1: 171636430-171636430 on Build GRCh38
rs749735333 Chr.1: 171636451-171636451 on Build GRCh38
rs61730975 Chr.1: 171636252-171636252 on Build GRCh38
rs776030983 Chr.1: 171636375-171636375 on Build GRCh38
rs376895063 Chr.1: 171636502-171636502 on Build GRCh38
rs146606638 Chr.1: 171636585-171636585 on Build GRCh38
rs909669034 Chr.1: 171636436-171636436 on Build GRCh38
rs747174989 Chr.1: 171636343-171636343 on Build GRCh38
rs771200816 Chr.1: 171636250-171636250 on Build GRCh38
rs61745146 Chr.1: 171636386-171636386 on Build GRCh38
rs1033533679 Chr.1: 171636287-171636287 on Build GRCh38
rs942833087 Chr.1: 171636496-171636496 on Build GRCh38
rs757174636 Chr.1: 171636504-171636504 on Build GRCh38
rs28936694 Chr.1: 171636244-171636244 on Build GRCh38
rs149881467 Chr.1: 171636330-171636330 on Build GRCh38
rs766610033 Chr.1: 171636153-171636153 on Build GRCh38
rs1054929295 Chr.1: 171636498-171636498 on Build GRCh38
rs778293128 Chr.1: 171636606-171636606 on Build GRCh38
rs781356204 Chr.1: 171636510-171636510 on Build GRCh38
rs121909194 Chr.1: 171636302-171636302 on Build GRCh38
rs762244947 Chr.1: 171636459-171636459 on Build GRCh38
rs745360975 Chr.1: 171636616-171636616 on Build GRCh38
rs775982158 Chr.1: 171636448-171636448 on Build GRCh38
rs749164354 Chr.1: 171636201-171636201 on Build GRCh38
rs748516657 Chr.1: 171636590-171636590 on Build GRCh38
rs181923440 Chr.1: 171636576-171636576 on Build GRCh38
rs139122673 Chr.1: 171636562-171636562 on Build GRCh38
rs1050657565 Chr.1: 171636156-171636156 on Build GRCh38
rs137853277 Chr.1: 171636382-171636382 on Build GRCh38
rs748621461 Chr.1: 171636542-171636542 on Build GRCh38
rs145934417 Chr.1: 171636553-171636553 on Build GRCh38
rs755824609 Chr.1: 171636190-171636190 on Build GRCh38
rs148433908 Chr.1: 171636534-171636534 on Build GRCh38
rs146391864 Chr.1: 171636455-171636455 on Build GRCh38
rs137853276 Chr.1: 171636449-171636449 on Build GRCh38
rs755706856 Chr.1: 171636298-171636298 on Build GRCh38
rs765809047 Chr.1: 171636255-171636255 on Build GRCh38
rs143474164 Chr.1: 171636559-171636559 on Build GRCh38
rs759128833 Chr.1: 171636479-171636479 on Build GRCh38
rs201206951 Chr.1: 171636439-171636439 on Build GRCh38
rs56314834 Chr.1: 171636247-171636247 on Build GRCh38
rs758257704 Chr.1: 171636425-171636425 on Build GRCh38
rs147122394 Chr.1: 171636466-171636466 on Build GRCh38
rs746048852 Chr.1: 171636442-171636442 on Build GRCh38
rs751113505 Chr.1: 171636176-171636176 on Build GRCh38
rs746216832 Chr.1: 171636588-171636588 on Build GRCh38
rs140112372 Chr.1: 171636309-171636309 on Build GRCh38
rs989306028 Chr.1: 171636321-171636321 on Build GRCh38
rs74315330 Chr.1: 171636331-171636331 on Build GRCh38
rs866938399 Chr.1: 171636369-171636369 on Build GRCh38
rs74315329 Chr.1: 171636338-171636338 on Build GRCh38
rs759114694 Chr.1: 171636554-171636554 on Build GRCh38
rs766321647 Chr.1: 171636484-171636484 on Build GRCh38

View Full Product Details