Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96231674-96232189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UQCRB
Entrez Gene ID
7381
Gene Name
ubiquinol-cytochrome c reductase binding protein
Gene Aliases
MC3DN3, QCR7, QP-C, QPC, UQBC, UQBP, UQCR6, UQPC
UniGene
Hs.131255
Gene Chromosome Location
Chr.8: 96226676-96235634 on Build GRCh38
Transcripts
NM_001199975.2 NM_001254752.1 NM_006294.4 NR_045639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1102760 c.136G>T p.E46* Chr.8: 96231896-96231896 on Build GRCh38
6484662 c.172G>T p.D58Y Chr.8: 96231860-96231860 on Build GRCh38
1102759 c.223A>C p.I75L Chr.8: 96231809-96231809 on Build GRCh38
6362386 c.179T>G p.M60R Chr.8: 96231853-96231853 on Build GRCh38
3835185 c.146G>A p.R49K Chr.8: 96231886-96231886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759047335 Chr.8: 96231899-96231899 on Build GRCh38
rs946282587 Chr.8: 96231715-96231715 on Build GRCh38
rs765687499 Chr.8: 96231983-96231983 on Build GRCh38
rs750762015 Chr.8: 96231985-96231985 on Build GRCh38
rs368089982 Chr.8: 96232138-96232138 on Build GRCh38
rs756562701 Chr.8: 96231814-96231814 on Build GRCh38
rs544653898 Chr.8: 96232017-96232017 on Build GRCh38
rs141279562 Chr.8: 96232067-96232067 on Build GRCh38
rs1027992971 Chr.8: 96232070-96232070 on Build GRCh38
rs375663407 Chr.8: 96231756-96231756 on Build GRCh38
rs776521344 Chr.8: 96231777-96231777 on Build GRCh38
rs755322780 Chr.8: 96231988-96231988 on Build GRCh38
rs756035969 Chr.8: 96231991-96231991 on Build GRCh38
rs373917947 Chr.8: 96231916-96231916 on Build GRCh38
rs367861933 Chr.8: 96231976-96231976 on Build GRCh38
rs117599689 Chr.8: 96231719-96231719 on Build GRCh38
rs765910006 Chr.8: 96231808-96231808 on Build GRCh38
rs780172310 Chr.8: 96231925-96231925 on Build GRCh38
rs564927481 Chr.8: 96232050-96232050 on Build GRCh38
rs536858169 Chr.8: 96231740-96231740 on Build GRCh38
rs201818465 Chr.8: 96231873-96231873 on Build GRCh38
rs764178627 Chr.8: 96231772-96231772 on Build GRCh38
rs768870614 Chr.8: 96231931-96231931 on Build GRCh38
rs765967155 Chr.8: 96231890-96231890 on Build GRCh38
rs73269963 Chr.8: 96231969-96231969 on Build GRCh38
rs577844002 Chr.8: 96231920-96231920 on Build GRCh38
rs142060471 Chr.8: 96231989-96231990 on Build GRCh38
rs747676206 Chr.8: 96231941-96231942 on Build GRCh38
rs1048820211 Chr.8: 96232049-96232049 on Build GRCh38
rs376891713 Chr.8: 96231944-96231944 on Build GRCh38
rs769661824 Chr.8: 96231947-96231947 on Build GRCh38
rs111646807 Chr.8: 96231942-96231942 on Build GRCh38
rs762685731 Chr.8: 96231951-96231951 on Build GRCh38
rs372659618 Chr.8: 96231732-96231732 on Build GRCh38
rs757827287 Chr.8: 96231914-96231914 on Build GRCh38
rs781198686 Chr.8: 96231990-96231990 on Build GRCh38
rs1044383098 Chr.8: 96232031-96232031 on Build GRCh38
rs147795609 Chr.8: 96232058-96232058 on Build GRCh38
rs781393195 Chr.8: 96231989-96231989 on Build GRCh38
rs754413288 Chr.8: 96231916-96231917 on Build GRCh38
rs750105381 Chr.8: 96231793-96231793 on Build GRCh38
rs769702943 Chr.8: 96231869-96231869 on Build GRCh38
rs577597817 Chr.8: 96231996-96231996 on Build GRCh38
rs777625043 Chr.8: 96231990-96231990 on Build GRCh38
rs370012842 Chr.8: 96231767-96231767 on Build GRCh38
rs947813674 Chr.8: 96232041-96232041 on Build GRCh38
rs1804263 Chr.8: 96231826-96231826 on Build GRCh38
rs11558865 Chr.8: 96231837-96231837 on Build GRCh38
rs773885026 Chr.8: 96231962-96231962 on Build GRCh38
rs762638352 Chr.8: 96231885-96231885 on Build GRCh38
rs754329213 Chr.8: 96231908-96231908 on Build GRCh38
rs530276576 Chr.8: 96232118-96232118 on Build GRCh38
rs371247478 Chr.8: 96231949-96231949 on Build GRCh38
rs1008430383 Chr.8: 96232131-96232131 on Build GRCh38
rs1014823761 Chr.8: 96231909-96231909 on Build GRCh38
rs779304514 Chr.8: 96231915-96231915 on Build GRCh38
rs761734666 Chr.8: 96231782-96231782 on Build GRCh38
rs901749231 Chr.8: 96232143-96232143 on Build GRCh38
rs752243166 Chr.8: 96231970-96231970 on Build GRCh38
rs761646712 Chr.8: 96231861-96231861 on Build GRCh38
rs189786465 Chr.8: 96232111-96232111 on Build GRCh38
rs531483355 Chr.8: 96232012-96232012 on Build GRCh38
rs201177719 Chr.8: 96231955-96231955 on Build GRCh38
rs751036864 Chr.8: 96231896-96231896 on Build GRCh38
rs766565491 Chr.8: 96231986-96231986 on Build GRCh38
rs778282692 Chr.8: 96231823-96231823 on Build GRCh38
rs775497059 Chr.8: 96231759-96231759 on Build GRCh38
rs764523435 Chr.8: 96231901-96231901 on Build GRCh38
rs751073751 Chr.8: 96231809-96231809 on Build GRCh38
rs145974195 Chr.8: 96231854-96231854 on Build GRCh38
rs776718899 Chr.8: 96231860-96231860 on Build GRCh38
rs771101419 Chr.8: 96231736-96231736 on Build GRCh38
rs139851271 Chr.8: 96231932-96231932 on Build GRCh38
rs145828292 Chr.8: 96231783-96231783 on Build GRCh38
rs139283183 Chr.8: 96231832-96231832 on Build GRCh38
rs771974856 Chr.8: 96231847-96231847 on Build GRCh38
rs751822953 Chr.8: 96231987-96231987 on Build GRCh38
rs759239500 Chr.8: 96231964-96231964 on Build GRCh38
rs778519031 Chr.8: 96231939-96231940 on Build GRCh38
rs116946612 Chr.8: 96232011-96232011 on Build GRCh38
rs757627180 Chr.8: 96231834-96231834 on Build GRCh38
rs758600342 Chr.8: 96231922-96231922 on Build GRCh38
rs1056370228 Chr.8: 96231807-96231807 on Build GRCh38
rs779199448 Chr.8: 96231836-96231836 on Build GRCh38
rs758045138 Chr.8: 96231806-96231806 on Build GRCh38
rs567823218 Chr.8: 96231739-96231739 on Build GRCh38
rs755471110 Chr.8: 96231919-96231933 on Build GRCh38
rs772220316 Chr.8: 96231846-96231846 on Build GRCh38
rs747018908 Chr.8: 96231930-96231930 on Build GRCh38

View Full Product Details