Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98045166-98045670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL30
Entrez Gene ID
6156
Gene Name
ribosomal protein L30
Gene Aliases
L30
UniGene
Hs.400295
Gene Chromosome Location
Chr.8: 98041710-98045590 on Build GRCh38
Transcripts
NM_000989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769222 c.10G>A p.A4T Chr.8: 98045358-98045358 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922595539 Chr.8: 98045524-98045524 on Build GRCh38
rs777764208 Chr.8: 98045399-98045399 on Build GRCh38
rs919597776 Chr.8: 98045290-98045290 on Build GRCh38
rs746892759 Chr.8: 98045616-98045616 on Build GRCh38
rs776261510 Chr.8: 98045247-98045247 on Build GRCh38
rs138167986 Chr.8: 98045337-98045337 on Build GRCh38
rs74971161 Chr.8: 98045187-98045187 on Build GRCh38
rs566163443 Chr.8: 98045421-98045421 on Build GRCh38
rs980150813 Chr.8: 98045493-98045493 on Build GRCh38
rs954771989 Chr.8: 98045401-98045401 on Build GRCh38
rs913065748 Chr.8: 98045556-98045556 on Build GRCh38
rs812 Chr.8: 98045298-98045298 on Build GRCh38
rs371636770 Chr.8: 98045316-98045316 on Build GRCh38
rs115595912 Chr.8: 98045403-98045403 on Build GRCh38
rs147738892 Chr.8: 98045216-98045216 on Build GRCh38
rs904515958 Chr.8: 98045390-98045390 on Build GRCh38
rs142660222 Chr.8: 98045554-98045554 on Build GRCh38
rs77033333 Chr.8: 98045487-98045487 on Build GRCh38
rs1049965855 Chr.8: 98045224-98045224 on Build GRCh38
rs374627007 Chr.8: 98045617-98045617 on Build GRCh38
rs996127283 Chr.8: 98045594-98045594 on Build GRCh38
rs1044073510 Chr.8: 98045293-98045293 on Build GRCh38
rs920809115 Chr.8: 98045469-98045469 on Build GRCh38
rs763213200 Chr.8: 98045357-98045357 on Build GRCh38
rs369869528 Chr.8: 98045410-98045410 on Build GRCh38
rs551163149 Chr.8: 98045330-98045330 on Build GRCh38
rs1029527722 Chr.8: 98045211-98045211 on Build GRCh38
rs575910020 Chr.8: 98045629-98045629 on Build GRCh38
rs1011048653 Chr.8: 98045268-98045268 on Build GRCh38
rs932604026 Chr.8: 98045534-98045534 on Build GRCh38
rs10591 Chr.8: 98045379-98045379 on Build GRCh38
rs569133755 Chr.8: 98045465-98045465 on Build GRCh38
rs951900660 Chr.8: 98045433-98045433 on Build GRCh38
rs573322519 Chr.8: 98045615-98045615 on Build GRCh38
rs370054567 Chr.8: 98045259-98045259 on Build GRCh38
rs939101675 Chr.8: 98045576-98045576 on Build GRCh38
rs553395463 Chr.8: 98045613-98045613 on Build GRCh38
rs752684032 Chr.8: 98045392-98045392 on Build GRCh38
rs755965282 Chr.8: 98045394-98045394 on Build GRCh38
rs751552137 Chr.8: 98045369-98045369 on Build GRCh38
rs904400000 Chr.8: 98045591-98045591 on Build GRCh38
rs1050359933 Chr.8: 98045598-98045598 on Build GRCh38
rs745658879 Chr.8: 98045409-98045410 on Build GRCh38
rs962210891 Chr.8: 98045458-98045458 on Build GRCh38
rs962242644 Chr.8: 98045255-98045255 on Build GRCh38
rs11539268 Chr.8: 98045431-98045431 on Build GRCh38
rs938398654 Chr.8: 98045517-98045517 on Build GRCh38
rs1027839804 Chr.8: 98045643-98045643 on Build GRCh38
rs112294464 Chr.8: 98045546-98045546 on Build GRCh38
rs961791524 Chr.8: 98045516-98045516 on Build GRCh38
rs757006806 Chr.8: 98045408-98045408 on Build GRCh38
rs112858018 Chr.8: 98045548-98045548 on Build GRCh38
rs1042178165 Chr.8: 98045281-98045281 on Build GRCh38
rs769883554 Chr.8: 98045649-98045649 on Build GRCh38
rs548909700 Chr.8: 98045441-98045441 on Build GRCh38
rs890488667 Chr.8: 98045605-98045605 on Build GRCh38
rs765657504 Chr.8: 98045333-98045333 on Build GRCh38
rs1041470349 Chr.8: 98045199-98045199 on Build GRCh38
rs985722224 Chr.8: 98045545-98045545 on Build GRCh38
rs555743648 Chr.8: 98045622-98045622 on Build GRCh38
rs771897951 Chr.8: 98045526-98045526 on Build GRCh38
rs766416798 Chr.8: 98045358-98045358 on Build GRCh38
rs894818835 Chr.8: 98045611-98045612 on Build GRCh38
rs932933930 Chr.8: 98045220-98045220 on Build GRCh38
rs113903310 Chr.8: 98045332-98045332 on Build GRCh38
rs816 Chr.8: 98045276-98045276 on Build GRCh38
rs908071586 Chr.8: 98045249-98045249 on Build GRCh38
rs112468901 Chr.8: 98045460-98045460 on Build GRCh38
rs1042614096 Chr.8: 98045562-98045562 on Build GRCh38
rs1010135448 Chr.8: 98045368-98045368 on Build GRCh38
rs987561910 Chr.8: 98045235-98045235 on Build GRCh38
rs118025902 Chr.8: 98045569-98045569 on Build GRCh38
rs6984226 Chr.8: 98045513-98045513 on Build GRCh38
rs755106560 Chr.8: 98045370-98045370 on Build GRCh38
rs945905682 Chr.8: 98045561-98045561 on Build GRCh38
rs764702613 Chr.8: 98045325-98045325 on Build GRCh38
rs1028953350 Chr.8: 98045393-98045393 on Build GRCh38
rs771619783 Chr.8: 98045414-98045414 on Build GRCh38
rs774051404 Chr.8: 98045302-98045302 on Build GRCh38
rs1056924758 Chr.8: 98045540-98045540 on Build GRCh38
rs974093757 Chr.8: 98045272-98045272 on Build GRCh38
rs535351227 Chr.8: 98045439-98045439 on Build GRCh38
rs931005965 Chr.8: 98045291-98045291 on Build GRCh38
rs201982680 Chr.8: 98045381-98045381 on Build GRCh38

View Full Product Details