Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123231586-123232099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf76
Entrez Gene ID
84933
Gene Name
chromosome 8 open reading frame 76
Gene Aliases
-
UniGene
Hs.707401
Gene Chromosome Location
Chr.8: 123219956-123241398 on Build GRCh38
Transcripts
NM_032847.2 NM_001204180.1

Gene Symbol
ZHX1-C8orf76
Entrez Gene ID
100533106
Gene Name
ZHX1-C8orf76 readthrough
Gene Aliases
-
UniGene
Hs.707401
Gene Chromosome Location
Chr.8: 123226189-123274487 on Build GRCh38
Transcripts
NM_032847.2 NM_001204180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5750227 c.498C>G p.N166K Chr.8: 123231617-123231617 on Build GRCh38
5683883 c.393G>T p.T131T Chr.8: 123231722-123231722 on Build GRCh38
4496739 c.481C>T p.P161S Chr.8: 123231634-123231634 on Build GRCh38
254593 c.448A>G p.I150V Chr.8: 123231667-123231667 on Build GRCh38
6616747 c.392C>T p.T131M Chr.8: 123231723-123231723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35968111 Chr.8: 123231912-123231913 on Build GRCh38
rs781288876 Chr.8: 123231836-123231836 on Build GRCh38
rs567657005 Chr.8: 123231956-123231957 on Build GRCh38
rs752592288 Chr.8: 123231771-123231771 on Build GRCh38
rs775991570 Chr.8: 123231711-123231711 on Build GRCh38
rs534967592 Chr.8: 123231967-123231967 on Build GRCh38
rs778672959 Chr.8: 123231770-123231770 on Build GRCh38
rs767252268 Chr.8: 123231721-123231721 on Build GRCh38
rs751697928 Chr.8: 123231723-123231723 on Build GRCh38
rs777308267 Chr.8: 123231700-123231700 on Build GRCh38
rs750007760 Chr.8: 123231618-123231618 on Build GRCh38
rs776063848 Chr.8: 123231766-123231766 on Build GRCh38
rs936035350 Chr.8: 123232054-123232054 on Build GRCh38
rs964919164 Chr.8: 123232063-123232063 on Build GRCh38
rs763228154 Chr.8: 123231765-123231766 on Build GRCh38
rs184745962 Chr.8: 123231833-123231833 on Build GRCh38
rs748318333 Chr.8: 123231670-123231670 on Build GRCh38
rs374725759 Chr.8: 123231763-123231763 on Build GRCh38
rs923714051 Chr.8: 123231906-123231906 on Build GRCh38
rs756748601 Chr.8: 123231788-123231789 on Build GRCh38
rs1038568941 Chr.8: 123231939-123231939 on Build GRCh38
rs780010612 Chr.8: 123231629-123231629 on Build GRCh38
rs375138837 Chr.8: 123231817-123231817 on Build GRCh38
rs1056075095 Chr.8: 123231921-123231921 on Build GRCh38
rs916467311 Chr.8: 123231923-123231923 on Build GRCh38
rs1029875836 Chr.8: 123231776-123231776 on Build GRCh38
rs1047791076 Chr.8: 123231958-123231958 on Build GRCh38
rs945301679 Chr.8: 123231949-123231949 on Build GRCh38
rs201885819 Chr.8: 123231784-123231784 on Build GRCh38
rs915415599 Chr.8: 123231867-123231867 on Build GRCh38
rs35816159 Chr.8: 123231749-123231750 on Build GRCh38
rs749045273 Chr.8: 123231636-123231636 on Build GRCh38
rs980504543 Chr.8: 123231893-123231893 on Build GRCh38
rs759923053 Chr.8: 123231702-123231702 on Build GRCh38
rs200399778 Chr.8: 123231623-123231623 on Build GRCh38
rs764600082 Chr.8: 123231776-123231776 on Build GRCh38
rs779047019 Chr.8: 123231657-123231657 on Build GRCh38
rs771532120 Chr.8: 123231699-123231699 on Build GRCh38
rs1056296549 Chr.8: 123231691-123231691 on Build GRCh38
rs77482613 Chr.8: 123231766-123231766 on Build GRCh38
rs765684278 Chr.8: 123231745-123231752 on Build GRCh38
rs754793441 Chr.8: 123231646-123231646 on Build GRCh38
rs199631843 Chr.8: 123231681-123231681 on Build GRCh38
rs771156288 Chr.8: 123231808-123231808 on Build GRCh38
rs781737466 Chr.8: 123231809-123231809 on Build GRCh38
rs572205575 Chr.8: 123231811-123231811 on Build GRCh38
rs1041710479 Chr.8: 123231634-123231634 on Build GRCh38
rs760267905 Chr.8: 123231741-123231741 on Build GRCh38
rs560911652 Chr.8: 123231877-123231877 on Build GRCh38
rs549081517 Chr.8: 123231812-123231812 on Build GRCh38
rs147625869 Chr.8: 123231696-123231696 on Build GRCh38
rs772402528 Chr.8: 123231684-123231684 on Build GRCh38
rs992406933 Chr.8: 123231906-123231906 on Build GRCh38
rs368935182 Chr.8: 123231882-123231882 on Build GRCh38
rs913786312 Chr.8: 123231947-123231947 on Build GRCh38
rs3214807 Chr.8: 123231775-123231775 on Build GRCh38
rs766259331 Chr.8: 123231748-123231748 on Build GRCh38
rs1018965221 Chr.8: 123232058-123232058 on Build GRCh38
rs747020189 Chr.8: 123231807-123231807 on Build GRCh38
rs974874635 Chr.8: 123231855-123231855 on Build GRCh38
rs759884352 Chr.8: 123231724-123231724 on Build GRCh38
rs947979448 Chr.8: 123231930-123231933 on Build GRCh38
rs1038217840 Chr.8: 123231714-123231714 on Build GRCh38
rs113759997 Chr.8: 123231804-123231804 on Build GRCh38
rs370613823 Chr.8: 123231722-123231722 on Build GRCh38
rs189931490 Chr.8: 123231690-123231690 on Build GRCh38
rs761359957 Chr.8: 123231719-123231719 on Build GRCh38
rs758525817 Chr.8: 123231777-123231777 on Build GRCh38
rs765701081 Chr.8: 123231703-123231703 on Build GRCh38
rs904518547 Chr.8: 123231985-123231985 on Build GRCh38
rs968661243 Chr.8: 123231879-123231879 on Build GRCh38
rs756721731 Chr.8: 123231617-123231617 on Build GRCh38
rs752027404 Chr.8: 123231780-123231782 on Build GRCh38
rs926395191 Chr.8: 123231901-123231901 on Build GRCh38
rs553672150 Chr.8: 123232055-123232055 on Build GRCh38
rs939198555 Chr.8: 123231902-123231902 on Build GRCh38
rs753783530 Chr.8: 123231761-123231761 on Build GRCh38
rs111843338 Chr.8: 123231656-123231656 on Build GRCh38
rs982439652 Chr.8: 123231830-123231830 on Build GRCh38

View Full Product Details