Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 132614845-132615391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC6
Entrez Gene ID
23639
Gene Name
leucine rich repeat containing 6
Gene Aliases
CILD19, LRTP, TSLRP
UniGene
Hs.591865
Gene Chromosome Location
Chr.8: 132570419-132685039 on Build GRCh38
Transcripts
NM_001321961.1 NM_001321962.1 NM_001321963.1 NM_001321964.1 NM_001321965.1 NM_001321966.1 NM_012472.5 NR_073525.2 NR_135905.1 NR_135906.1 NR_135907.1 NR_135908.1 NR_135909.1 NR_135910.1 NR_135911.1 NR_135912.1 NR_135913.1 XM_006716538.3 XM_011516950.2 XM_017013296.1 XM_017013297.1 XM_017013298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755569 c.926C>T p.S309F Chr.8: 132615086-132615086 on Build GRCh38
1700648 c.924C>A p.F308L Chr.8: 132615088-132615088 on Build GRCh38
7375645 c.949A>G p.I317V Chr.8: 132615063-132615063 on Build GRCh38
4437246 c.974+2T>C p.? Chr.8: 132615036-132615036 on Build GRCh38
7213587 c.960C>A p.D320E Chr.8: 132615052-132615052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773789654 Chr.8: 132615065-132615065 on Build GRCh38
rs763711548 Chr.8: 132615013-132615013 on Build GRCh38
rs576683877 Chr.8: 132615256-132615256 on Build GRCh38
rs1008520815 Chr.8: 132615007-132615007 on Build GRCh38
rs996164379 Chr.8: 132615248-132615248 on Build GRCh38
rs76110264 Chr.8: 132614874-132614874 on Build GRCh38
rs75851179 Chr.8: 132614997-132614997 on Build GRCh38
rs943611081 Chr.8: 132614912-132614912 on Build GRCh38
rs945529088 Chr.8: 132614939-132614939 on Build GRCh38
rs778865895 Chr.8: 132615120-132615121 on Build GRCh38
rs751135026 Chr.8: 132615015-132615015 on Build GRCh38
rs758084942 Chr.8: 132615004-132615004 on Build GRCh38
rs761129792 Chr.8: 132615066-132615066 on Build GRCh38
rs777027062 Chr.8: 132615076-132615076 on Build GRCh38
rs369861605 Chr.8: 132615028-132615028 on Build GRCh38
rs761114246 Chr.8: 132614993-132614993 on Build GRCh38
rs761412567 Chr.8: 132615105-132615105 on Build GRCh38
rs146067670 Chr.8: 132615063-132615063 on Build GRCh38
rs967922654 Chr.8: 132615349-132615349 on Build GRCh38
rs755119635 Chr.8: 132615093-132615094 on Build GRCh38
rs753754799 Chr.8: 132615143-132615143 on Build GRCh38
rs373165179 Chr.8: 132615123-132615123 on Build GRCh38
rs913389954 Chr.8: 132615108-132615108 on Build GRCh38
rs190862296 Chr.8: 132614894-132614894 on Build GRCh38
rs768137707 Chr.8: 132614988-132614988 on Build GRCh38
rs928298463 Chr.8: 132615139-132615139 on Build GRCh38
rs55641476 Chr.8: 132615231-132615231 on Build GRCh38
rs886906189 Chr.8: 132615228-132615228 on Build GRCh38
rs981657725 Chr.8: 132615124-132615124 on Build GRCh38
rs779210545 Chr.8: 132615147-132615147 on Build GRCh38
rs750711824 Chr.8: 132615133-132615133 on Build GRCh38
rs772224728 Chr.8: 132615149-132615149 on Build GRCh38
rs779000571 Chr.8: 132615041-132615041 on Build GRCh38
rs771987194 Chr.8: 132615064-132615064 on Build GRCh38
rs139090169 Chr.8: 132615352-132615352 on Build GRCh38
rs771394029 Chr.8: 132615073-132615073 on Build GRCh38
rs754771874 Chr.8: 132614947-132614947 on Build GRCh38
rs4602866 Chr.8: 132615273-132615273 on Build GRCh38
rs763799383 Chr.8: 132615093-132615093 on Build GRCh38
rs754753691 Chr.8: 132615146-132615146 on Build GRCh38
rs371099222 Chr.8: 132615233-132615233 on Build GRCh38
rs780127916 Chr.8: 132615129-132615129 on Build GRCh38
rs769060474 Chr.8: 132615029-132615029 on Build GRCh38
rs748069129 Chr.8: 132615021-132615021 on Build GRCh38
rs773525376 Chr.8: 132614989-132614989 on Build GRCh38
rs182037288 Chr.8: 132614925-132614925 on Build GRCh38
rs936431057 Chr.8: 132615341-132615341 on Build GRCh38
rs182830760 Chr.8: 132615247-132615247 on Build GRCh38
rs576461946 Chr.8: 132615367-132615367 on Build GRCh38
rs1055454831 Chr.8: 132615167-132615167 on Build GRCh38
rs925426046 Chr.8: 132615304-132615304 on Build GRCh38
rs1020459566 Chr.8: 132615297-132615297 on Build GRCh38
rs551327320 Chr.8: 132614983-132614983 on Build GRCh38
rs200544802 Chr.8: 132615116-132615116 on Build GRCh38
rs756172763 Chr.8: 132615127-132615127 on Build GRCh38
rs972963097 Chr.8: 132615135-132615135 on Build GRCh38
rs957674260 Chr.8: 132615072-132615072 on Build GRCh38
rs544058059 Chr.8: 132615258-132615258 on Build GRCh38
rs753902965 Chr.8: 132615092-132615093 on Build GRCh38
rs748268973 Chr.8: 132615148-132615148 on Build GRCh38
rs529096167 Chr.8: 132614879-132614879 on Build GRCh38
rs2293978 Chr.8: 132615357-132615357 on Build GRCh38
rs919721154 Chr.8: 132615187-132615187 on Build GRCh38
rs756008255 Chr.8: 132615022-132615022 on Build GRCh38
rs555074391 Chr.8: 132615289-132615289 on Build GRCh38
rs751102396 Chr.8: 132615095-132615095 on Build GRCh38
rs770497274 Chr.8: 132615232-132615232 on Build GRCh38
rs759877163 Chr.8: 132615077-132615077 on Build GRCh38
rs766680793 Chr.8: 132614995-132614995 on Build GRCh38
rs535646544 Chr.8: 132615210-132615210 on Build GRCh38
rs117077775 Chr.8: 132614916-132614916 on Build GRCh38
rs931053393 Chr.8: 132615204-132615204 on Build GRCh38
rs777136238 Chr.8: 132615150-132615150 on Build GRCh38
rs952123459 Chr.8: 132615215-132615215 on Build GRCh38
rs376302322 Chr.8: 132615126-132615127 on Build GRCh38
rs780037666 Chr.8: 132615024-132615024 on Build GRCh38
rs185791758 Chr.8: 132615020-132615020 on Build GRCh38

View Full Product Details