Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21077805-21078242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNB1
Entrez Gene ID
3456
Gene Name
interferon beta 1
Gene Aliases
IFB, IFF, IFN-beta, IFNB
UniGene
Hs.93177
Gene Chromosome Location
Chr.9: 21077105-21077963 on Build GRCh38
Transcripts
NM_002176.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202037 c.9C>A p.N3K Chr.9: 21077861-21077861 on Build GRCh38
1107772 c.63C>A p.S21S Chr.9: 21077807-21077807 on Build GRCh38
6847630 c.39G>C p.L13F Chr.9: 21077831-21077831 on Build GRCh38
5922744 c.23A>G p.Q8R Chr.9: 21077847-21077847 on Build GRCh38
4691830 c.54A>G p.T18T Chr.9: 21077816-21077816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369041139 Chr.9: 21077871-21077871 on Build GRCh38
rs913267987 Chr.9: 21077875-21077875 on Build GRCh38
rs749835966 Chr.9: 21077858-21077858 on Build GRCh38
rs981862747 Chr.9: 21078001-21078001 on Build GRCh38
rs762636789 Chr.9: 21078074-21078074 on Build GRCh38
rs111329783 Chr.9: 21078165-21078169 on Build GRCh38
rs41309796 Chr.9: 21078174-21078174 on Build GRCh38
rs780381074 Chr.9: 21077856-21077856 on Build GRCh38
rs752635319 Chr.9: 21077904-21077904 on Build GRCh38
rs41309794 Chr.9: 21077881-21077881 on Build GRCh38
rs765105483 Chr.9: 21077898-21077898 on Build GRCh38
rs911201985 Chr.9: 21078158-21078158 on Build GRCh38
rs994345763 Chr.9: 21078187-21078187 on Build GRCh38
rs1056616528 Chr.9: 21077844-21077844 on Build GRCh38
rs1018659189 Chr.9: 21078148-21078148 on Build GRCh38
rs1004541371 Chr.9: 21078140-21078140 on Build GRCh38
rs992149429 Chr.9: 21077983-21077983 on Build GRCh38
rs762373714 Chr.9: 21077905-21077905 on Build GRCh38
rs1047223367 Chr.9: 21078193-21078193 on Build GRCh38
rs1045326163 Chr.9: 21078054-21078054 on Build GRCh38
rs573455496 Chr.9: 21078213-21078213 on Build GRCh38
rs529016888 Chr.9: 21078140-21078140 on Build GRCh38
rs373788631 Chr.9: 21077880-21077880 on Build GRCh38
rs946601124 Chr.9: 21078118-21078118 on Build GRCh38
rs969559249 Chr.9: 21077990-21077990 on Build GRCh38
rs1029201247 Chr.9: 21077953-21077953 on Build GRCh38
rs756874732 Chr.9: 21077916-21077916 on Build GRCh38
rs1005226773 Chr.9: 21077981-21077981 on Build GRCh38
rs948344350 Chr.9: 21077999-21077999 on Build GRCh38
rs751155447 Chr.9: 21077841-21077841 on Build GRCh38
rs886217104 Chr.9: 21078115-21078115 on Build GRCh38
rs935315376 Chr.9: 21077980-21077980 on Build GRCh38
rs763600638 Chr.9: 21077909-21077909 on Build GRCh38
rs538690297 Chr.9: 21077866-21077866 on Build GRCh38
rs1023273888 Chr.9: 21077989-21077989 on Build GRCh38
rs994855505 Chr.9: 21077930-21077930 on Build GRCh38
rs1048821471 Chr.9: 21078099-21078099 on Build GRCh38
rs901371957 Chr.9: 21078182-21078182 on Build GRCh38
rs967914622 Chr.9: 21077954-21077954 on Build GRCh38
rs915159630 Chr.9: 21078103-21078103 on Build GRCh38
rs1043050957 Chr.9: 21078134-21078134 on Build GRCh38
rs370094551 Chr.9: 21077891-21077891 on Build GRCh38
rs112752385 Chr.9: 21077939-21077939 on Build GRCh38
rs927792843 Chr.9: 21077857-21077857 on Build GRCh38
rs979456857 Chr.9: 21077957-21077957 on Build GRCh38
rs777321944 Chr.9: 21077834-21077834 on Build GRCh38
rs760969308 Chr.9: 21077890-21077890 on Build GRCh38
rs898627518 Chr.9: 21078066-21078066 on Build GRCh38
rs759593196 Chr.9: 21077897-21077897 on Build GRCh38
rs751227111 Chr.9: 21077910-21077910 on Build GRCh38
rs1045674540 Chr.9: 21077861-21077861 on Build GRCh38
rs147529998 Chr.9: 21078217-21078217 on Build GRCh38
rs1005779500 Chr.9: 21078214-21078214 on Build GRCh38
rs776533488 Chr.9: 21077892-21077892 on Build GRCh38
rs924045247 Chr.9: 21077977-21077977 on Build GRCh38
rs148878013 Chr.9: 21077850-21077850 on Build GRCh38
rs968261414 Chr.9: 21078185-21078185 on Build GRCh38
rs888710473 Chr.9: 21078197-21078197 on Build GRCh38
rs988055793 Chr.9: 21078100-21078100 on Build GRCh38
rs927814724 Chr.9: 21078008-21078008 on Build GRCh38
rs562612025 Chr.9: 21077974-21077974 on Build GRCh38
rs564695755 Chr.9: 21078178-21078178 on Build GRCh38
rs918162039 Chr.9: 21077988-21077988 on Build GRCh38
rs901323785 Chr.9: 21077929-21077929 on Build GRCh38
rs531418929 Chr.9: 21078097-21078097 on Build GRCh38
rs773323040 Chr.9: 21077886-21077886 on Build GRCh38
rs542749153 Chr.9: 21077944-21077944 on Build GRCh38
rs574896435 Chr.9: 21077924-21077924 on Build GRCh38
rs201099689 Chr.9: 21077918-21077918 on Build GRCh38
rs867273416 Chr.9: 21077923-21077923 on Build GRCh38
rs201629499 Chr.9: 21077877-21077877 on Build GRCh38

View Full Product Details