Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32785979-32786485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM215
Entrez Gene ID
401498
Gene Name
transmembrane protein 215
Gene Aliases
-
UniGene
Hs.522063
Gene Chromosome Location
Chr.9: 32783499-32789201 on Build GRCh38
Transcripts
NM_212558.2

SNP Detail

SNP ID Chromosome Location
rs920769293 Chr.9: 32786420-32786420 on Build GRCh38
rs555362941 Chr.9: 32786175-32786175 on Build GRCh38
rs903985094 Chr.9: 32786073-32786073 on Build GRCh38
rs1017843460 Chr.9: 32786399-32786399 on Build GRCh38
rs374805713 Chr.9: 32786450-32786450 on Build GRCh38
rs571899465 Chr.9: 32786187-32786187 on Build GRCh38
rs964879405 Chr.9: 32786387-32786387 on Build GRCh38
rs916905428 Chr.9: 32786177-32786177 on Build GRCh38
rs905805335 Chr.9: 32786305-32786305 on Build GRCh38
rs1034322290 Chr.9: 32786090-32786090 on Build GRCh38
rs569751937 Chr.9: 32786101-32786101 on Build GRCh38
rs144444168 Chr.9: 32786288-32786288 on Build GRCh38
rs535165880 Chr.9: 32786156-32786156 on Build GRCh38
rs929071321 Chr.9: 32786109-32786109 on Build GRCh38
rs985238930 Chr.9: 32786287-32786287 on Build GRCh38
rs1054299065 Chr.9: 32786342-32786342 on Build GRCh38
rs780505478 Chr.9: 32786373-32786373 on Build GRCh38
rs1009646466 Chr.9: 32786354-32786354 on Build GRCh38
rs180756640 Chr.9: 32786415-32786415 on Build GRCh38
rs954676997 Chr.9: 32786385-32786385 on Build GRCh38
rs538568361 Chr.9: 32786357-32786357 on Build GRCh38
rs1000403763 Chr.9: 32786324-32786324 on Build GRCh38
rs556322814 Chr.9: 32786019-32786019 on Build GRCh38
rs1011320535 Chr.9: 32786046-32786046 on Build GRCh38
rs891663752 Chr.9: 32786345-32786345 on Build GRCh38
rs1044271866 Chr.9: 32786264-32786264 on Build GRCh38
rs923169904 Chr.9: 32786436-32786436 on Build GRCh38
rs987712049 Chr.9: 32786010-32786010 on Build GRCh38
rs1021081928 Chr.9: 32786381-32786381 on Build GRCh38
rs891540230 Chr.9: 32786024-32786024 on Build GRCh38
rs1002389762 Chr.9: 32786080-32786080 on Build GRCh38
rs1013577013 Chr.9: 32786115-32786115 on Build GRCh38
rs1042815203 Chr.9: 32786065-32786065 on Build GRCh38
rs1047506595 Chr.9: 32786162-32786162 on Build GRCh38
rs974630156 Chr.9: 32786408-32786408 on Build GRCh38
rs372198004 Chr.9: 32786367-32786367 on Build GRCh38
rs761138406 Chr.9: 32786210-32786210 on Build GRCh38

View Full Product Details