Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113011056-113011553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF883
Entrez Gene ID
169834
Gene Name
zinc finger protein 883
Gene Aliases
-
UniGene
Hs.192877
Gene Chromosome Location
Chr.9: 112997120-113012192 on Build GRCh38
Transcripts
NM_001101338.1

SNP Detail

SNP ID Chromosome Location
rs984360885 Chr.9: 113011245-113011245 on Build GRCh38
rs895459184 Chr.9: 113011122-113011122 on Build GRCh38
rs895594045 Chr.9: 113011521-113011521 on Build GRCh38
rs149340126 Chr.9: 113011195-113011195 on Build GRCh38
rs759125239 Chr.9: 113011197-113011197 on Build GRCh38
rs369198869 Chr.9: 113011389-113011389 on Build GRCh38
rs951187702 Chr.9: 113011381-113011381 on Build GRCh38
rs548452685 Chr.9: 113011262-113011262 on Build GRCh38
rs41280199 Chr.9: 113011287-113011287 on Build GRCh38
rs192322373 Chr.9: 113011259-113011259 on Build GRCh38
rs905320175 Chr.9: 113011459-113011459 on Build GRCh38
rs763093456 Chr.9: 113011358-113011358 on Build GRCh38
rs555880763 Chr.9: 113011503-113011503 on Build GRCh38
rs561738312 Chr.9: 113011277-113011277 on Build GRCh38
rs13299990 Chr.9: 113011326-113011326 on Build GRCh38
rs1025551708 Chr.9: 113011192-113011192 on Build GRCh38
rs939516411 Chr.9: 113011465-113011465 on Build GRCh38
rs1035439670 Chr.9: 113011086-113011086 on Build GRCh38
rs138347691 Chr.9: 113011425-113011425 on Build GRCh38
rs776968582 Chr.9: 113011209-113011209 on Build GRCh38
rs144676214 Chr.9: 113011345-113011345 on Build GRCh38
rs13300013 Chr.9: 113011375-113011375 on Build GRCh38
rs13300144 Chr.9: 113011405-113011405 on Build GRCh38
rs1011049394 Chr.9: 113011282-113011283 on Build GRCh38
rs372778600 Chr.9: 113011517-113011517 on Build GRCh38
rs1048809513 Chr.9: 113011158-113011158 on Build GRCh38
rs13301450 Chr.9: 113011395-113011395 on Build GRCh38
rs918312692 Chr.9: 113011347-113011347 on Build GRCh38
rs935305683 Chr.9: 113011306-113011306 on Build GRCh38
rs929798217 Chr.9: 113011080-113011080 on Build GRCh38
rs549446668 Chr.9: 113011409-113011409 on Build GRCh38
rs530712353 Chr.9: 113011313-113011313 on Build GRCh38
rs190479453 Chr.9: 113011453-113011453 on Build GRCh38
rs185558096 Chr.9: 113011332-113011332 on Build GRCh38
rs771990854 Chr.9: 113011271-113011271 on Build GRCh38
rs796247836 Chr.9: 113011199-113011200 on Build GRCh38
rs993844073 Chr.9: 113011183-113011183 on Build GRCh38
rs568252981 Chr.9: 113011477-113011480 on Build GRCh38
rs964231692 Chr.9: 113011270-113011270 on Build GRCh38
rs13300137 Chr.9: 113011361-113011361 on Build GRCh38
rs916355585 Chr.9: 113011388-113011388 on Build GRCh38
rs892650574 Chr.9: 113011280-113011280 on Build GRCh38

View Full Product Details