Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120962589-120963096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5
Entrez Gene ID
727
Gene Name
complement component 5
Gene Aliases
C5D, C5a, C5b, CPAMD4, ECLZB
UniGene
Hs.494997
Gene Chromosome Location
Chr.9: 120952335-121075174 on Build GRCh38
Transcripts
NM_001317163.1 NM_001735.2 XM_011518980.2 XM_017015102.1 XM_017015103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4628821 c.4459T>C p.F1487L Chr.9: 120962716-120962716 on Build GRCh38
3653757 c.4426C>G p.R1476G Chr.9: 120962749-120962749 on Build GRCh38
1319666 c.4458G>T p.G1486G Chr.9: 120962717-120962717 on Build GRCh38
3322667 c.4426C>T p.R1476* Chr.9: 120962749-120962749 on Build GRCh38
3653758 c.4404C>T p.P1468P Chr.9: 120962771-120962771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762741385 Chr.9: 120962988-120962988 on Build GRCh38
rs775939939 Chr.9: 120962742-120962742 on Build GRCh38
rs761105830 Chr.9: 120962940-120962940 on Build GRCh38
rs576334993 Chr.9: 120962985-120962985 on Build GRCh38
rs777014012 Chr.9: 120962963-120962963 on Build GRCh38
rs55649448 Chr.9: 120962721-120962721 on Build GRCh38
rs912643424 Chr.9: 120963044-120963044 on Build GRCh38
rs372486611 Chr.9: 120962816-120962816 on Build GRCh38
rs765216720 Chr.9: 120962785-120962785 on Build GRCh38
rs375227918 Chr.9: 120962657-120962657 on Build GRCh38
rs751384128 Chr.9: 120962851-120962851 on Build GRCh38
rs868488283 Chr.9: 120962998-120962998 on Build GRCh38
rs375865204 Chr.9: 120962975-120962975 on Build GRCh38
rs750356445 Chr.9: 120962806-120962806 on Build GRCh38
rs113011708 Chr.9: 120962892-120962892 on Build GRCh38
rs771390003 Chr.9: 120962701-120962701 on Build GRCh38
rs137867011 Chr.9: 120962667-120962667 on Build GRCh38
rs574469025 Chr.9: 120962974-120962974 on Build GRCh38
rs762012629 Chr.9: 120962773-120962773 on Build GRCh38
rs756358379 Chr.9: 120962775-120962776 on Build GRCh38
rs1032063037 Chr.9: 120962723-120962723 on Build GRCh38
rs777301249 Chr.9: 120962901-120962901 on Build GRCh38
rs758430233 Chr.9: 120962820-120962820 on Build GRCh38
rs746367534 Chr.9: 120962902-120962902 on Build GRCh38
rs780932441 Chr.9: 120962862-120962862 on Build GRCh38
rs534785208 Chr.9: 120962672-120962672 on Build GRCh38
rs949696280 Chr.9: 120962633-120962633 on Build GRCh38
rs569109691 Chr.9: 120962624-120962624 on Build GRCh38
rs750576128 Chr.9: 120962771-120962772 on Build GRCh38
rs537359464 Chr.9: 120962638-120962638 on Build GRCh38
rs892646565 Chr.9: 120962859-120962859 on Build GRCh38
rs370935824 Chr.9: 120962687-120962687 on Build GRCh38
rs376245744 Chr.9: 120962686-120962686 on Build GRCh38
rs965313093 Chr.9: 120962894-120962894 on Build GRCh38
rs952512953 Chr.9: 120962951-120962951 on Build GRCh38
rs1019722923 Chr.9: 120962838-120962838 on Build GRCh38
rs758246973 Chr.9: 120962660-120962660 on Build GRCh38
rs754827684 Chr.9: 120963004-120963004 on Build GRCh38
rs121909588 Chr.9: 120962749-120962749 on Build GRCh38
rs74749615 Chr.9: 120962708-120962708 on Build GRCh38
rs1036788887 Chr.9: 120962618-120962618 on Build GRCh38
rs767353203 Chr.9: 120963005-120963005 on Build GRCh38
rs762167893 Chr.9: 120962964-120962964 on Build GRCh38
rs763961337 Chr.9: 120962660-120962661 on Build GRCh38
rs983931912 Chr.9: 120963007-120963007 on Build GRCh38
rs369606211 Chr.9: 120962663-120962663 on Build GRCh38
rs772690228 Chr.9: 120962906-120962906 on Build GRCh38
rs954549642 Chr.9: 120963003-120963003 on Build GRCh38
rs975770185 Chr.9: 120962900-120962900 on Build GRCh38
rs1053007328 Chr.9: 120962843-120962843 on Build GRCh38
rs373515982 Chr.9: 120962705-120962705 on Build GRCh38
rs368329850 Chr.9: 120962760-120962760 on Build GRCh38
rs747610446 Chr.9: 120962887-120962887 on Build GRCh38
rs780059425 Chr.9: 120962983-120962983 on Build GRCh38
rs377213091 Chr.9: 120962917-120962917 on Build GRCh38
rs775658749 Chr.9: 120962923-120962923 on Build GRCh38
rs138933092 Chr.9: 120962743-120962743 on Build GRCh38
rs1039760315 Chr.9: 120962802-120962802 on Build GRCh38
rs371393264 Chr.9: 120962946-120962946 on Build GRCh38
rs754524189 Chr.9: 120962858-120962858 on Build GRCh38
rs552463689 Chr.9: 120962648-120962648 on Build GRCh38
rs575092054 Chr.9: 120963063-120963063 on Build GRCh38
rs780707722 Chr.9: 120962893-120962893 on Build GRCh38
rs1001034253 Chr.9: 120962668-120962668 on Build GRCh38
rs751276454 Chr.9: 120962995-120962995 on Build GRCh38
rs535154384 Chr.9: 120962989-120962989 on Build GRCh38
rs904070902 Chr.9: 120962803-120962803 on Build GRCh38
rs756395867 Chr.9: 120962621-120962621 on Build GRCh38
rs747665589 Chr.9: 120962696-120962696 on Build GRCh38
rs369172301 Chr.9: 120963015-120963015 on Build GRCh38
rs942770289 Chr.9: 120963050-120963050 on Build GRCh38
rs528221911 Chr.9: 120962627-120962627 on Build GRCh38
rs779701089 Chr.9: 120962824-120962824 on Build GRCh38

View Full Product Details