Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 25003899-25004440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARX
Entrez Gene ID
170302
Gene Name
aristaless related homeobox
Gene Aliases
CT121, EIEE1, ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS
UniGene
Hs.300304
Gene Chromosome Location
Chr.X: 25003694-25015948 on Build GRCh38
Transcripts
NM_139058.2

SNP Detail

SNP ID Chromosome Location
rs771348025 Chr.X: 25004292-25004292 on Build GRCh38
rs869211274 Chr.X: 25004001-25004002 on Build GRCh38
rs1038585538 Chr.X: 25004273-25004273 on Build GRCh38
rs952399658 Chr.X: 25003948-25003948 on Build GRCh38
rs1006681318 Chr.X: 25003978-25003978 on Build GRCh38
rs916388011 Chr.X: 25004263-25004263 on Build GRCh38
rs936756316 Chr.X: 25004241-25004241 on Build GRCh38
rs773507925 Chr.X: 25004261-25004264 on Build GRCh38
rs964935996 Chr.X: 25003984-25003988 on Build GRCh38
rs888144695 Chr.X: 25004362-25004362 on Build GRCh38
rs1055171463 Chr.X: 25004257-25004257 on Build GRCh38
rs566976212 Chr.X: 25004337-25004337 on Build GRCh38
rs67950419 Chr.X: 25003987-25003987 on Build GRCh38
rs199975831 Chr.X: 25003987-25003988 on Build GRCh38
rs77688950 Chr.X: 25003986-25003986 on Build GRCh38
rs1026657514 Chr.X: 25003939-25003939 on Build GRCh38
rs866962208 Chr.X: 25004181-25004181 on Build GRCh38
rs773853529 Chr.X: 25004315-25004315 on Build GRCh38
rs764605075 Chr.X: 25004367-25004367 on Build GRCh38
rs925290898 Chr.X: 25004237-25004237 on Build GRCh38
rs1046679941 Chr.X: 25004290-25004290 on Build GRCh38
rs766015643 Chr.X: 25004281-25004281 on Build GRCh38
rs1018180095 Chr.X: 25003984-25003986 on Build GRCh38
rs774656902 Chr.X: 25004308-25004308 on Build GRCh38
rs769672754 Chr.X: 25004002-25004002 on Build GRCh38
rs754327749 Chr.X: 25004412-25004412 on Build GRCh38
rs1048087304 Chr.X: 25004329-25004329 on Build GRCh38
rs1009678018 Chr.X: 25004417-25004417 on Build GRCh38
rs978030923 Chr.X: 25004224-25004224 on Build GRCh38
rs180803666 Chr.X: 25004143-25004143 on Build GRCh38
rs1035032312 Chr.X: 25004406-25004406 on Build GRCh38
rs1047659049 Chr.X: 25004065-25004065 on Build GRCh38
rs762724492 Chr.X: 25004269-25004269 on Build GRCh38
rs1006630281 Chr.X: 25004396-25004396 on Build GRCh38
rs933753543 Chr.X: 25003997-25003997 on Build GRCh38
rs887324369 Chr.X: 25004167-25004167 on Build GRCh38
rs960768415 Chr.X: 25004414-25004414 on Build GRCh38
rs772860962 Chr.X: 25004010-25004010 on Build GRCh38
rs966945731 Chr.X: 25004185-25004185 on Build GRCh38
rs922330719 Chr.X: 25003950-25003950 on Build GRCh38

View Full Product Details