Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185300296-185300833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IVNS1ABP
Entrez Gene ID
10625
Gene Name
influenza virus NS1A binding protein
Gene Aliases
FLARA3, HSPC068, KLHL39, ND1, NS-1, NS1-BP, NS1BP
UniGene
Hs.497183
Gene Chromosome Location
Chr.1: 185292367-185317343 on Build GRCh38
Transcripts
NM_006469.4 XM_005244843.3 XM_011509079.2 XM_011509080.2 XM_017000084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295591 c.1225C>G p.Q409E Chr.1: 185300454-185300454 on Build GRCh38
4601025 c.1163A>G p.Y388C Chr.1: 185300516-185300516 on Build GRCh38
5522930 c.1187C>T p.S396F Chr.1: 185300492-185300492 on Build GRCh38
1211204 c.1181A>G p.H394R Chr.1: 185300498-185300498 on Build GRCh38
424794 c.1253_1254delAT p.Y418fs*11 Chr.1: 185300332-185300333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779759885 Chr.1: 185300387-185300388 on Build GRCh38
rs561065057 Chr.1: 185300807-185300807 on Build GRCh38
rs763214704 Chr.1: 185300347-185300347 on Build GRCh38
rs539708977 Chr.1: 185300414-185300414 on Build GRCh38
rs926153323 Chr.1: 185300563-185300563 on Build GRCh38
rs760496710 Chr.1: 185300613-185300613 on Build GRCh38
rs1032398637 Chr.1: 185300485-185300485 on Build GRCh38
rs759651752 Chr.1: 185300746-185300746 on Build GRCh38
rs906366397 Chr.1: 185300676-185300676 on Build GRCh38
rs1037506860 Chr.1: 185300722-185300723 on Build GRCh38
rs969566300 Chr.1: 185300439-185300439 on Build GRCh38
rs758056702 Chr.1: 185300536-185300536 on Build GRCh38
rs777578199 Chr.1: 185300552-185300552 on Build GRCh38
rs554160376 Chr.1: 185300404-185300404 on Build GRCh38
rs766411765 Chr.1: 185300418-185300418 on Build GRCh38
rs774792652 Chr.1: 185300406-185300406 on Build GRCh38
rs10798006 Chr.1: 185300784-185300784 on Build GRCh38
rs753632316 Chr.1: 185300520-185300520 on Build GRCh38
rs16823897 Chr.1: 185300725-185300725 on Build GRCh38
rs138170199 Chr.1: 185300400-185300400 on Build GRCh38
rs374873846 Chr.1: 185300582-185300582 on Build GRCh38
rs765398037 Chr.1: 185300695-185300698 on Build GRCh38
rs760543632 Chr.1: 185300484-185300484 on Build GRCh38
rs540731830 Chr.1: 185300646-185300646 on Build GRCh38
rs894579593 Chr.1: 185300612-185300612 on Build GRCh38
rs994039918 Chr.1: 185300317-185300317 on Build GRCh38
rs755982128 Chr.1: 185300368-185300368 on Build GRCh38
rs988507151 Chr.1: 185300563-185300564 on Build GRCh38
rs763160067 Chr.1: 185300430-185300430 on Build GRCh38
rs563630816 Chr.1: 185300783-185300783 on Build GRCh38
rs34393280 Chr.1: 185300394-185300394 on Build GRCh38
rs143068443 Chr.1: 185300431-185300431 on Build GRCh38
rs1045316128 Chr.1: 185300791-185300791 on Build GRCh38
rs752990930 Chr.1: 185300346-185300346 on Build GRCh38
rs764790670 Chr.1: 185300525-185300525 on Build GRCh38
rs770276163 Chr.1: 185300608-185300608 on Build GRCh38
rs747325526 Chr.1: 185300579-185300579 on Build GRCh38
rs747780478 Chr.1: 185300393-185300393 on Build GRCh38
rs777212809 Chr.1: 185300420-185300420 on Build GRCh38
rs979614365 Chr.1: 185300482-185300482 on Build GRCh38
rs371309946 Chr.1: 185300808-185300808 on Build GRCh38
rs1046253595 Chr.1: 185300620-185300620 on Build GRCh38
rs747380218 Chr.1: 185300408-185300408 on Build GRCh38
rs976526907 Chr.1: 185300788-185300788 on Build GRCh38
rs558897444 Chr.1: 185300698-185300698 on Build GRCh38
rs750468870 Chr.1: 185300389-185300389 on Build GRCh38
rs752321106 Chr.1: 185300531-185300531 on Build GRCh38
rs538644369 Chr.1: 185300524-185300524 on Build GRCh38
rs766186509 Chr.1: 185300501-185300501 on Build GRCh38
rs200488794 Chr.1: 185300427-185300427 on Build GRCh38
rs1014234033 Chr.1: 185300791-185300792 on Build GRCh38
rs780056191 Chr.1: 185300382-185300382 on Build GRCh38
rs967541183 Chr.1: 185300799-185300799 on Build GRCh38
rs566086596 Chr.1: 185300410-185300410 on Build GRCh38
rs772935019 Chr.1: 185300450-185300450 on Build GRCh38
rs150401241 Chr.1: 185300363-185300363 on Build GRCh38
rs375862719 Chr.1: 185300421-185300421 on Build GRCh38
rs544962462 Chr.1: 185300734-185300734 on Build GRCh38
rs765324156 Chr.1: 185300337-185300337 on Build GRCh38
rs775484020 Chr.1: 185300609-185300609 on Build GRCh38
rs144406498 Chr.1: 185300507-185300507 on Build GRCh38
rs767290254 Chr.1: 185300361-185300361 on Build GRCh38
rs769898706 Chr.1: 185300424-185300424 on Build GRCh38
rs754774460 Chr.1: 185300649-185300649 on Build GRCh38
rs993925260 Chr.1: 185300703-185300703 on Build GRCh38
rs138975587 Chr.1: 185300798-185300798 on Build GRCh38
rs1017928300 Chr.1: 185300738-185300738 on Build GRCh38
rs778534833 Chr.1: 185300724-185300724 on Build GRCh38
rs1025520275 Chr.1: 185300722-185300722 on Build GRCh38
rs750632493 Chr.1: 185300771-185300771 on Build GRCh38
rs929112208 Chr.1: 185300746-185300746 on Build GRCh38
rs370600226 Chr.1: 185300390-185300390 on Build GRCh38
rs781447606 Chr.1: 185300587-185300587 on Build GRCh38
rs146142240 Chr.1: 185300596-185300596 on Build GRCh38

View Full Product Details