Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109642453-109642960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs111616134 Chr.X: 109642885-109642885 on Build GRCh38
rs190157038 Chr.X: 109642795-109642795 on Build GRCh38
rs929071495 Chr.X: 109642903-109642903 on Build GRCh38
rs372686883 Chr.X: 109642561-109642561 on Build GRCh38
rs181617170 Chr.X: 109642678-109642678 on Build GRCh38
rs774490336 Chr.X: 109642482-109642482 on Build GRCh38
rs774382151 Chr.X: 109642579-109642580 on Build GRCh38
rs750624131 Chr.X: 109642671-109642671 on Build GRCh38
rs986020871 Chr.X: 109642634-109642634 on Build GRCh38
rs55955620 Chr.X: 109642670-109642670 on Build GRCh38
rs752654601 Chr.X: 109642589-109642589 on Build GRCh38
rs910348971 Chr.X: 109642749-109642749 on Build GRCh38
rs370638294 Chr.X: 109642680-109642680 on Build GRCh38
rs759628048 Chr.X: 109642555-109642555 on Build GRCh38
rs946010249 Chr.X: 109642930-109642930 on Build GRCh38
rs922705545 Chr.X: 109642927-109642927 on Build GRCh38
rs185398001 Chr.X: 109642783-109642783 on Build GRCh38
rs752697391 Chr.X: 109642855-109642855 on Build GRCh38
rs1027297065 Chr.X: 109642604-109642605 on Build GRCh38
rs754851853 Chr.X: 109642737-109642737 on Build GRCh38
rs375133823 Chr.X: 109642852-109642852 on Build GRCh38
rs377139823 Chr.X: 109642614-109642614 on Build GRCh38

View Full Product Details