Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225494188-225494700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENAH
Entrez Gene ID
55740
Gene Name
enabled homolog (Drosophila)
Gene Aliases
ENA, MENA, NDPP1
UniGene
Hs.497893
Gene Chromosome Location
Chr.1: 225486832-225660045 on Build GRCh38
Transcripts
NM_001008493.1 NM_018212.4 XM_005273187.2 XM_011544229.2 XM_017001746.1 XM_017001747.1 XM_017001748.1 XM_017001749.1 XM_017001750.1 XM_017001751.1 XM_017001752.1 XM_017001753.1 XM_017001754.1 XM_017001755.1 XM_017001756.1 XM_017001757.1

SNP Detail

SNP ID Chromosome Location
rs531522245 Chr.1: 225494316-225494316 on Build GRCh38
rs989564236 Chr.1: 225494250-225494250 on Build GRCh38
rs978165049 Chr.1: 225494491-225494492 on Build GRCh38
rs894830051 Chr.1: 225494257-225494257 on Build GRCh38
rs533345009 Chr.1: 225494402-225494402 on Build GRCh38
rs911320504 Chr.1: 225494251-225494251 on Build GRCh38
rs1018126917 Chr.1: 225494457-225494457 on Build GRCh38
rs777533344 Chr.1: 225494396-225494396 on Build GRCh38
rs150313735 Chr.1: 225494593-225494593 on Build GRCh38
rs12146028 Chr.1: 225494323-225494323 on Build GRCh38
rs550021615 Chr.1: 225494370-225494370 on Build GRCh38
rs541639459 Chr.1: 225494660-225494660 on Build GRCh38
rs1033368489 Chr.1: 225494539-225494539 on Build GRCh38
rs752564223 Chr.1: 225494383-225494383 on Build GRCh38
rs1006766530 Chr.1: 225494433-225494433 on Build GRCh38
rs751407926 Chr.1: 225494415-225494415 on Build GRCh38
rs964131684 Chr.1: 225494613-225494613 on Build GRCh38
rs967057799 Chr.1: 225494514-225494514 on Build GRCh38
rs567815843 Chr.1: 225494533-225494533 on Build GRCh38
rs560707845 Chr.1: 225494216-225494216 on Build GRCh38
rs538182448 Chr.1: 225494573-225494573 on Build GRCh38
rs780281409 Chr.1: 225494669-225494669 on Build GRCh38
rs746128085 Chr.1: 225494501-225494501 on Build GRCh38
rs549609288 Chr.1: 225494526-225494526 on Build GRCh38
rs552851039 Chr.1: 225494490-225494490 on Build GRCh38
rs957197370 Chr.1: 225494572-225494572 on Build GRCh38
rs941029787 Chr.1: 225494574-225494576 on Build GRCh38
rs577473791 Chr.1: 225494623-225494623 on Build GRCh38
rs141360537 Chr.1: 225494395-225494395 on Build GRCh38
rs916821013 Chr.1: 225494564-225494564 on Build GRCh38
rs942652618 Chr.1: 225494353-225494353 on Build GRCh38
rs566970731 Chr.1: 225494479-225494479 on Build GRCh38
rs986385469 Chr.1: 225494582-225494582 on Build GRCh38
rs930235057 Chr.1: 225494385-225494385 on Build GRCh38
rs2039620 Chr.1: 225494473-225494473 on Build GRCh38
rs940977233 Chr.1: 225494295-225494300 on Build GRCh38
rs1010842578 Chr.1: 225494478-225494478 on Build GRCh38
rs911269446 Chr.1: 225494586-225494586 on Build GRCh38
rs776433931 Chr.1: 225494290-225494290 on Build GRCh38
rs1764346 Chr.1: 225494483-225494483 on Build GRCh38

View Full Product Details