Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72868611-72869108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCU
Entrez Gene ID
90550
Gene Name
mitochondrial calcium uniporter
Gene Aliases
C10orf42, CCDC109A
UniGene
Hs.591366
Gene Chromosome Location
Chr.10: 72692131-72887694 on Build GRCh38
Transcripts
NM_001270679.1 NM_001270680.1 NM_138357.2 NR_073062.1 XM_017016882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7152058 c.499C>T p.L167F Chr.10: 72868705-72868705 on Build GRCh38
1199791 c.654A>C p.E218D Chr.10: 72868860-72868860 on Build GRCh38
2161587 c.582G>A p.Q194Q Chr.10: 72868788-72868788 on Build GRCh38
6710143 c.526C>A p.L176M Chr.10: 72868732-72868732 on Build GRCh38
1662726 c.565A>C p.T189P Chr.10: 72868771-72868771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766991402 Chr.10: 72868902-72868902 on Build GRCh38
rs763392876 Chr.10: 72868879-72868879 on Build GRCh38
rs986902184 Chr.10: 72869060-72869060 on Build GRCh38
rs754255006 Chr.10: 72868687-72868687 on Build GRCh38
rs561387101 Chr.10: 72868883-72868883 on Build GRCh38
rs765739740 Chr.10: 72868876-72868876 on Build GRCh38
rs747785506 Chr.10: 72868764-72868764 on Build GRCh38
rs879113463 Chr.10: 72868957-72868957 on Build GRCh38
rs775683336 Chr.10: 72868696-72868696 on Build GRCh38
rs772083063 Chr.10: 72868667-72868667 on Build GRCh38
rs932840355 Chr.10: 72868990-72868990 on Build GRCh38
rs140182415 Chr.10: 72868730-72868730 on Build GRCh38
rs763412680 Chr.10: 72868745-72868745 on Build GRCh38
rs974763676 Chr.10: 72868927-72868927 on Build GRCh38
rs764384067 Chr.10: 72868902-72868903 on Build GRCh38
rs1050138269 Chr.10: 72868665-72868665 on Build GRCh38
rs763420697 Chr.10: 72868882-72868882 on Build GRCh38
rs766921426 Chr.10: 72868852-72868852 on Build GRCh38
rs369032318 Chr.10: 72868731-72868731 on Build GRCh38
rs537793734 Chr.10: 72868729-72868729 on Build GRCh38
rs962739662 Chr.10: 72868925-72868925 on Build GRCh38
rs573284159 Chr.10: 72868941-72868941 on Build GRCh38
rs201005434 Chr.10: 72868679-72868679 on Build GRCh38
rs932789503 Chr.10: 72868645-72868645 on Build GRCh38
rs202242355 Chr.10: 72868770-72868770 on Build GRCh38
rs941260682 Chr.10: 72869067-72869067 on Build GRCh38
rs749211583 Chr.10: 72868774-72868774 on Build GRCh38
rs191859204 Chr.10: 72868670-72868670 on Build GRCh38
rs754540135 Chr.10: 72868892-72868892 on Build GRCh38
rs201174780 Chr.10: 72868780-72868780 on Build GRCh38
rs762108402 Chr.10: 72868857-72868857 on Build GRCh38
rs982874337 Chr.10: 72868887-72868887 on Build GRCh38
rs752386146 Chr.10: 72868915-72868915 on Build GRCh38
rs532837008 Chr.10: 72869016-72869016 on Build GRCh38
rs775442121 Chr.10: 72868834-72868834 on Build GRCh38
rs118015950 Chr.10: 72869086-72869086 on Build GRCh38
rs145860328 Chr.10: 72868854-72868854 on Build GRCh38
rs1000562490 Chr.10: 72868748-72868748 on Build GRCh38
rs754600073 Chr.10: 72868752-72868752 on Build GRCh38
rs766851746 Chr.10: 72868881-72868881 on Build GRCh38
rs200035470 Chr.10: 72868762-72868762 on Build GRCh38
rs377256530 Chr.10: 72868715-72868715 on Build GRCh38
rs762924871 Chr.10: 72868659-72868659 on Build GRCh38
rs760737128 Chr.10: 72868838-72868838 on Build GRCh38
rs972358190 Chr.10: 72869084-72869084 on Build GRCh38
rs764350225 Chr.10: 72868680-72868680 on Build GRCh38
rs1012714967 Chr.10: 72868773-72868773 on Build GRCh38
rs751032622 Chr.10: 72868740-72868740 on Build GRCh38
rs921478310 Chr.10: 72868966-72868966 on Build GRCh38
rs773588968 Chr.10: 72868878-72868878 on Build GRCh38
rs761568643 Chr.10: 72868825-72868825 on Build GRCh38
rs377054382 Chr.10: 72868673-72868673 on Build GRCh38
rs779463538 Chr.10: 72868732-72868732 on Build GRCh38
rs770681583 Chr.10: 72868666-72868666 on Build GRCh38
rs540804535 Chr.10: 72868974-72868974 on Build GRCh38
rs185042686 Chr.10: 72868988-72868988 on Build GRCh38

View Full Product Details