Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 82865187-82865691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRG3
Entrez Gene ID
10718
Gene Name
neuregulin 3
Gene Aliases
HRG3, pro-NRG3
UniGene
Hs.125119
Gene Chromosome Location
Chr.10: 81875298-82989979 on Build GRCh38
Transcripts
NM_001010848.3 NM_001165972.1 NM_001165973.1 XM_005269444.4 XM_011539172.2 XM_011539173.2 XM_011539175.2 XM_017015573.1 XM_017015574.1 XM_017015575.1 XM_017015576.1 XM_017015577.1 XM_017015578.1 XM_017015579.1 XM_017015580.1 XM_017015581.1 XM_017015582.1 XM_017015583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3665918 c.385T>C p.F129L Chr.10: 82865431-82865431 on Build GRCh38
4457568 c.1050C>T p.F350F Chr.10: 82865433-82865433 on Build GRCh38
3686987 c.365C>T p.T122I Chr.10: 82865411-82865411 on Build GRCh38
4708157 c.376G>T p.G126W Chr.10: 82865422-82865422 on Build GRCh38
3686986 c.1028C>T p.T343I Chr.10: 82865411-82865411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907588452 Chr.10: 82865378-82865378 on Build GRCh38
rs142131041 Chr.10: 82865425-82865425 on Build GRCh38
rs150985246 Chr.10: 82865419-82865419 on Build GRCh38
rs748723239 Chr.10: 82865380-82865380 on Build GRCh38
rs976786681 Chr.10: 82865533-82865533 on Build GRCh38
rs946811176 Chr.10: 82865605-82865605 on Build GRCh38
rs1029909940 Chr.10: 82865348-82865348 on Build GRCh38
rs371756880 Chr.10: 82865522-82865522 on Build GRCh38
rs899843287 Chr.10: 82865653-82865653 on Build GRCh38
rs893655633 Chr.10: 82865590-82865590 on Build GRCh38
rs945314805 Chr.10: 82865298-82865298 on Build GRCh38
rs1018213444 Chr.10: 82865328-82865328 on Build GRCh38
rs965410151 Chr.10: 82865516-82865516 on Build GRCh38
rs545018033 Chr.10: 82865347-82865347 on Build GRCh38
rs976693150 Chr.10: 82865253-82865253 on Build GRCh38
rs757268565 Chr.10: 82865664-82865664 on Build GRCh38
rs935132854 Chr.10: 82865575-82865575 on Build GRCh38
rs999402953 Chr.10: 82865215-82865215 on Build GRCh38
rs781399792 Chr.10: 82865513-82865513 on Build GRCh38
rs982020856 Chr.10: 82865565-82865565 on Build GRCh38
rs770367069 Chr.10: 82865397-82865397 on Build GRCh38
rs367893412 Chr.10: 82865212-82865212 on Build GRCh38
rs1052296238 Chr.10: 82865583-82865583 on Build GRCh38
rs1039798813 Chr.10: 82865611-82865611 on Build GRCh38
rs978415886 Chr.10: 82865560-82865560 on Build GRCh38
rs925146877 Chr.10: 82865323-82865323 on Build GRCh38
rs963177687 Chr.10: 82865459-82865459 on Build GRCh38
rs752689864 Chr.10: 82865657-82865657 on Build GRCh38
rs115447589 Chr.10: 82865334-82865334 on Build GRCh38
rs926887058 Chr.10: 82865210-82865210 on Build GRCh38
rs575062813 Chr.10: 82865261-82865261 on Build GRCh38
rs755142589 Chr.10: 82865492-82865492 on Build GRCh38
rs894867429 Chr.10: 82865330-82865330 on Build GRCh38
rs774927520 Chr.10: 82865469-82865469 on Build GRCh38
rs774094908 Chr.10: 82865445-82865445 on Build GRCh38
rs973172497 Chr.10: 82865471-82865471 on Build GRCh38
rs781413474 Chr.10: 82865379-82865379 on Build GRCh38
rs542374225 Chr.10: 82865667-82865667 on Build GRCh38
rs1011303296 Chr.10: 82865315-82865315 on Build GRCh38
rs537560589 Chr.10: 82865314-82865314 on Build GRCh38
rs998143810 Chr.10: 82865661-82865661 on Build GRCh38
rs967402337 Chr.10: 82865559-82865559 on Build GRCh38
rs527364566 Chr.10: 82865364-82865364 on Build GRCh38
rs927904018 Chr.10: 82865571-82865571 on Build GRCh38
rs998385602 Chr.10: 82865342-82865342 on Build GRCh38
rs478010 Chr.10: 82865351-82865351 on Build GRCh38
rs751777933 Chr.10: 82865448-82865448 on Build GRCh38
rs916234576 Chr.10: 82865529-82865529 on Build GRCh38
rs893905289 Chr.10: 82865278-82865278 on Build GRCh38
rs947734395 Chr.10: 82865544-82865544 on Build GRCh38
rs949142136 Chr.10: 82865633-82865633 on Build GRCh38
rs888616837 Chr.10: 82865398-82865398 on Build GRCh38
rs1046173501 Chr.10: 82865658-82865658 on Build GRCh38
rs771179786 Chr.10: 82865435-82865435 on Build GRCh38
rs982979216 Chr.10: 82865355-82865355 on Build GRCh38
rs371409110 Chr.10: 82865333-82865333 on Build GRCh38
rs936477897 Chr.10: 82865326-82865326 on Build GRCh38
rs771628932 Chr.10: 82865464-82865464 on Build GRCh38
rs760614798 Chr.10: 82865470-82865470 on Build GRCh38
rs759185982 Chr.10: 82865460-82865460 on Build GRCh38
rs925188867 Chr.10: 82865573-82865573 on Build GRCh38

View Full Product Details