Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84197706-84198199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR1
Entrez Gene ID
92211
Gene Name
cadherin related family member 1
Gene Aliases
CORD15, PCDH21, PRCAD, RP65
UniGene
Hs.137556
Gene Chromosome Location
Chr.10: 84194635-84219621 on Build GRCh38
Transcripts
NM_001171971.2 NM_033100.3 XM_011540337.1 XM_011540338.1 XM_011540339.1 XM_011540340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400517 c.304G>C p.D102H Chr.10: 84197792-84197792 on Build GRCh38
6625214 c.343A>C p.N115H Chr.10: 84197831-84197831 on Build GRCh38
1585155 c.316G>T p.A106S Chr.10: 84197804-84197804 on Build GRCh38
298245 c.306T>C p.D102D Chr.10: 84197794-84197794 on Build GRCh38
6625215 c.343A>C p.N115H Chr.10: 84197831-84197831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758613827 Chr.10: 84197906-84197906 on Build GRCh38
rs750669992 Chr.10: 84197789-84197789 on Build GRCh38
rs1010230410 Chr.10: 84197854-84197854 on Build GRCh38
rs930560733 Chr.10: 84197979-84197979 on Build GRCh38
rs770542148 Chr.10: 84197861-84197861 on Build GRCh38
rs778600684 Chr.10: 84197865-84197865 on Build GRCh38
rs770271238 Chr.10: 84197742-84197742 on Build GRCh38
rs770505990 Chr.10: 84197750-84197750 on Build GRCh38
rs969001681 Chr.10: 84197851-84197851 on Build GRCh38
rs751630535 Chr.10: 84197828-84197828 on Build GRCh38
rs775408178 Chr.10: 84197874-84197874 on Build GRCh38
rs537821794 Chr.10: 84197727-84197727 on Build GRCh38
rs953620269 Chr.10: 84198148-84198148 on Build GRCh38
rs556564516 Chr.10: 84197872-84197872 on Build GRCh38
rs571828279 Chr.10: 84197888-84197888 on Build GRCh38
rs140543050 Chr.10: 84198121-84198121 on Build GRCh38
rs747425652 Chr.10: 84197825-84197825 on Build GRCh38
rs755472490 Chr.10: 84197980-84197980 on Build GRCh38
rs962133040 Chr.10: 84197959-84197959 on Build GRCh38
rs370015988 Chr.10: 84197903-84197903 on Build GRCh38
rs551687330 Chr.10: 84197743-84197743 on Build GRCh38
rs748588550 Chr.10: 84197741-84197741 on Build GRCh38
rs533998856 Chr.10: 84197821-84197821 on Build GRCh38
rs904673895 Chr.10: 84198079-84198079 on Build GRCh38
rs760563696 Chr.10: 84197877-84197877 on Build GRCh38
rs763109043 Chr.10: 84197806-84197806 on Build GRCh38
rs937365001 Chr.10: 84198004-84198004 on Build GRCh38
rs537943056 Chr.10: 84197868-84197868 on Build GRCh38
rs542823648 Chr.10: 84197977-84197977 on Build GRCh38
rs1057440025 Chr.10: 84198010-84198010 on Build GRCh38
rs968724888 Chr.10: 84197892-84197892 on Build GRCh38
rs1029237751 Chr.10: 84198147-84198147 on Build GRCh38
rs921620049 Chr.10: 84197866-84197866 on Build GRCh38
rs1020471139 Chr.10: 84198061-84198061 on Build GRCh38
rs987092557 Chr.10: 84197905-84197905 on Build GRCh38
rs561317992 Chr.10: 84198094-84198094 on Build GRCh38
rs372683059 Chr.10: 84197751-84197751 on Build GRCh38
rs567648155 Chr.10: 84197913-84197913 on Build GRCh38
rs1010699322 Chr.10: 84198043-84198043 on Build GRCh38
rs1010439411 Chr.10: 84197791-84197791 on Build GRCh38
rs537000214 Chr.10: 84197843-84197843 on Build GRCh38
rs138798963 Chr.10: 84197738-84197738 on Build GRCh38
rs267602595 Chr.10: 84197834-84197834 on Build GRCh38
rs199864430 Chr.10: 84197760-84197760 on Build GRCh38
rs371416045 Chr.10: 84197764-84197764 on Build GRCh38
rs905093542 Chr.10: 84197988-84197988 on Build GRCh38
rs572359295 Chr.10: 84197940-84197940 on Build GRCh38
rs200333496 Chr.10: 84197761-84197761 on Build GRCh38
rs200738075 Chr.10: 84197924-84197925 on Build GRCh38
rs781607008 Chr.10: 84197838-84197838 on Build GRCh38
rs11200915 Chr.10: 84197925-84197925 on Build GRCh38
rs759068203 Chr.10: 84197757-84197757 on Build GRCh38
rs756554786 Chr.10: 84197846-84197846 on Build GRCh38
rs549505124 Chr.10: 84197856-84197856 on Build GRCh38
rs755080034 Chr.10: 84197835-84197835 on Build GRCh38
rs376315843 Chr.10: 84197847-84197847 on Build GRCh38
rs374945865 Chr.10: 84197767-84197767 on Build GRCh38
rs895910283 Chr.10: 84198013-84198013 on Build GRCh38

View Full Product Details