Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6270915-6271419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCKBR
Entrez Gene ID
887
Gene Name
cholecystokinin B receptor
Gene Aliases
CCK-B, CCK2R, GASR
UniGene
Hs.203
Gene Chromosome Location
Chr.11: 6259611-6272127 on Build GRCh38
Transcripts
NM_001318029.1 NM_176875.3 XM_005253210.1 XM_017018516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319300 c.1136C>T p.T379M Chr.11: 6271128-6271128 on Build GRCh38
6537221 c.838C>G p.R280G Chr.11: 6271037-6271037 on Build GRCh38
3451176 c.844G>A p.E282K Chr.11: 6271043-6271043 on Build GRCh38
6531056 c.1119G>C p.L373L Chr.11: 6271111-6271111 on Build GRCh38
4670166 c.1039C>T p.R347C Chr.11: 6271031-6271031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748178922 Chr.11: 6270973-6270973 on Build GRCh38
rs771124037 Chr.11: 6270984-6270984 on Build GRCh38
rs773727589 Chr.11: 6271152-6271152 on Build GRCh38
rs780808122 Chr.11: 6270971-6270975 on Build GRCh38
rs201958014 Chr.11: 6270989-6270989 on Build GRCh38
rs758665918 Chr.11: 6271177-6271177 on Build GRCh38
rs753116644 Chr.11: 6271386-6271386 on Build GRCh38
rs201876764 Chr.11: 6271347-6271347 on Build GRCh38
rs777883079 Chr.11: 6271052-6271052 on Build GRCh38
rs200339576 Chr.11: 6271086-6271086 on Build GRCh38
rs575229231 Chr.11: 6270966-6270966 on Build GRCh38
rs200973903 Chr.11: 6271056-6271056 on Build GRCh38
rs540451185 Chr.11: 6271071-6271071 on Build GRCh38
rs750156310 Chr.11: 6270936-6270936 on Build GRCh38
rs758929674 Chr.11: 6271215-6271215 on Build GRCh38
rs1805001 Chr.11: 6271155-6271155 on Build GRCh38
rs777122005 Chr.11: 6271193-6271193 on Build GRCh38
rs145637819 Chr.11: 6271327-6271327 on Build GRCh38
rs556976466 Chr.11: 6270949-6270949 on Build GRCh38
rs200480495 Chr.11: 6271383-6271383 on Build GRCh38
rs914042542 Chr.11: 6271298-6271298 on Build GRCh38
rs774873372 Chr.11: 6271264-6271265 on Build GRCh38
rs201721347 Chr.11: 6271385-6271385 on Build GRCh38
rs918426340 Chr.11: 6271260-6271260 on Build GRCh38
rs749320440 Chr.11: 6271053-6271053 on Build GRCh38
rs201626406 Chr.11: 6271140-6271140 on Build GRCh38
rs138218747 Chr.11: 6271339-6271339 on Build GRCh38
rs753601546 Chr.11: 6271226-6271226 on Build GRCh38
rs755275816 Chr.11: 6271380-6271380 on Build GRCh38
rs754731863 Chr.11: 6271037-6271037 on Build GRCh38
rs745569675 Chr.11: 6271121-6271122 on Build GRCh38
rs535599390 Chr.11: 6271267-6271267 on Build GRCh38
rs201854899 Chr.11: 6271204-6271204 on Build GRCh38
rs957774181 Chr.11: 6271198-6271198 on Build GRCh38
rs767203414 Chr.11: 6271216-6271216 on Build GRCh38
rs190601811 Chr.11: 6271219-6271219 on Build GRCh38
rs199552178 Chr.11: 6271129-6271129 on Build GRCh38
rs747287727 Chr.11: 6271137-6271137 on Build GRCh38
rs201162773 Chr.11: 6271150-6271150 on Build GRCh38
rs201338739 Chr.11: 6271362-6271362 on Build GRCh38
rs201237450 Chr.11: 6271136-6271136 on Build GRCh38
rs199930350 Chr.11: 6271011-6271011 on Build GRCh38
rs756376172 Chr.11: 6271394-6271394 on Build GRCh38
rs993321475 Chr.11: 6271400-6271400 on Build GRCh38
rs199874559 Chr.11: 6271213-6271213 on Build GRCh38
rs748652241 Chr.11: 6271391-6271393 on Build GRCh38
rs781003043 Chr.11: 6271039-6271039 on Build GRCh38
rs765220669 Chr.11: 6271344-6271344 on Build GRCh38
rs762508957 Chr.11: 6271075-6271075 on Build GRCh38
rs780352247 Chr.11: 6271272-6271272 on Build GRCh38
rs751887877 Chr.11: 6271264-6271264 on Build GRCh38
rs376332235 Chr.11: 6271128-6271128 on Build GRCh38
rs113168010 Chr.11: 6271154-6271154 on Build GRCh38
rs771833429 Chr.11: 6271302-6271302 on Build GRCh38
rs145064936 Chr.11: 6271397-6271397 on Build GRCh38
rs557999450 Chr.11: 6271034-6271034 on Build GRCh38
rs140350741 Chr.11: 6271014-6271014 on Build GRCh38
rs778076555 Chr.11: 6271292-6271292 on Build GRCh38
rs777652032 Chr.11: 6271102-6271102 on Build GRCh38
rs749374907 Chr.11: 6271103-6271103 on Build GRCh38
rs544323198 Chr.11: 6271148-6271148 on Build GRCh38
rs141279039 Chr.11: 6271357-6271357 on Build GRCh38
rs199896310 Chr.11: 6271331-6271331 on Build GRCh38
rs771543762 Chr.11: 6271300-6271300 on Build GRCh38
rs201659480 Chr.11: 6270997-6270997 on Build GRCh38
rs748172133 Chr.11: 6271045-6271045 on Build GRCh38
rs75576436 Chr.11: 6271220-6271220 on Build GRCh38
rs765032135 Chr.11: 6271254-6271254 on Build GRCh38
rs200865839 Chr.11: 6271051-6271051 on Build GRCh38
rs201036187 Chr.11: 6271023-6271023 on Build GRCh38
rs1042045 Chr.11: 6271135-6271135 on Build GRCh38
rs200736965 Chr.11: 6271172-6271172 on Build GRCh38
rs779087210 Chr.11: 6271169-6271169 on Build GRCh38
rs762933442 Chr.11: 6271358-6271358 on Build GRCh38
rs367917726 Chr.11: 6271021-6271021 on Build GRCh38
rs199633246 Chr.11: 6271163-6271163 on Build GRCh38
rs200919066 Chr.11: 6271340-6271340 on Build GRCh38
rs201953391 Chr.11: 6271228-6271228 on Build GRCh38
rs772679349 Chr.11: 6271396-6271404 on Build GRCh38
rs202204639 Chr.11: 6271119-6271119 on Build GRCh38
rs745396843 Chr.11: 6271295-6271295 on Build GRCh38
rs755245323 Chr.11: 6271265-6271265 on Build GRCh38
rs965159112 Chr.11: 6271249-6271249 on Build GRCh38
rs756071960 Chr.11: 6271101-6271101 on Build GRCh38
rs758118124 Chr.11: 6270944-6270944 on Build GRCh38
rs200202419 Chr.11: 6271060-6271060 on Build GRCh38
rs149146333 Chr.11: 6270983-6270983 on Build GRCh38
rs752415795 Chr.11: 6271097-6271097 on Build GRCh38
rs767387348 Chr.11: 6271087-6271087 on Build GRCh38
rs953750132 Chr.11: 6271188-6271188 on Build GRCh38
rs144317623 Chr.11: 6271031-6271031 on Build GRCh38
rs989706426 Chr.11: 6270999-6270999 on Build GRCh38
rs751163301 Chr.11: 6271083-6271083 on Build GRCh38
rs202141196 Chr.11: 6271038-6271038 on Build GRCh38
rs200262698 Chr.11: 6271143-6271143 on Build GRCh38
rs760363464 Chr.11: 6271318-6271318 on Build GRCh38
rs371750197 Chr.11: 6271064-6271064 on Build GRCh38
rs748729612 Chr.11: 6271210-6271210 on Build GRCh38
rs759831741 Chr.11: 6271059-6271059 on Build GRCh38
rs200543193 Chr.11: 6271225-6271225 on Build GRCh38
rs766435444 Chr.11: 6271359-6271359 on Build GRCh38
rs1017770527 Chr.11: 6271196-6271196 on Build GRCh38
rs766130597 Chr.11: 6271081-6271081 on Build GRCh38
rs145313566 Chr.11: 6271032-6271032 on Build GRCh38
rs773278468 Chr.11: 6270979-6270979 on Build GRCh38
rs200976920 Chr.11: 6271066-6271066 on Build GRCh38
rs958277749 Chr.11: 6271153-6271153 on Build GRCh38
rs140742189 Chr.11: 6271257-6271257 on Build GRCh38
rs757091759 Chr.11: 6270965-6270966 on Build GRCh38
rs773936454 Chr.11: 6271214-6271214 on Build GRCh38
rs201673722 Chr.11: 6271159-6271159 on Build GRCh38
rs749921079 Chr.11: 6271027-6271027 on Build GRCh38
rs937985143 Chr.11: 6271338-6271338 on Build GRCh38
rs200097559 Chr.11: 6271261-6271261 on Build GRCh38
rs897640161 Chr.11: 6271392-6271392 on Build GRCh38
rs910332356 Chr.11: 6270975-6270975 on Build GRCh38
rs755843090 Chr.11: 6271161-6271161 on Build GRCh38
rs200384464 Chr.11: 6271356-6271356 on Build GRCh38
rs149197660 Chr.11: 6271054-6271054 on Build GRCh38
rs1046862255 Chr.11: 6271348-6271348 on Build GRCh38
rs185405927 Chr.11: 6271185-6271185 on Build GRCh38
rs777207956 Chr.11: 6271147-6271147 on Build GRCh38
rs201374522 Chr.11: 6271157-6271157 on Build GRCh38
rs200700735 Chr.11: 6271186-6271186 on Build GRCh38
rs757183608 Chr.11: 6271166-6271166 on Build GRCh38
rs767915510 Chr.11: 6271001-6271001 on Build GRCh38
rs764032535 Chr.11: 6271162-6271162 on Build GRCh38
rs780033579 Chr.11: 6270945-6270945 on Build GRCh38
rs200830731 Chr.11: 6270992-6270992 on Build GRCh38
rs758173202 Chr.11: 6271029-6271029 on Build GRCh38
rs200060380 Chr.11: 6270991-6270991 on Build GRCh38
rs151322840 Chr.11: 6271164-6271164 on Build GRCh38
rs202203262 Chr.11: 6271376-6271376 on Build GRCh38
rs769083241 Chr.11: 6271189-6271189 on Build GRCh38
rs201375824 Chr.11: 6271309-6271309 on Build GRCh38
rs761785922 Chr.11: 6271240-6271240 on Build GRCh38
rs142419010 Chr.11: 6271112-6271112 on Build GRCh38
rs748476238 Chr.11: 6271275-6271275 on Build GRCh38
rs770366601 Chr.11: 6271279-6271279 on Build GRCh38
rs753191090 Chr.11: 6271006-6271006 on Build GRCh38
rs769278521 Chr.11: 6271220-6271221 on Build GRCh38
rs747382508 Chr.11: 6271187-6271187 on Build GRCh38

View Full Product Details