Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34658443-34658946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHF
Entrez Gene ID
26298
Gene Name
ETS homologous factor
Gene Aliases
ESE3, ESE3B, ESEJ
UniGene
Hs.653859
Gene Chromosome Location
Chr.11: 34621041-34663288 on Build GRCh38
Transcripts
NM_001206615.1 NM_001206616.1 NM_012153.5 XM_005252857.1 XM_005252860.1 XM_005252861.1 XM_005252862.1 XM_011519983.1 XM_011519984.1 XM_011519985.1 XM_017017534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238761 c.640C>T p.R214C Chr.11: 34658565-34658565 on Build GRCh38
6551912 c.878G>T p.R293L Chr.11: 34658906-34658906 on Build GRCh38
4333269 c.794G>A p.R265Q Chr.11: 34658719-34658719 on Build GRCh38
1205082 c.832C>T p.R278C Chr.11: 34658860-34658860 on Build GRCh38
4416977 c.825T>G p.I275M Chr.11: 34658853-34658853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199838917 Chr.11: 34658631-34658631 on Build GRCh38
rs758960350 Chr.11: 34658742-34658742 on Build GRCh38
rs770260609 Chr.11: 34658510-34658510 on Build GRCh38
rs763605744 Chr.11: 34658535-34658535 on Build GRCh38
rs147664541 Chr.11: 34658736-34658736 on Build GRCh38
rs778842956 Chr.11: 34658623-34658623 on Build GRCh38
rs569349541 Chr.11: 34658567-34658567 on Build GRCh38
rs79800810 Chr.11: 34658551-34658551 on Build GRCh38
rs771519966 Chr.11: 34658711-34658711 on Build GRCh38
rs3818514 Chr.11: 34658506-34658506 on Build GRCh38
rs368391797 Chr.11: 34658813-34658813 on Build GRCh38
rs950653325 Chr.11: 34658537-34658537 on Build GRCh38
rs747121315 Chr.11: 34658490-34658490 on Build GRCh38
rs201722091 Chr.11: 34658562-34658562 on Build GRCh38
rs746432576 Chr.11: 34658863-34658863 on Build GRCh38
rs550910897 Chr.11: 34658513-34658513 on Build GRCh38
rs994665794 Chr.11: 34658529-34658529 on Build GRCh38
rs1027549870 Chr.11: 34658534-34658534 on Build GRCh38
rs1055880629 Chr.11: 34658498-34658498 on Build GRCh38
rs760123736 Chr.11: 34658906-34658906 on Build GRCh38
rs371615222 Chr.11: 34658797-34658797 on Build GRCh38
rs760413147 Chr.11: 34658750-34658750 on Build GRCh38
rs568623948 Chr.11: 34658796-34658796 on Build GRCh38
rs759142735 Chr.11: 34658566-34658566 on Build GRCh38
rs757056736 Chr.11: 34658790-34658790 on Build GRCh38
rs775169543 Chr.11: 34658861-34658861 on Build GRCh38
rs77580726 Chr.11: 34658553-34658553 on Build GRCh38
rs532117870 Chr.11: 34658620-34658620 on Build GRCh38
rs1053012686 Chr.11: 34658812-34658812 on Build GRCh38
rs761591997 Chr.11: 34658917-34658917 on Build GRCh38
rs752457173 Chr.11: 34658581-34658581 on Build GRCh38
rs368425107 Chr.11: 34658502-34658502 on Build GRCh38
rs749279269 Chr.11: 34658483-34658483 on Build GRCh38
rs777562842 Chr.11: 34658482-34658482 on Build GRCh38
rs948397964 Chr.11: 34658747-34658747 on Build GRCh38
rs1004729806 Chr.11: 34658548-34658548 on Build GRCh38
rs200647493 Chr.11: 34658803-34658803 on Build GRCh38
rs747269254 Chr.11: 34658504-34658506 on Build GRCh38
rs765958099 Chr.11: 34658536-34658536 on Build GRCh38
rs201673153 Chr.11: 34658669-34658669 on Build GRCh38
rs781454395 Chr.11: 34658754-34658754 on Build GRCh38
rs981064678 Chr.11: 34658761-34658761 on Build GRCh38
rs781267458 Chr.11: 34658645-34658645 on Build GRCh38
rs768710849 Chr.11: 34658491-34658491 on Build GRCh38
rs201700802 Chr.11: 34658880-34658880 on Build GRCh38
rs139008764 Chr.11: 34658859-34658859 on Build GRCh38
rs768238883 Chr.11: 34658907-34658907 on Build GRCh38
rs141336422 Chr.11: 34658568-34658568 on Build GRCh38
rs1018573848 Chr.11: 34658570-34658570 on Build GRCh38
rs763995950 Chr.11: 34658609-34658609 on Build GRCh38
rs368204105 Chr.11: 34658708-34658708 on Build GRCh38
rs770034763 Chr.11: 34658680-34658680 on Build GRCh38
rs1002027388 Chr.11: 34658497-34658497 on Build GRCh38
rs767109210 Chr.11: 34658748-34658748 on Build GRCh38
rs376033943 Chr.11: 34658622-34658622 on Build GRCh38
rs549658997 Chr.11: 34658514-34658514 on Build GRCh38
rs755844845 Chr.11: 34658595-34658595 on Build GRCh38
rs184420633 Chr.11: 34658767-34658767 on Build GRCh38
rs375639873 Chr.11: 34658875-34658875 on Build GRCh38
rs192787555 Chr.11: 34658487-34658487 on Build GRCh38
rs374420735 Chr.11: 34658489-34658489 on Build GRCh38
rs776269897 Chr.11: 34658910-34658910 on Build GRCh38
rs774981383 Chr.11: 34658749-34658749 on Build GRCh38
rs756322371 Chr.11: 34658838-34658838 on Build GRCh38
rs747033466 Chr.11: 34658630-34658630 on Build GRCh38
rs974266726 Chr.11: 34658608-34658608 on Build GRCh38
rs775022168 Chr.11: 34658878-34658878 on Build GRCh38
rs752832931 Chr.11: 34658805-34658805 on Build GRCh38
rs776494379 Chr.11: 34658689-34658689 on Build GRCh38
rs286891 Chr.11: 34658853-34658853 on Build GRCh38
rs757126229 Chr.11: 34658719-34658719 on Build GRCh38
rs1045781764 Chr.11: 34658470-34658470 on Build GRCh38
rs773755546 Chr.11: 34658692-34658692 on Build GRCh38
rs1017671285 Chr.11: 34658476-34658476 on Build GRCh38
rs753579116 Chr.11: 34658769-34658769 on Build GRCh38
rs757363463 Chr.11: 34658522-34658522 on Build GRCh38

View Full Product Details