Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46382211-46382704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDK
Entrez Gene ID
4192
Gene Name
midkine (neurite growth-promoting factor 2)
Gene Aliases
ARAP, MK, NEGF2
UniGene
Hs.82045
Gene Chromosome Location
Chr.11: 46380784-46383837 on Build GRCh38
Transcripts
NM_001012333.2 NM_001012334.2 NM_001270550.1 NM_001270551.1 NM_001270552.1 NM_002391.4 NR_073039.1 XM_011520116.2 XM_017017764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050831 c.279G>A p.A93A Chr.11: 46382621-46382621 on Build GRCh38
5372502 c.304G>A p.V102I Chr.11: 46382646-46382646 on Build GRCh38
4842845 c.106G>C p.E36Q Chr.11: 46382323-46382323 on Build GRCh38
7376733 c.94G>T p.G32C Chr.11: 46382311-46382311 on Build GRCh38
3869187 c.128G>A p.G43E Chr.11: 46382345-46382345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533427797 Chr.11: 46382323-46382323 on Build GRCh38
rs535929130 Chr.11: 46382411-46382411 on Build GRCh38
rs549989631 Chr.11: 46382307-46382307 on Build GRCh38
rs971229303 Chr.11: 46382581-46382581 on Build GRCh38
rs1020019180 Chr.11: 46382516-46382516 on Build GRCh38
rs767636741 Chr.11: 46382647-46382647 on Build GRCh38
rs754744084 Chr.11: 46382255-46382255 on Build GRCh38
rs201944015 Chr.11: 46382633-46382633 on Build GRCh38
rs766378741 Chr.11: 46382250-46382250 on Build GRCh38
rs774088935 Chr.11: 46382593-46382593 on Build GRCh38
rs755127433 Chr.11: 46382549-46382549 on Build GRCh38
rs756667060 Chr.11: 46382342-46382342 on Build GRCh38
rs760416643 Chr.11: 46382495-46382495 on Build GRCh38
rs375682194 Chr.11: 46382574-46382574 on Build GRCh38
rs771419039 Chr.11: 46382677-46382677 on Build GRCh38
rs373502349 Chr.11: 46382331-46382331 on Build GRCh38
rs781748273 Chr.11: 46382423-46382423 on Build GRCh38
rs752407888 Chr.11: 46382355-46382355 on Build GRCh38
rs145465168 Chr.11: 46382663-46382663 on Build GRCh38
rs1004989965 Chr.11: 46382346-46382346 on Build GRCh38
rs766682049 Chr.11: 46382510-46382510 on Build GRCh38
rs879050410 Chr.11: 46382463-46382463 on Build GRCh38
rs938231531 Chr.11: 46382636-46382636 on Build GRCh38
rs202080796 Chr.11: 46382279-46382279 on Build GRCh38
rs758418356 Chr.11: 46382400-46382400 on Build GRCh38
rs745914347 Chr.11: 46382274-46382274 on Build GRCh38
rs879134006 Chr.11: 46382462-46382462 on Build GRCh38
rs758618039 Chr.11: 46382505-46382505 on Build GRCh38
rs780996116 Chr.11: 46382405-46382405 on Build GRCh38
rs967091312 Chr.11: 46382254-46382254 on Build GRCh38
rs200063024 Chr.11: 46382392-46382392 on Build GRCh38
rs771631882 Chr.11: 46382612-46382612 on Build GRCh38
rs751569691 Chr.11: 46382539-46382539 on Build GRCh38
rs778278101 Chr.11: 46382578-46382578 on Build GRCh38
rs775434897 Chr.11: 46382494-46382494 on Build GRCh38
rs762326985 Chr.11: 46382293-46382293 on Build GRCh38
rs750669640 Chr.11: 46382374-46382374 on Build GRCh38
rs995590952 Chr.11: 46382412-46382412 on Build GRCh38
rs7481848 Chr.11: 46382319-46382319 on Build GRCh38
rs779063659 Chr.11: 46382358-46382358 on Build GRCh38
rs1007467012 Chr.11: 46382553-46382553 on Build GRCh38
rs749727193 Chr.11: 46382583-46382583 on Build GRCh38
rs760612922 Chr.11: 46382616-46382616 on Build GRCh38
rs370175046 Chr.11: 46382452-46382452 on Build GRCh38
rs751840359 Chr.11: 46382632-46382632 on Build GRCh38
rs748676546 Chr.11: 46382573-46382573 on Build GRCh38
rs748494356 Chr.11: 46382428-46382428 on Build GRCh38
rs766479252 Chr.11: 46382631-46382631 on Build GRCh38
rs769018894 Chr.11: 46382575-46382576 on Build GRCh38
rs747672612 Chr.11: 46382301-46382303 on Build GRCh38
rs112695639 Chr.11: 46382387-46382387 on Build GRCh38
rs899387472 Chr.11: 46382393-46382393 on Build GRCh38
rs761717296 Chr.11: 46382627-46382627 on Build GRCh38
rs879159016 Chr.11: 46382291-46382291 on Build GRCh38
rs759131374 Chr.11: 46382471-46382471 on Build GRCh38
rs377442266 Chr.11: 46382448-46382448 on Build GRCh38
rs762502808 Chr.11: 46382315-46382315 on Build GRCh38
rs184181824 Chr.11: 46382235-46382235 on Build GRCh38
rs745326945 Chr.11: 46382598-46382598 on Build GRCh38
rs767544613 Chr.11: 46382348-46382348 on Build GRCh38
rs759385996 Chr.11: 46382336-46382336 on Build GRCh38
rs749634952 Chr.11: 46382665-46382665 on Build GRCh38
rs769002341 Chr.11: 46382283-46382283 on Build GRCh38
rs942114318 Chr.11: 46382380-46382380 on Build GRCh38
rs765998798 Chr.11: 46382330-46382330 on Build GRCh38
rs988636977 Chr.11: 46382534-46382534 on Build GRCh38
rs746514716 Chr.11: 46382276-46382283 on Build GRCh38
rs769549708 Chr.11: 46382298-46382298 on Build GRCh38
rs770303856 Chr.11: 46382431-46382431 on Build GRCh38
rs751472591 Chr.11: 46382253-46382253 on Build GRCh38
rs953320775 Chr.11: 46382543-46382543 on Build GRCh38
rs748952493 Chr.11: 46382442-46382442 on Build GRCh38
rs772939800 Chr.11: 46382436-46382436 on Build GRCh38
rs971890724 Chr.11: 46382577-46382577 on Build GRCh38
rs372172569 Chr.11: 46382260-46382260 on Build GRCh38
rs745549795 Chr.11: 46382674-46382674 on Build GRCh38
rs770464409 Chr.11: 46382588-46382588 on Build GRCh38
rs756675404 Chr.11: 46382652-46382652 on Build GRCh38
rs373593241 Chr.11: 46382492-46382492 on Build GRCh38
rs558385181 Chr.11: 46382672-46382672 on Build GRCh38
rs1019947035 Chr.11: 46382245-46382245 on Build GRCh38
rs61731885 Chr.11: 46382357-46382357 on Build GRCh38
rs926649994 Chr.11: 46382587-46382587 on Build GRCh38
rs1027952408 Chr.11: 46382493-46382493 on Build GRCh38
rs779153713 Chr.11: 46382264-46382264 on Build GRCh38
rs776856042 Chr.11: 46382284-46382284 on Build GRCh38
rs982131054 Chr.11: 46382586-46382586 on Build GRCh38
rs997155538 Chr.11: 46382523-46382523 on Build GRCh38
rs552093574 Chr.11: 46382499-46382499 on Build GRCh38
rs947290562 Chr.11: 46382546-46382546 on Build GRCh38
rs764089666 Chr.11: 46382621-46382621 on Build GRCh38
rs774453078 Chr.11: 46382593-46382594 on Build GRCh38
rs757669386 Chr.11: 46382669-46382669 on Build GRCh38
rs919242088 Chr.11: 46382422-46382422 on Build GRCh38
rs749651858 Chr.11: 46382568-46382569 on Build GRCh38
rs78369317 Chr.11: 46382524-46382524 on Build GRCh38
rs955879945 Chr.11: 46382533-46382533 on Build GRCh38
rs757452961 Chr.11: 46382262-46382262 on Build GRCh38
rs902943489 Chr.11: 46382519-46382519 on Build GRCh38
rs915512883 Chr.11: 46382673-46382673 on Build GRCh38
rs769031420 Chr.11: 46382416-46382416 on Build GRCh38
rs781513243 Chr.11: 46382561-46382561 on Build GRCh38
rs767324227 Chr.11: 46382472-46382472 on Build GRCh38
rs754954347 Chr.11: 46382640-46382640 on Build GRCh38
rs772174456 Chr.11: 46382277-46382277 on Build GRCh38
rs866043166 Chr.11: 46382594-46382594 on Build GRCh38
rs752891556 Chr.11: 46382649-46382649 on Build GRCh38
rs780586599 Chr.11: 46382551-46382581 on Build GRCh38

View Full Product Details