Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34607978-34608545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
UniGene
Hs.356647
Gene Chromosome Location
Chr.14: 34561409-34630160 on Build GRCh38
Transcripts
NM_021249.3 NM_152233.2 XM_006720224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6435525 c.222C>A p.N74K Chr.14: 34608114-34608114 on Build GRCh38
3711497 c.263G>T p.G88V Chr.14: 34608037-34608037 on Build GRCh38
6367518 c.260C>T p.A87V Chr.14: 34608040-34608040 on Build GRCh38
433030 c.194C>T p.S65L Chr.14: 34608106-34608106 on Build GRCh38
3495801 c.270C>T p.I90I Chr.14: 34608030-34608030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552329633 Chr.14: 34608505-34608505 on Build GRCh38
rs1023669040 Chr.14: 34608149-34608149 on Build GRCh38
rs760125606 Chr.14: 34608021-34608021 on Build GRCh38
rs753435792 Chr.14: 34608027-34608027 on Build GRCh38
rs775988867 Chr.14: 34608148-34608148 on Build GRCh38
rs555202849 Chr.14: 34608229-34608229 on Build GRCh38
rs758232592 Chr.14: 34608049-34608049 on Build GRCh38
rs542014445 Chr.14: 34608236-34608236 on Build GRCh38
rs956641307 Chr.14: 34608303-34608303 on Build GRCh38
rs147318968 Chr.14: 34608247-34608247 on Build GRCh38
rs751714642 Chr.14: 34608185-34608186 on Build GRCh38
rs981591025 Chr.14: 34608389-34608389 on Build GRCh38
rs763600661 Chr.14: 34608023-34608023 on Build GRCh38
rs908417821 Chr.14: 34608325-34608325 on Build GRCh38
rs988756622 Chr.14: 34608313-34608313 on Build GRCh38
rs1029233949 Chr.14: 34608234-34608234 on Build GRCh38
rs756155484 Chr.14: 34608111-34608111 on Build GRCh38
rs756779754 Chr.14: 34608423-34608423 on Build GRCh38
rs546295977 Chr.14: 34608150-34608150 on Build GRCh38
rs369985339 Chr.14: 34608414-34608414 on Build GRCh38
rs751295409 Chr.14: 34608046-34608046 on Build GRCh38
rs765031095 Chr.14: 34608443-34608443 on Build GRCh38
rs997303222 Chr.14: 34608231-34608231 on Build GRCh38
rs1044087885 Chr.14: 34608486-34608486 on Build GRCh38
rs772373075 Chr.14: 34608139-34608139 on Build GRCh38
rs764967493 Chr.14: 34608032-34608032 on Build GRCh38
rs762859145 Chr.14: 34608185-34608185 on Build GRCh38
rs377266343 Chr.14: 34608105-34608105 on Build GRCh38
rs772955829 Chr.14: 34608183-34608183 on Build GRCh38
rs550946721 Chr.14: 34608302-34608302 on Build GRCh38
rs780166850 Chr.14: 34608097-34608097 on Build GRCh38
rs913262883 Chr.14: 34608361-34608361 on Build GRCh38
rs953177071 Chr.14: 34608246-34608246 on Build GRCh38
rs1004520181 Chr.14: 34608179-34608179 on Build GRCh38
rs116322269 Chr.14: 34608189-34608189 on Build GRCh38
rs755367176 Chr.14: 34608458-34608458 on Build GRCh38
rs1050148961 Chr.14: 34608488-34608488 on Build GRCh38
rs531998252 Chr.14: 34608454-34608454 on Build GRCh38
rs921151313 Chr.14: 34608301-34608301 on Build GRCh38
rs1012970156 Chr.14: 34608455-34608455 on Build GRCh38
rs370482210 Chr.14: 34608114-34608114 on Build GRCh38
rs531244245 Chr.14: 34608259-34608259 on Build GRCh38
rs141847426 Chr.14: 34608153-34608153 on Build GRCh38
rs1048669217 Chr.14: 34608307-34608307 on Build GRCh38
rs997671766 Chr.14: 34608464-34608464 on Build GRCh38
rs775935774 Chr.14: 34608012-34608012 on Build GRCh38
rs975342827 Chr.14: 34608293-34608293 on Build GRCh38
rs1056834332 Chr.14: 34608418-34608418 on Build GRCh38
rs761050264 Chr.14: 34608039-34608039 on Build GRCh38
rs761202673 Chr.14: 34608160-34608160 on Build GRCh38
rs940944787 Chr.14: 34608445-34608445 on Build GRCh38
rs772608847 Chr.14: 34608005-34608005 on Build GRCh38
rs374118473 Chr.14: 34608128-34608128 on Build GRCh38
rs964729306 Chr.14: 34608292-34608292 on Build GRCh38
rs746263408 Chr.14: 34608135-34608135 on Build GRCh38
rs927370510 Chr.14: 34608400-34608400 on Build GRCh38
rs765018579 Chr.14: 34608167-34608167 on Build GRCh38
rs776259372 Chr.14: 34608159-34608159 on Build GRCh38
rs911891471 Chr.14: 34608260-34608260 on Build GRCh38
rs971881788 Chr.14: 34608383-34608383 on Build GRCh38
rs141002693 Chr.14: 34608453-34608453 on Build GRCh38
rs777978976 Chr.14: 34608113-34608113 on Build GRCh38
rs1049269384 Chr.14: 34608415-34608415 on Build GRCh38
rs749317511 Chr.14: 34608118-34608118 on Build GRCh38
rs766213457 Chr.14: 34608187-34608187 on Build GRCh38
rs771193546 Chr.14: 34608120-34608120 on Build GRCh38
rs888879348 Chr.14: 34608514-34608514 on Build GRCh38
rs577143688 Chr.14: 34608030-34608030 on Build GRCh38

View Full Product Details