Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118530550-118531050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929089
Entrez Gene ID
101929089
Gene Name
uncharacterized LOC101929089
Gene Aliases
-
UniGene
Hs.666633
Gene Chromosome Location
Chr.11: 118511918-118531094 on Build GRCh38
Transcripts
NR_120572.1 NR_120573.1 NR_120574.1 NR_120575.1 NR_120576.1 NM_001080441.1

Gene Symbol
TTC36
Entrez Gene ID
143941
Gene Name
tetratricopeptide repeat domain 36
Gene Aliases
HBP21
UniGene
Hs.653449
Gene Chromosome Location
Chr.11: 118527495-118531025 on Build GRCh38
Transcripts
NR_120572.1 NR_120573.1 NR_120574.1 NR_120575.1 NR_120576.1 NM_001080441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6021025 c.353G>A p.R118Q Chr.11: 118530699-118530699 on Build GRCh38
7412858 c.534G>A p.M178I Chr.11: 118530880-118530880 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782530484 Chr.11: 118530724-118530724 on Build GRCh38
rs782773703 Chr.11: 118530737-118530737 on Build GRCh38
rs1010721417 Chr.11: 118530630-118530630 on Build GRCh38
rs552582178 Chr.11: 118530695-118530695 on Build GRCh38
rs782739514 Chr.11: 118530675-118530675 on Build GRCh38
rs928994472 Chr.11: 118530572-118530572 on Build GRCh38
rs1038788652 Chr.11: 118530616-118530616 on Build GRCh38
rs781902931 Chr.11: 118530703-118530703 on Build GRCh38
rs781861285 Chr.11: 118530756-118530756 on Build GRCh38
rs973944908 Chr.11: 118530741-118530741 on Build GRCh38
rs371638343 Chr.11: 118530814-118530814 on Build GRCh38
rs564421103 Chr.11: 118530809-118530809 on Build GRCh38
rs900361874 Chr.11: 118530629-118530629 on Build GRCh38
rs1043912927 Chr.11: 118530902-118530902 on Build GRCh38
rs548111134 Chr.11: 118530607-118530607 on Build GRCh38
rs970211856 Chr.11: 118530720-118530720 on Build GRCh38
rs929586914 Chr.11: 118530757-118530757 on Build GRCh38
rs1047808834 Chr.11: 118530582-118530582 on Build GRCh38
rs782394025 Chr.11: 118530854-118530854 on Build GRCh38
rs782681179 Chr.11: 118530752-118530752 on Build GRCh38
rs900224666 Chr.11: 118530878-118530878 on Build GRCh38
rs1022226579 Chr.11: 118530664-118530664 on Build GRCh38
rs782704541 Chr.11: 118530698-118530698 on Build GRCh38
rs782129430 Chr.11: 118530637-118530637 on Build GRCh38
rs999876849 Chr.11: 118530931-118530931 on Build GRCh38
rs549927235 Chr.11: 118530936-118530936 on Build GRCh38
rs782684650 Chr.11: 118530827-118530827 on Build GRCh38
rs782443327 Chr.11: 118530825-118530825 on Build GRCh38
rs911590087 Chr.11: 118530778-118530778 on Build GRCh38
rs1038821306 Chr.11: 118530848-118530848 on Build GRCh38
rs782066530 Chr.11: 118530680-118530680 on Build GRCh38
rs781971818 Chr.11: 118530879-118530879 on Build GRCh38
rs1010748681 Chr.11: 118530888-118530888 on Build GRCh38
rs781798700 Chr.11: 118530575-118530575 on Build GRCh38
rs782352035 Chr.11: 118530808-118530808 on Build GRCh38
rs140403049 Chr.11: 118530841-118530841 on Build GRCh38
rs1034880346 Chr.11: 118530948-118530948 on Build GRCh38
rs187027064 Chr.11: 118530894-118530894 on Build GRCh38
rs375349391 Chr.11: 118530774-118530774 on Build GRCh38
rs890122754 Chr.11: 118530593-118530593 on Build GRCh38
rs915971323 Chr.11: 118530699-118530699 on Build GRCh38
rs11600045 Chr.11: 118530668-118530668 on Build GRCh38
rs782599969 Chr.11: 118530794-118530794 on Build GRCh38
rs145595390 Chr.11: 118531029-118531029 on Build GRCh38
rs960047127 Chr.11: 118531012-118531012 on Build GRCh38
rs568336829 Chr.11: 118531024-118531024 on Build GRCh38
rs966997374 Chr.11: 118530671-118530671 on Build GRCh38
rs902344768 Chr.11: 118530922-118530922 on Build GRCh38

View Full Product Details