Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122979314-122979804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGFOD2
Entrez Gene ID
79676
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 2
Gene Aliases
-
UniGene
Hs.524817
Gene Chromosome Location
Chr.12: 122974703-122980041 on Build GRCh38
Transcripts
NM_001304833.1 NM_001304834.1 NM_001304835.1 NM_001304836.1 NM_001304837.1 NM_001304838.1 NM_024623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5840420 c.1021C>T p.P341S Chr.12: 122979314-122979314 on Build GRCh38
6733101 c.1023C>T p.P341P Chr.12: 122979316-122979316 on Build GRCh38
2225589 c.1030G>A p.V344M Chr.12: 122979323-122979323 on Build GRCh38
936370 c.848C>T p.T283M Chr.12: 122979321-122979321 on Build GRCh38
1196569 c.850G>A p.V284M Chr.12: 122979323-122979323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002151142 Chr.12: 122979743-122979743 on Build GRCh38
rs552268277 Chr.12: 122979672-122979672 on Build GRCh38
rs571243415 Chr.12: 122979773-122979773 on Build GRCh38
rs920628398 Chr.12: 122979608-122979608 on Build GRCh38
rs540643937 Chr.12: 122979626-122979626 on Build GRCh38
rs61546241 Chr.12: 122979452-122979452 on Build GRCh38
rs986963424 Chr.12: 122979490-122979490 on Build GRCh38
rs777852422 Chr.12: 122979336-122979336 on Build GRCh38
rs925285470 Chr.12: 122979604-122979604 on Build GRCh38
rs949179306 Chr.12: 122979717-122979717 on Build GRCh38
rs987890397 Chr.12: 122979470-122979470 on Build GRCh38
rs772855464 Chr.12: 122979366-122979366 on Build GRCh38
rs775961876 Chr.12: 122979355-122979355 on Build GRCh38
rs768536413 Chr.12: 122979495-122979495 on Build GRCh38
rs955619351 Chr.12: 122979487-122979487 on Build GRCh38
rs375318596 Chr.12: 122979337-122979337 on Build GRCh38
rs987602407 Chr.12: 122979526-122979526 on Build GRCh38
rs557570566 Chr.12: 122979392-122979392 on Build GRCh38
rs181026664 Chr.12: 122979715-122979715 on Build GRCh38
rs964379482 Chr.12: 122979441-122979441 on Build GRCh38
rs758362986 Chr.12: 122979396-122979396 on Build GRCh38
rs369470455 Chr.12: 122979373-122979373 on Build GRCh38
rs34436013 Chr.12: 122979345-122979346 on Build GRCh38
rs759195482 Chr.12: 122979382-122979382 on Build GRCh38
rs528460680 Chr.12: 122979589-122979589 on Build GRCh38
rs189082092 Chr.12: 122979361-122979361 on Build GRCh38
rs373405921 Chr.12: 122979356-122979356 on Build GRCh38
rs753427371 Chr.12: 122979741-122979741 on Build GRCh38
rs764991120 Chr.12: 122979383-122979383 on Build GRCh38
rs1016360202 Chr.12: 122979425-122979425 on Build GRCh38
rs893507043 Chr.12: 122979777-122979777 on Build GRCh38
rs752439477 Chr.12: 122979391-122979391 on Build GRCh38
rs185662215 Chr.12: 122979728-122979728 on Build GRCh38
rs1032422368 Chr.12: 122979747-122979747 on Build GRCh38
rs896440755 Chr.12: 122979657-122979657 on Build GRCh38
rs914065101 Chr.12: 122979478-122979478 on Build GRCh38
rs3759115 Chr.12: 122979732-122979732 on Build GRCh38
rs943325748 Chr.12: 122979352-122979352 on Build GRCh38
rs1002250193 Chr.12: 122979435-122979435 on Build GRCh38
rs754257944 Chr.12: 122979374-122979375 on Build GRCh38
rs565186301 Chr.12: 122979669-122979669 on Build GRCh38
rs141715131 Chr.12: 122979753-122979753 on Build GRCh38
rs958009836 Chr.12: 122979459-122979459 on Build GRCh38
rs1055128872 Chr.12: 122979613-122979613 on Build GRCh38
rs778698112 Chr.12: 122979713-122979713 on Build GRCh38
rs563172345 Chr.12: 122979698-122979698 on Build GRCh38
rs545233205 Chr.12: 122979381-122979381 on Build GRCh38
rs534631529 Chr.12: 122979338-122979338 on Build GRCh38
rs914055626 Chr.12: 122979546-122979546 on Build GRCh38
rs746086798 Chr.12: 122979343-122979343 on Build GRCh38
rs532682045 Chr.12: 122979686-122979686 on Build GRCh38
rs567246926 Chr.12: 122979733-122979733 on Build GRCh38
rs764390775 Chr.12: 122979366-122979366 on Build GRCh38
rs899215916 Chr.12: 122979358-122979358 on Build GRCh38
rs371453887 Chr.12: 122979688-122979688 on Build GRCh38
rs188695145 Chr.12: 122979755-122979755 on Build GRCh38
rs896324780 Chr.12: 122979354-122979354 on Build GRCh38
rs770119016 Chr.12: 122979345-122979345 on Build GRCh38
rs561248446 Chr.12: 122979574-122979574 on Build GRCh38
rs954841671 Chr.12: 122979500-122979500 on Build GRCh38
rs969352751 Chr.12: 122979400-122979400 on Build GRCh38
rs1005949809 Chr.12: 122979380-122979380 on Build GRCh38
rs937988830 Chr.12: 122979609-122979609 on Build GRCh38
rs910769992 Chr.12: 122979558-122979559 on Build GRCh38
rs975643064 Chr.12: 122979442-122979442 on Build GRCh38

View Full Product Details