Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16132201-16132697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA2
Entrez Gene ID
1969
Gene Name
EPH receptor A2
Gene Aliases
ARCC2, CTPA, CTPP1, CTRCT6, ECK
UniGene
Hs.171596
Gene Chromosome Location
Chr.1: 16124337-16156104 on Build GRCh38
Transcripts
XM_017000534.1 XM_017000535.1 XM_017000536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5453972 c.2054-8C>A p.? Chr.1: 16132447-16132447 on Build GRCh38
3477572 c.2060C>T p.P687L Chr.1: 16132433-16132433 on Build GRCh38
1335888 c.2127C>T p.G709G Chr.1: 16132262-16132262 on Build GRCh38
3733930 c.2062A>C p.M688L Chr.1: 16132431-16132431 on Build GRCh38
3477571 c.2061C>T p.P687P Chr.1: 16132432-16132432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776884554 Chr.1: 16132419-16132419 on Build GRCh38
rs28394876 Chr.1: 16132353-16132353 on Build GRCh38
rs1042053606 Chr.1: 16132452-16132452 on Build GRCh38
rs749155445 Chr.1: 16132379-16132379 on Build GRCh38
rs758656920 Chr.1: 16132289-16132289 on Build GRCh38
rs780992981 Chr.1: 16132304-16132304 on Build GRCh38
rs373342266 Chr.1: 16132344-16132344 on Build GRCh38
rs372487104 Chr.1: 16132315-16132315 on Build GRCh38
rs546458254 Chr.1: 16132546-16132546 on Build GRCh38
rs372370904 Chr.1: 16132509-16132509 on Build GRCh38
rs373768600 Chr.1: 16132361-16132361 on Build GRCh38
rs776614918 Chr.1: 16132226-16132226 on Build GRCh38
rs769676253 Chr.1: 16132444-16132445 on Build GRCh38
rs1000430607 Chr.1: 16132607-16132607 on Build GRCh38
rs770343045 Chr.1: 16132420-16132420 on Build GRCh38
rs745684787 Chr.1: 16132398-16132398 on Build GRCh38
rs767606255 Chr.1: 16132450-16132450 on Build GRCh38
rs565871197 Chr.1: 16132317-16132317 on Build GRCh38
rs1055023170 Chr.1: 16132615-16132615 on Build GRCh38
rs781287716 Chr.1: 16132354-16132354 on Build GRCh38
rs775915325 Chr.1: 16132429-16132429 on Build GRCh38
rs963915589 Chr.1: 16132644-16132644 on Build GRCh38
rs775323762 Chr.1: 16132492-16132513 on Build GRCh38
rs554687807 Chr.1: 16132330-16132330 on Build GRCh38
rs768514997 Chr.1: 16132380-16132380 on Build GRCh38
rs577508347 Chr.1: 16132438-16132438 on Build GRCh38
rs750586785 Chr.1: 16132451-16132451 on Build GRCh38
rs866424508 Chr.1: 16132399-16132399 on Build GRCh38
rs771544356 Chr.1: 16132331-16132331 on Build GRCh38
rs753762655 Chr.1: 16132466-16132466 on Build GRCh38
rs774463742 Chr.1: 16132446-16132446 on Build GRCh38
rs765586631 Chr.1: 16132279-16132279 on Build GRCh38
rs368263231 Chr.1: 16132252-16132252 on Build GRCh38
rs762147989 Chr.1: 16132449-16132449 on Build GRCh38
rs763307879 Chr.1: 16132431-16132431 on Build GRCh38
rs368942223 Chr.1: 16132334-16132334 on Build GRCh38
rs766434233 Chr.1: 16132462-16132462 on Build GRCh38
rs1002428075 Chr.1: 16132494-16132494 on Build GRCh38
rs997423547 Chr.1: 16132653-16132653 on Build GRCh38
rs775040710 Chr.1: 16132234-16132234 on Build GRCh38
rs768403185 Chr.1: 16132253-16132253 on Build GRCh38
rs760138875 Chr.1: 16132333-16132333 on Build GRCh38
rs756136771 Chr.1: 16132460-16132460 on Build GRCh38
rs116506614 Chr.1: 16132227-16132227 on Build GRCh38
rs376787040 Chr.1: 16132228-16132228 on Build GRCh38
rs377555887 Chr.1: 16132349-16132349 on Build GRCh38
rs941278640 Chr.1: 16132370-16132370 on Build GRCh38
rs768491047 Chr.1: 16132320-16132320 on Build GRCh38
rs757300830 Chr.1: 16132300-16132300 on Build GRCh38
rs772818925 Chr.1: 16132413-16132413 on Build GRCh38
rs149080726 Chr.1: 16132396-16132396 on Build GRCh38
rs148264105 Chr.1: 16132403-16132403 on Build GRCh38
rs900035188 Chr.1: 16132647-16132647 on Build GRCh38
rs753038083 Chr.1: 16132282-16132282 on Build GRCh38
rs144576767 Chr.1: 16132388-16132388 on Build GRCh38
rs1033776063 Chr.1: 16132520-16132520 on Build GRCh38
rs1009670403 Chr.1: 16132434-16132434 on Build GRCh38
rs143440993 Chr.1: 16132675-16132675 on Build GRCh38
rs2291805 Chr.1: 16132319-16132319 on Build GRCh38
rs141594918 Chr.1: 16132393-16132393 on Build GRCh38
rs140708366 Chr.1: 16132262-16132262 on Build GRCh38
rs941167688 Chr.1: 16132560-16132560 on Build GRCh38
rs761405706 Chr.1: 16132255-16132255 on Build GRCh38
rs750451596 Chr.1: 16132357-16132357 on Build GRCh38
rs777965114 Chr.1: 16132290-16132290 on Build GRCh38
rs756119956 Chr.1: 16132358-16132358 on Build GRCh38
rs377546819 Chr.1: 16132369-16132369 on Build GRCh38
rs774135538 Chr.1: 16132328-16132328 on Build GRCh38
rs375203031 Chr.1: 16132298-16132298 on Build GRCh38
rs1038557517 Chr.1: 16132658-16132658 on Build GRCh38
rs372553014 Chr.1: 16132278-16132278 on Build GRCh38

View Full Product Details