Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76137445-76137943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
UniGene
Hs.661411
Gene Chromosome Location
Chr.15: 76059929-76204963 on Build GRCh38
Transcripts
NM_152335.2 XM_005254160.2 XM_017021915.1 XM_017021916.1 XM_017021917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5570193 c.176C>T p.T59M Chr.15: 76137844-76137844 on Build GRCh38
4057038 c.96A>C p.E32D Chr.15: 76137764-76137764 on Build GRCh38
4057037 c.94G>A p.E32K Chr.15: 76137762-76137762 on Build GRCh38
6614362 c.54A>C p.G18G Chr.15: 76137722-76137722 on Build GRCh38
4420114 c.207C>T p.D69D Chr.15: 76137875-76137875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758162612 Chr.15: 76137844-76137844 on Build GRCh38
rs752239257 Chr.15: 76137914-76137914 on Build GRCh38
rs777543871 Chr.15: 76137560-76137560 on Build GRCh38
rs573411058 Chr.15: 76137583-76137583 on Build GRCh38
rs1044547272 Chr.15: 76137780-76137780 on Build GRCh38
rs750138418 Chr.15: 76137696-76137696 on Build GRCh38
rs201363747 Chr.15: 76137868-76137868 on Build GRCh38
rs370445042 Chr.15: 76137741-76137741 on Build GRCh38
rs747935485 Chr.15: 76137720-76137720 on Build GRCh38
rs187813842 Chr.15: 76137884-76137884 on Build GRCh38
rs907605756 Chr.15: 76137609-76137609 on Build GRCh38
rs540647010 Chr.15: 76137811-76137811 on Build GRCh38
rs148471385 Chr.15: 76137559-76137559 on Build GRCh38
rs771077402 Chr.15: 76137753-76137753 on Build GRCh38
rs182426528 Chr.15: 76137694-76137694 on Build GRCh38
rs537501127 Chr.15: 76137628-76137628 on Build GRCh38
rs562019254 Chr.15: 76137875-76137875 on Build GRCh38
rs758292920 Chr.15: 76137917-76137917 on Build GRCh38
rs754449801 Chr.15: 76137897-76137897 on Build GRCh38
rs942207020 Chr.15: 76137764-76137764 on Build GRCh38
rs971145002 Chr.15: 76137885-76137885 on Build GRCh38
rs374327200 Chr.15: 76137858-76137858 on Build GRCh38
rs377726177 Chr.15: 76137704-76137704 on Build GRCh38
rs747209908 Chr.15: 76137743-76137743 on Build GRCh38
rs779577432 Chr.15: 76137703-76137703 on Build GRCh38
rs778828569 Chr.15: 76137705-76137705 on Build GRCh38
rs1010202517 Chr.15: 76137860-76137860 on Build GRCh38
rs577119878 Chr.15: 76137565-76137565 on Build GRCh38
rs926330571 Chr.15: 76137701-76137701 on Build GRCh38
rs900090487 Chr.15: 76137833-76137833 on Build GRCh38
rs757068587 Chr.15: 76137921-76137921 on Build GRCh38
rs376759726 Chr.15: 76137693-76137693 on Build GRCh38
rs910487381 Chr.15: 76137654-76137654 on Build GRCh38
rs138484129 Chr.15: 76137654-76137654 on Build GRCh38
rs777571379 Chr.15: 76137845-76137845 on Build GRCh38
rs868104115 Chr.15: 76137552-76137552 on Build GRCh38
rs927226129 Chr.15: 76137653-76137653 on Build GRCh38
rs202112486 Chr.15: 76137640-76137640 on Build GRCh38
rs778812089 Chr.15: 76137834-76137834 on Build GRCh38
rs759691310 Chr.15: 76137770-76137770 on Build GRCh38
rs906095906 Chr.15: 76137480-76137480 on Build GRCh38
rs776782113 Chr.15: 76137854-76137854 on Build GRCh38
rs200729077 Chr.15: 76137847-76137847 on Build GRCh38
rs569425236 Chr.15: 76137533-76137533 on Build GRCh38
rs776711597 Chr.15: 76137762-76137762 on Build GRCh38
rs754552309 Chr.15: 76137831-76137831 on Build GRCh38
rs765850065 Chr.15: 76137658-76137658 on Build GRCh38
rs763813971 Chr.15: 76137724-76137731 on Build GRCh38
rs764772320 Chr.15: 76137898-76137898 on Build GRCh38
rs560165284 Chr.15: 76137785-76137785 on Build GRCh38
rs776108925 Chr.15: 76137859-76137859 on Build GRCh38
rs771915334 Chr.15: 76137726-76137726 on Build GRCh38
rs762095902 Chr.15: 76137876-76137876 on Build GRCh38
rs753279696 Chr.15: 76137823-76137823 on Build GRCh38
rs979257052 Chr.15: 76137573-76137573 on Build GRCh38
rs555987589 Chr.15: 76137652-76137652 on Build GRCh38
rs764539432 Chr.15: 76137870-76137870 on Build GRCh38
rs35624427 Chr.15: 76137707-76137707 on Build GRCh38
rs200391773 Chr.15: 76137810-76137810 on Build GRCh38
rs981276680 Chr.15: 76137629-76137629 on Build GRCh38
rs776195376 Chr.15: 76137790-76137790 on Build GRCh38
rs1002848655 Chr.15: 76137486-76137486 on Build GRCh38
rs760463765 Chr.15: 76137675-76137676 on Build GRCh38
rs749789131 Chr.15: 76137603-76137603 on Build GRCh38
rs746808752 Chr.15: 76137920-76137920 on Build GRCh38
rs928955471 Chr.15: 76137714-76137714 on Build GRCh38
rs753553131 Chr.15: 76137891-76137891 on Build GRCh38
rs1035681799 Chr.15: 76137532-76137532 on Build GRCh38
rs763386965 Chr.15: 76137808-76137808 on Build GRCh38
rs765962968 Chr.15: 76137815-76137815 on Build GRCh38
rs372015383 Chr.15: 76137853-76137853 on Build GRCh38
rs566493500 Chr.15: 76137558-76137558 on Build GRCh38
rs891192320 Chr.15: 76137748-76137748 on Build GRCh38
rs778383237 Chr.15: 76137616-76137616 on Build GRCh38
rs189814130 Chr.15: 76137543-76137543 on Build GRCh38
rs753041041 Chr.15: 76137672-76137672 on Build GRCh38
rs756914056 Chr.15: 76137586-76137586 on Build GRCh38
rs745928129 Chr.15: 76137856-76137856 on Build GRCh38
rs551773217 Chr.15: 76137551-76137551 on Build GRCh38

View Full Product Details