Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35720693-35721192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMT2B
Entrez Gene ID
9757
Gene Name
lysine methyltransferase 2B
Gene Aliases
CXXC10, HRX2, MLL1B, MLL2, MLL4, TRX2, WBP-7, WBP7
UniGene
Hs.676457
Gene Chromosome Location
Chr.19: 35717818-35738880 on Build GRCh38
Transcripts
NM_014727.2 XM_011527561.2 XM_011527562.2 XM_017027544.1 XM_017027545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5686028 c.1780G>T p.V594F Chr.19: 35721127-35721127 on Build GRCh38
4471509 c.1722C>T p.P574P Chr.19: 35721069-35721069 on Build GRCh38
4466455 c.1435C>T p.R479W Chr.19: 35720782-35720782 on Build GRCh38
6003592 c.1504G>A p.A502T Chr.19: 35720851-35720851 on Build GRCh38
4646267 c.1374T>C p.P458P Chr.19: 35720721-35720721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775276995 Chr.19: 35720997-35720997 on Build GRCh38
rs748289035 Chr.19: 35720954-35720954 on Build GRCh38
rs1008249304 Chr.19: 35720787-35720787 on Build GRCh38
rs116170996 Chr.19: 35720922-35720922 on Build GRCh38
rs760508345 Chr.19: 35721100-35721100 on Build GRCh38
rs779053204 Chr.19: 35720940-35720940 on Build GRCh38
rs527331134 Chr.19: 35720951-35720951 on Build GRCh38
rs755199038 Chr.19: 35720934-35720934 on Build GRCh38
rs766738948 Chr.19: 35720862-35720862 on Build GRCh38
rs773918376 Chr.19: 35721067-35721067 on Build GRCh38
rs752887254 Chr.19: 35721018-35721018 on Build GRCh38
rs753528166 Chr.19: 35720782-35720782 on Build GRCh38
rs564031735 Chr.19: 35721015-35721015 on Build GRCh38
rs773186530 Chr.19: 35721168-35721168 on Build GRCh38
rs1057519279 Chr.19: 35720980-35720980 on Build GRCh38
rs909409167 Chr.19: 35720786-35720786 on Build GRCh38
rs920292369 Chr.19: 35721136-35721136 on Build GRCh38
rs368178075 Chr.19: 35721045-35721045 on Build GRCh38
rs768154617 Chr.19: 35721062-35721062 on Build GRCh38
rs764912329 Chr.19: 35720880-35720880 on Build GRCh38
rs761948097 Chr.19: 35720785-35720785 on Build GRCh38
rs372702715 Chr.19: 35721154-35721154 on Build GRCh38
rs745622279 Chr.19: 35720931-35720931 on Build GRCh38
rs757068223 Chr.19: 35720825-35720825 on Build GRCh38
rs781774179 Chr.19: 35721116-35721116 on Build GRCh38
rs751482151 Chr.19: 35720917-35720917 on Build GRCh38
rs909482859 Chr.19: 35721098-35721098 on Build GRCh38
rs758647357 Chr.19: 35721106-35721106 on Build GRCh38
rs772190237 Chr.19: 35720957-35720957 on Build GRCh38
rs920487561 Chr.19: 35720743-35720743 on Build GRCh38
rs753586954 Chr.19: 35721105-35721105 on Build GRCh38
rs202218975 Chr.19: 35720981-35720981 on Build GRCh38
rs759290935 Chr.19: 35720878-35720878 on Build GRCh38
rs375618290 Chr.19: 35720883-35720883 on Build GRCh38
rs767973202 Chr.19: 35720738-35720738 on Build GRCh38
rs200719562 Chr.19: 35721075-35721075 on Build GRCh38
rs748160305 Chr.19: 35720853-35720853 on Build GRCh38
rs746354562 Chr.19: 35721117-35721117 on Build GRCh38
rs773760714 Chr.19: 35720856-35720856 on Build GRCh38
rs574808075 Chr.19: 35720722-35720722 on Build GRCh38
rs368369291 Chr.19: 35721026-35721026 on Build GRCh38
rs767552559 Chr.19: 35721087-35721087 on Build GRCh38
rs1057519283 Chr.19: 35721037-35721037 on Build GRCh38
rs775330360 Chr.19: 35721059-35721059 on Build GRCh38
rs775294431 Chr.19: 35720996-35720997 on Build GRCh38
rs767291593 Chr.19: 35721009-35721009 on Build GRCh38
rs138918689 Chr.19: 35720745-35720745 on Build GRCh38
rs770250640 Chr.19: 35721056-35721056 on Build GRCh38
rs773635592 Chr.19: 35720958-35720958 on Build GRCh38
rs80216638 Chr.19: 35720840-35720840 on Build GRCh38
rs750233315 Chr.19: 35721010-35721010 on Build GRCh38
rs1001821773 Chr.19: 35720905-35720905 on Build GRCh38
rs761171846 Chr.19: 35721083-35721083 on Build GRCh38
rs771300360 Chr.19: 35720969-35720969 on Build GRCh38
rs965503785 Chr.19: 35720949-35720949 on Build GRCh38
rs747570960 Chr.19: 35720959-35720959 on Build GRCh38
rs781065031 Chr.19: 35720929-35720929 on Build GRCh38
rs777053465 Chr.19: 35720977-35720977 on Build GRCh38
rs576728234 Chr.19: 35720751-35720751 on Build GRCh38
rs762590352 Chr.19: 35720998-35720998 on Build GRCh38
rs2242519 Chr.19: 35721107-35721107 on Build GRCh38
rs868680634 Chr.19: 35720867-35720867 on Build GRCh38
rs757967105 Chr.19: 35720884-35720884 on Build GRCh38
rs777708173 Chr.19: 35720804-35720804 on Build GRCh38
rs377239282 Chr.19: 35721092-35721092 on Build GRCh38
rs780178514 Chr.19: 35720837-35720837 on Build GRCh38
rs773862731 Chr.19: 35721148-35721148 on Build GRCh38
rs761455591 Chr.19: 35721149-35721149 on Build GRCh38
rs764003155 Chr.19: 35720901-35720901 on Build GRCh38
rs749549494 Chr.19: 35721137-35721137 on Build GRCh38
rs775752899 Chr.19: 35720774-35720774 on Build GRCh38
rs541943377 Chr.19: 35720868-35720868 on Build GRCh38
rs772584640 Chr.19: 35720855-35720855 on Build GRCh38
rs376336021 Chr.19: 35720871-35720871 on Build GRCh38
rs757278967 Chr.19: 35721115-35721115 on Build GRCh38
rs759918812 Chr.19: 35720978-35720978 on Build GRCh38
rs760631410 Chr.19: 35721170-35721170 on Build GRCh38
rs768819878 Chr.19: 35720850-35720850 on Build GRCh38
rs185072239 Chr.19: 35720818-35720818 on Build GRCh38
rs770375335 Chr.19: 35721132-35721132 on Build GRCh38
rs907295056 Chr.19: 35720832-35720832 on Build GRCh38
rs755894161 Chr.19: 35721012-35721012 on Build GRCh38
rs367816667 Chr.19: 35721046-35721046 on Build GRCh38
rs761014480 Chr.19: 35720858-35720858 on Build GRCh38
rs371471934 Chr.19: 35721054-35721054 on Build GRCh38
rs762659366 Chr.19: 35721110-35721112 on Build GRCh38
rs763637244 Chr.19: 35721002-35721002 on Build GRCh38
rs374728315 Chr.19: 35721038-35721038 on Build GRCh38
rs761843749 Chr.19: 35721008-35721008 on Build GRCh38
rs545651739 Chr.19: 35720793-35720793 on Build GRCh38
rs768693126 Chr.19: 35720715-35720715 on Build GRCh38
rs374554860 Chr.19: 35720851-35720851 on Build GRCh38
rs371783473 Chr.19: 35721035-35721035 on Build GRCh38
rs762689767 Chr.19: 35721061-35721061 on Build GRCh38
rs985065417 Chr.19: 35721071-35721071 on Build GRCh38
rs766007976 Chr.19: 35720761-35720761 on Build GRCh38
rs1007147127 Chr.19: 35720921-35720921 on Build GRCh38
rs973051945 Chr.19: 35721126-35721126 on Build GRCh38
rs1050214389 Chr.19: 35720770-35720770 on Build GRCh38
rs758411036 Chr.19: 35720803-35720803 on Build GRCh38
rs866671941 Chr.19: 35720933-35720933 on Build GRCh38
rs751715820 Chr.19: 35721112-35721112 on Build GRCh38
rs781760377 Chr.19: 35721053-35721053 on Build GRCh38
rs533922071 Chr.19: 35720725-35720725 on Build GRCh38
rs764859385 Chr.19: 35720792-35720792 on Build GRCh38
rs780597908 Chr.19: 35721133-35721133 on Build GRCh38
rs754581856 Chr.19: 35720784-35720784 on Build GRCh38
rs189407972 Chr.19: 35721101-35721101 on Build GRCh38
rs750985731 Chr.19: 35721005-35721005 on Build GRCh38
rs113085641 Chr.19: 35720861-35720861 on Build GRCh38
rs1050328540 Chr.19: 35721171-35721171 on Build GRCh38

View Full Product Details