Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53873812-53874306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYADM
Entrez Gene ID
91663
Gene Name
myeloid associated differentiation marker
Gene Aliases
SB135
UniGene
Hs.380906
Gene Chromosome Location
Chr.19: 53865584-53876435 on Build GRCh38
Transcripts
NM_001020818.2 NM_001020819.2 NM_001020820.2 NM_001020821.2 NM_001290188.1 NM_001290189.1 NM_001290190.1 NM_001290191.1 NM_001290192.1 NM_001290193.1 NM_001290194.1 NM_138373.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4081105 c.385T>C p.S129P Chr.19: 53873914-53873914 on Build GRCh38
5642533 c.635T>C p.I212T Chr.19: 53874164-53874164 on Build GRCh38
4081107 c.566C>T p.A189V Chr.19: 53874095-53874095 on Build GRCh38
6996131 c.408C>T p.H136H Chr.19: 53873937-53873937 on Build GRCh38
1396153 c.410C>T p.A137V Chr.19: 53873939-53873939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562245107 Chr.19: 53874220-53874220 on Build GRCh38
rs764639876 Chr.19: 53873934-53873934 on Build GRCh38
rs1036529376 Chr.19: 53874195-53874195 on Build GRCh38
rs761711160 Chr.19: 53874218-53874218 on Build GRCh38
rs767544336 Chr.19: 53874135-53874135 on Build GRCh38
rs781546645 Chr.19: 53873998-53873999 on Build GRCh38
rs372653211 Chr.19: 53873977-53873977 on Build GRCh38
rs573155428 Chr.19: 53873954-53873954 on Build GRCh38
rs367969185 Chr.19: 53874153-53874153 on Build GRCh38
rs761638066 Chr.19: 53874132-53874132 on Build GRCh38
rs750593806 Chr.19: 53874045-53874046 on Build GRCh38
rs772753619 Chr.19: 53873943-53873943 on Build GRCh38
rs909453043 Chr.19: 53874183-53874183 on Build GRCh38
rs752509164 Chr.19: 53874015-53874015 on Build GRCh38
rs777895511 Chr.19: 53873895-53873895 on Build GRCh38
rs142869931 Chr.19: 53874186-53874186 on Build GRCh38
rs200301241 Chr.19: 53873833-53873833 on Build GRCh38
rs756149686 Chr.19: 53874145-53874145 on Build GRCh38
rs755142485 Chr.19: 53874238-53874238 on Build GRCh38
rs746114179 Chr.19: 53873923-53873923 on Build GRCh38
rs774019269 Chr.19: 53874277-53874277 on Build GRCh38
rs575762816 Chr.19: 53873918-53873918 on Build GRCh38
rs758799496 Chr.19: 53874171-53874171 on Build GRCh38
rs747451838 Chr.19: 53874254-53874254 on Build GRCh38
rs1047065286 Chr.19: 53873901-53873901 on Build GRCh38
rs757903347 Chr.19: 53874255-53874255 on Build GRCh38
rs564819081 Chr.19: 53874063-53874063 on Build GRCh38
rs781342599 Chr.19: 53873968-53873968 on Build GRCh38
rs765535366 Chr.19: 53874244-53874244 on Build GRCh38
rs991202023 Chr.19: 53874133-53874133 on Build GRCh38
rs748040595 Chr.19: 53873888-53873888 on Build GRCh38
rs757232240 Chr.19: 53873967-53873967 on Build GRCh38
rs765030899 Chr.19: 53874013-53874013 on Build GRCh38
rs529752727 Chr.19: 53874247-53874247 on Build GRCh38
rs780654913 Chr.19: 53874264-53874264 on Build GRCh38
rs928388680 Chr.19: 53873937-53873937 on Build GRCh38
rs139333242 Chr.19: 53874288-53874288 on Build GRCh38
rs868620973 Chr.19: 53874265-53874265 on Build GRCh38
rs778872762 Chr.19: 53873882-53873882 on Build GRCh38
rs767334901 Chr.19: 53874219-53874219 on Build GRCh38
rs768855797 Chr.19: 53874075-53874075 on Build GRCh38
rs889083260 Chr.19: 53874016-53874016 on Build GRCh38
rs779349845 Chr.19: 53874076-53874076 on Build GRCh38
rs774683208 Chr.19: 53874008-53874008 on Build GRCh38
rs143914793 Chr.19: 53874018-53874018 on Build GRCh38
rs150876352 Chr.19: 53873856-53873856 on Build GRCh38
rs751435104 Chr.19: 53874131-53874131 on Build GRCh38
rs768166941 Chr.19: 53874267-53874267 on Build GRCh38
rs140541138 Chr.19: 53874100-53874100 on Build GRCh38
rs748886165 Chr.19: 53874187-53874187 on Build GRCh38
rs920039400 Chr.19: 53873847-53873847 on Build GRCh38
rs777609873 Chr.19: 53873961-53873961 on Build GRCh38
rs112052285 Chr.19: 53874125-53874125 on Build GRCh38
rs779944413 Chr.19: 53874272-53874274 on Build GRCh38
rs766380194 Chr.19: 53874226-53874226 on Build GRCh38
rs763468606 Chr.19: 53873844-53873844 on Build GRCh38
rs750474173 Chr.19: 53874139-53874139 on Build GRCh38
rs901926968 Chr.19: 53874007-53874007 on Build GRCh38
rs550331558 Chr.19: 53874126-53874126 on Build GRCh38
rs200527300 Chr.19: 53874179-53874179 on Build GRCh38
rs915591463 Chr.19: 53874156-53874156 on Build GRCh38
rs748605110 Chr.19: 53874169-53874169 on Build GRCh38
rs773999438 Chr.19: 53874197-53874197 on Build GRCh38
rs533715842 Chr.19: 53874154-53874154 on Build GRCh38
rs751392745 Chr.19: 53874032-53874032 on Build GRCh38
rs757219485 Chr.19: 53874034-53874034 on Build GRCh38
rs763726840 Chr.19: 53874129-53874129 on Build GRCh38
rs754692319 Chr.19: 53873948-53873948 on Build GRCh38
rs759860136 Chr.19: 53873832-53873832 on Build GRCh38
rs747781975 Chr.19: 53874279-53874279 on Build GRCh38
rs763442113 Chr.19: 53873930-53873930 on Build GRCh38
rs765993680 Chr.19: 53873865-53873865 on Build GRCh38
rs747539869 Chr.19: 53874184-53874184 on Build GRCh38
rs952331346 Chr.19: 53874158-53874158 on Build GRCh38
rs746019741 Chr.19: 53873969-53873969 on Build GRCh38
rs373472423 Chr.19: 53874108-53874108 on Build GRCh38
rs749546470 Chr.19: 53874051-53874051 on Build GRCh38
rs1036883212 Chr.19: 53874004-53874004 on Build GRCh38
rs985466484 Chr.19: 53874159-53874159 on Build GRCh38
rs756226376 Chr.19: 53874230-53874235 on Build GRCh38
rs779694099 Chr.19: 53874260-53874260 on Build GRCh38
rs923401316 Chr.19: 53874208-53874208 on Build GRCh38
rs780132482 Chr.19: 53874147-53874147 on Build GRCh38
rs766950916 Chr.19: 53873853-53873853 on Build GRCh38
rs963903213 Chr.19: 53874030-53874030 on Build GRCh38
rs775233357 Chr.19: 53874124-53874124 on Build GRCh38
rs375338651 Chr.19: 53873924-53873924 on Build GRCh38
rs940848866 Chr.19: 53873983-53873983 on Build GRCh38
rs140166646 Chr.19: 53874010-53874010 on Build GRCh38
rs1011921382 Chr.19: 53874037-53874037 on Build GRCh38
rs200027989 Chr.19: 53873952-53873952 on Build GRCh38
rs771155643 Chr.19: 53873919-53873919 on Build GRCh38
rs977593432 Chr.19: 53873920-53873920 on Build GRCh38
rs146783056 Chr.19: 53874043-53874043 on Build GRCh38
rs775841521 Chr.19: 53873927-53873927 on Build GRCh38
rs746997005 Chr.19: 53873963-53873963 on Build GRCh38
rs775929758 Chr.19: 53873842-53873842 on Build GRCh38
rs765057736 Chr.19: 53873951-53873951 on Build GRCh38
rs201905769 Chr.19: 53874042-53874042 on Build GRCh38
rs114795210 Chr.19: 53873946-53873946 on Build GRCh38
rs770065060 Chr.19: 53873970-53873970 on Build GRCh38
rs1027198696 Chr.19: 53874096-53874096 on Build GRCh38
rs892168506 Chr.19: 53873988-53873988 on Build GRCh38
rs769459181 Chr.19: 53874112-53874112 on Build GRCh38
rs369701605 Chr.19: 53874081-53874081 on Build GRCh38
rs758414883 Chr.19: 53873889-53873889 on Build GRCh38
rs754016935 Chr.19: 53874231-53874231 on Build GRCh38
rs780298853 Chr.19: 53874048-53874048 on Build GRCh38
rs758739416 Chr.19: 53874249-53874249 on Build GRCh38
rs755235316 Chr.19: 53874160-53874160 on Build GRCh38
rs908617153 Chr.19: 53874175-53874175 on Build GRCh38
rs780357518 Chr.19: 53874011-53874011 on Build GRCh38
rs372001283 Chr.19: 53873859-53873859 on Build GRCh38
rs769178252 Chr.19: 53873993-53873993 on Build GRCh38
rs557514172 Chr.19: 53873871-53873871 on Build GRCh38
rs754778239 Chr.19: 53873873-53873873 on Build GRCh38
rs376434685 Chr.19: 53873982-53873982 on Build GRCh38
rs778294247 Chr.19: 53874251-53874251 on Build GRCh38
rs779142146 Chr.19: 53874163-53874163 on Build GRCh38
rs759156943 Chr.19: 53874110-53874110 on Build GRCh38
rs768409589 Chr.19: 53874188-53874188 on Build GRCh38

View Full Product Details