Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19036594-19037096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR2
Entrez Gene ID
9993
Gene Name
DiGeorge syndrome critical region gene 2
Gene Aliases
DGS-C, IDD, LAN, SEZ-12
UniGene
Hs.517357
Gene Chromosome Location
Chr.22: 19036282-19122454 on Build GRCh38
Transcripts
NM_001173533.1 NM_001173534.1 NM_001184781.1 NM_005137.2 NR_033674.1

SNP Detail

SNP ID Chromosome Location
rs1007680708 Chr.22: 19037008-19037008 on Build GRCh38
rs971561114 Chr.22: 19037042-19037042 on Build GRCh38
rs973705296 Chr.22: 19036859-19036859 on Build GRCh38
rs977030127 Chr.22: 19036775-19036775 on Build GRCh38
rs1055556980 Chr.22: 19036747-19036747 on Build GRCh38
rs561592858 Chr.22: 19036922-19036922 on Build GRCh38
rs866394413 Chr.22: 19036670-19036670 on Build GRCh38
rs190793030 Chr.22: 19036999-19036999 on Build GRCh38
rs930222379 Chr.22: 19036873-19036873 on Build GRCh38
rs371700698 Chr.22: 19036908-19036908 on Build GRCh38
rs761245181 Chr.22: 19036960-19036960 on Build GRCh38
rs991285241 Chr.22: 19036804-19036804 on Build GRCh38
rs915626607 Chr.22: 19037026-19037026 on Build GRCh38
rs895340038 Chr.22: 19036642-19036642 on Build GRCh38
rs577090746 Chr.22: 19036729-19036730 on Build GRCh38
rs929615104 Chr.22: 19036713-19036714 on Build GRCh38
rs547571088 Chr.22: 19036799-19036799 on Build GRCh38
rs1054341296 Chr.22: 19036629-19036629 on Build GRCh38
rs928165961 Chr.22: 19036864-19036864 on Build GRCh38
rs1021947471 Chr.22: 19036817-19036817 on Build GRCh38
rs535877410 Chr.22: 19036880-19036880 on Build GRCh38
rs967428492 Chr.22: 19036821-19036821 on Build GRCh38
rs1047102014 Chr.22: 19036712-19036712 on Build GRCh38
rs544301849 Chr.22: 19036907-19036907 on Build GRCh38
rs113989059 Chr.22: 19036982-19036982 on Build GRCh38
rs967277243 Chr.22: 19036760-19036760 on Build GRCh38
rs973995495 Chr.22: 19037033-19037033 on Build GRCh38
rs906975403 Chr.22: 19036716-19036716 on Build GRCh38
rs925640287 Chr.22: 19036796-19036796 on Build GRCh38
rs548168810 Chr.22: 19036693-19036693 on Build GRCh38
rs997996469 Chr.22: 19036930-19036930 on Build GRCh38
rs1030337689 Chr.22: 19036992-19036993 on Build GRCh38
rs888503488 Chr.22: 19036713-19036713 on Build GRCh38
rs755240255 Chr.22: 19036927-19036927 on Build GRCh38
rs1046723404 Chr.22: 19036891-19036891 on Build GRCh38
rs919523146 Chr.22: 19036861-19036861 on Build GRCh38
rs539517126 Chr.22: 19036839-19036839 on Build GRCh38
rs982271563 Chr.22: 19037059-19037059 on Build GRCh38
rs928264474 Chr.22: 19036904-19036904 on Build GRCh38
rs963868936 Chr.22: 19036856-19036856 on Build GRCh38
rs569930800 Chr.22: 19036854-19036854 on Build GRCh38
rs1011805261 Chr.22: 19036928-19036929 on Build GRCh38
rs986308607 Chr.22: 19036671-19036671 on Build GRCh38
rs570671730 Chr.22: 19036823-19036823 on Build GRCh38
rs950971501 Chr.22: 19037073-19037074 on Build GRCh38
rs1029617583 Chr.22: 19036758-19036758 on Build GRCh38
rs1046679706 Chr.22: 19036682-19036682 on Build GRCh38
rs527481460 Chr.22: 19036743-19036743 on Build GRCh38
rs117455748 Chr.22: 19036619-19036619 on Build GRCh38
rs370014755 Chr.22: 19036747-19036748 on Build GRCh38
rs963474267 Chr.22: 19037014-19037014 on Build GRCh38
rs575621893 Chr.22: 19036757-19036757 on Build GRCh38
rs557896280 Chr.22: 19036957-19036957 on Build GRCh38
rs929533864 Chr.22: 19036882-19036882 on Build GRCh38
rs1029464657 Chr.22: 19036947-19036947 on Build GRCh38
rs1032628669 Chr.22: 19036998-19036998 on Build GRCh38
rs928133176 Chr.22: 19037073-19037073 on Build GRCh38
rs372503787 Chr.22: 19036986-19036986 on Build GRCh38
rs967244563 Chr.22: 19036961-19036961 on Build GRCh38
rs763310516 Chr.22: 19036749-19036749 on Build GRCh38
rs867138210 Chr.22: 19036981-19036981 on Build GRCh38
rs200923991 Chr.22: 19036729-19036729 on Build GRCh38
rs555631342 Chr.22: 19036888-19036888 on Build GRCh38
rs531364182 Chr.22: 19036688-19036688 on Build GRCh38
rs375602542 Chr.22: 19036748-19036749 on Build GRCh38
rs186078418 Chr.22: 19036853-19036853 on Build GRCh38
rs141810894 Chr.22: 19037010-19037010 on Build GRCh38
rs890456469 Chr.22: 19037007-19037007 on Build GRCh38

View Full Product Details