Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20976559-20977055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564359 c.1128_1129GG>AA p.G377S Chr.22: 20976748-20976749 on Build GRCh38
3390113 c.1218+5G>T p.? Chr.22: 20976943-20976943 on Build GRCh38
6227852 c.1074G>A p.T358T Chr.22: 20976676-20976676 on Build GRCh38
5366513 c.1141C>T p.R381C Chr.22: 20976743-20976743 on Build GRCh38
4564361 c.1092_1093GG>AA p.G365S Chr.22: 20976748-20976749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867788850 Chr.22: 20976756-20976756 on Build GRCh38
rs767000408 Chr.22: 20976851-20976851 on Build GRCh38
rs767637166 Chr.22: 20976943-20976944 on Build GRCh38
rs186295000 Chr.22: 20976905-20976905 on Build GRCh38
rs751783736 Chr.22: 20976997-20976997 on Build GRCh38
rs760162047 Chr.22: 20976975-20976975 on Build GRCh38
rs181406196 Chr.22: 20976633-20976633 on Build GRCh38
rs551037156 Chr.22: 20976938-20976938 on Build GRCh38
rs775285677 Chr.22: 20976752-20976752 on Build GRCh38
rs145883640 Chr.22: 20976898-20976898 on Build GRCh38
rs754933734 Chr.22: 20977014-20977014 on Build GRCh38
rs758033678 Chr.22: 20976616-20976616 on Build GRCh38
rs764514211 Chr.22: 20976995-20976995 on Build GRCh38
rs777305076 Chr.22: 20976618-20976618 on Build GRCh38
rs779417568 Chr.22: 20976792-20976792 on Build GRCh38
rs374843871 Chr.22: 20976648-20976648 on Build GRCh38
rs775234120 Chr.22: 20976684-20976684 on Build GRCh38
rs760738469 Chr.22: 20976605-20976605 on Build GRCh38
rs745868220 Chr.22: 20976746-20976746 on Build GRCh38
rs572514447 Chr.22: 20976764-20976764 on Build GRCh38
rs537276493 Chr.22: 20976706-20976706 on Build GRCh38
rs777938488 Chr.22: 20976807-20976807 on Build GRCh38
rs1033974929 Chr.22: 20976672-20976672 on Build GRCh38
rs778661498 Chr.22: 20977015-20977015 on Build GRCh38
rs374116315 Chr.22: 20976811-20976811 on Build GRCh38
rs368535113 Chr.22: 20976608-20976608 on Build GRCh38
rs189788146 Chr.22: 20976767-20976767 on Build GRCh38
rs45606234 Chr.22: 20976696-20976696 on Build GRCh38
rs868239600 Chr.22: 20976713-20976713 on Build GRCh38
rs142530985 Chr.22: 20976745-20976745 on Build GRCh38
rs756717490 Chr.22: 20976714-20976714 on Build GRCh38
rs751081103 Chr.22: 20976707-20976707 on Build GRCh38
rs753021490 Chr.22: 20976796-20976796 on Build GRCh38
rs780265128 Chr.22: 20976644-20976644 on Build GRCh38
rs752414779 Chr.22: 20976612-20976612 on Build GRCh38
rs1013789820 Chr.22: 20976694-20976694 on Build GRCh38
rs772263424 Chr.22: 20976744-20976744 on Build GRCh38
rs747853511 Chr.22: 20976925-20976925 on Build GRCh38
rs900361173 Chr.22: 20976596-20976596 on Build GRCh38
rs1018708541 Chr.22: 20976963-20976963 on Build GRCh38
rs565364047 Chr.22: 20976606-20976606 on Build GRCh38
rs764802691 Chr.22: 20976609-20976609 on Build GRCh38
rs178269 Chr.22: 20976754-20976754 on Build GRCh38
rs371098629 Chr.22: 20976723-20976723 on Build GRCh38
rs377355378 Chr.22: 20976981-20976981 on Build GRCh38
rs1014569101 Chr.22: 20976844-20976844 on Build GRCh38
rs767619338 Chr.22: 20976999-20976999 on Build GRCh38
rs917385279 Chr.22: 20976633-20976633 on Build GRCh38
rs373473269 Chr.22: 20976717-20976717 on Build GRCh38
rs145487001 Chr.22: 20976675-20976675 on Build GRCh38
rs771588412 Chr.22: 20976839-20976839 on Build GRCh38
rs776260420 Chr.22: 20976968-20976968 on Build GRCh38
rs754271331 Chr.22: 20976785-20976785 on Build GRCh38
rs373121171 Chr.22: 20976884-20976884 on Build GRCh38
rs559227367 Chr.22: 20976945-20976950 on Build GRCh38
rs201592533 Chr.22: 20976890-20976890 on Build GRCh38
rs778803055 Chr.22: 20976924-20976924 on Build GRCh38
rs766947115 Chr.22: 20976784-20976784 on Build GRCh38
rs774053295 Chr.22: 20976758-20976758 on Build GRCh38
rs762623722 Chr.22: 20976690-20976690 on Build GRCh38
rs746930152 Chr.22: 20976668-20976668 on Build GRCh38
rs749574773 Chr.22: 20976647-20976647 on Build GRCh38
rs1009288832 Chr.22: 20976602-20976602 on Build GRCh38
rs975977010 Chr.22: 20977004-20977004 on Build GRCh38
rs746643800 Chr.22: 20976902-20976902 on Build GRCh38
rs751028058 Chr.22: 20976619-20976619 on Build GRCh38
rs551098812 Chr.22: 20976670-20976670 on Build GRCh38
rs779898129 Chr.22: 20976815-20976815 on Build GRCh38
rs926234535 Chr.22: 20976688-20976688 on Build GRCh38
rs369219792 Chr.22: 20976676-20976676 on Build GRCh38
rs1004941440 Chr.22: 20976843-20976843 on Build GRCh38
rs770625010 Chr.22: 20976942-20976942 on Build GRCh38
rs769577965 Chr.22: 20976749-20976749 on Build GRCh38
rs755276108 Chr.22: 20976738-20976738 on Build GRCh38
rs780610191 Chr.22: 20976736-20976736 on Build GRCh38
rs750526067 Chr.22: 20976944-20976950 on Build GRCh38
rs139501949 Chr.22: 20976743-20976743 on Build GRCh38
rs1055770970 Chr.22: 20976864-20976864 on Build GRCh38
rs774607870 Chr.22: 20976659-20976659 on Build GRCh38
rs746220834 Chr.22: 20976634-20976634 on Build GRCh38
rs754463554 Chr.22: 20976858-20976858 on Build GRCh38
rs576119035 Chr.22: 20976814-20976814 on Build GRCh38
rs779222361 Chr.22: 20976740-20976740 on Build GRCh38
rs774106229 Chr.22: 20976827-20976827 on Build GRCh38
rs745971943 Chr.22: 20976939-20976939 on Build GRCh38
rs774554901 Chr.22: 20976604-20976604 on Build GRCh38
rs964569212 Chr.22: 20976983-20976983 on Build GRCh38
rs747784434 Chr.22: 20976769-20976769 on Build GRCh38
rs756565479 Chr.22: 20976640-20976640 on Build GRCh38
rs748191991 Chr.22: 20976664-20976664 on Build GRCh38
rs763159110 Chr.22: 20976986-20976986 on Build GRCh38
rs369396180 Chr.22: 20976841-20976841 on Build GRCh38
rs761354018 Chr.22: 20976760-20976760 on Build GRCh38
rs1042122717 Chr.22: 20976825-20976825 on Build GRCh38
rs199667436 Chr.22: 20976840-20976840 on Build GRCh38
rs952816331 Chr.22: 20976904-20976904 on Build GRCh38
rs571336795 Chr.22: 20977029-20977029 on Build GRCh38
rs768428267 Chr.22: 20976719-20976719 on Build GRCh38
rs144862097 Chr.22: 20976650-20976650 on Build GRCh38
rs769692208 Chr.22: 20977009-20977009 on Build GRCh38
rs758527929 Chr.22: 20976799-20976799 on Build GRCh38
rs749634759 Chr.22: 20976737-20976737 on Build GRCh38
rs762676617 Chr.22: 20976753-20976753 on Build GRCh38
rs756678786 Chr.22: 20976782-20976782 on Build GRCh38
rs751662410 Chr.22: 20976897-20976897 on Build GRCh38
rs371773561 Chr.22: 20976837-20976837 on Build GRCh38
rs757446473 Chr.22: 20976998-20976998 on Build GRCh38
rs765790583 Chr.22: 20976979-20976979 on Build GRCh38
rs758668022 Chr.22: 20976885-20976885 on Build GRCh38
rs771772853 Chr.22: 20976928-20976928 on Build GRCh38
rs369765956 Chr.22: 20977010-20977010 on Build GRCh38
rs749268990 Chr.22: 20976909-20976909 on Build GRCh38
rs186357127 Chr.22: 20976718-20976718 on Build GRCh38
rs528081577 Chr.22: 20976922-20976922 on Build GRCh38
rs749216020 Chr.22: 20976817-20976817 on Build GRCh38
rs765745859 Chr.22: 20976883-20976883 on Build GRCh38
rs372801515 Chr.22: 20976623-20976623 on Build GRCh38
rs755329071 Chr.22: 20976786-20976786 on Build GRCh38
rs140732562 Chr.22: 20976882-20976882 on Build GRCh38
rs749873712 Chr.22: 20976777-20976777 on Build GRCh38

View Full Product Details