Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76011091-76011587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2

SNP Detail

SNP ID Chromosome Location
rs906360825 Chr.4: 76011471-76011471 on Build GRCh38
rs557911345 Chr.4: 76011504-76011504 on Build GRCh38
rs529036187 Chr.4: 76011283-76011283 on Build GRCh38
rs1009334782 Chr.4: 76011247-76011247 on Build GRCh38
rs961477626 Chr.4: 76011319-76011319 on Build GRCh38
rs371029820 Chr.4: 76011461-76011461 on Build GRCh38
rs905030928 Chr.4: 76011228-76011228 on Build GRCh38
rs146114029 Chr.4: 76011429-76011430 on Build GRCh38
rs568705642 Chr.4: 76011369-76011369 on Build GRCh38
rs954254589 Chr.4: 76011258-76011258 on Build GRCh38
rs910018418 Chr.4: 76011141-76011141 on Build GRCh38
rs987060638 Chr.4: 76011498-76011498 on Build GRCh38
rs536072085 Chr.4: 76011442-76011442 on Build GRCh38
rs148909151 Chr.4: 76011356-76011356 on Build GRCh38
rs1037263055 Chr.4: 76011145-76011145 on Build GRCh38
rs868772547 Chr.4: 76011198-76011198 on Build GRCh38
rs1020681410 Chr.4: 76011375-76011375 on Build GRCh38
rs554360695 Chr.4: 76011443-76011443 on Build GRCh38
rs181818867 Chr.4: 76011293-76011293 on Build GRCh38
rs570278124 Chr.4: 76011246-76011246 on Build GRCh38
rs1033645979 Chr.4: 76011295-76011295 on Build GRCh38
rs943055095 Chr.4: 76011365-76011365 on Build GRCh38
rs954469404 Chr.4: 76011467-76011467 on Build GRCh38
rs971073863 Chr.4: 76011115-76011115 on Build GRCh38
rs912716165 Chr.4: 76011276-76011276 on Build GRCh38
rs896312542 Chr.4: 76011333-76011333 on Build GRCh38
rs959481232 Chr.4: 76011135-76011135 on Build GRCh38
rs143533268 Chr.4: 76011500-76011500 on Build GRCh38
rs547153110 Chr.4: 76011288-76011288 on Build GRCh38
rs77037165 Chr.4: 76011193-76011193 on Build GRCh38
rs934267039 Chr.4: 76011208-76011208 on Build GRCh38
rs940184893 Chr.4: 76011142-76011142 on Build GRCh38
rs756028249 Chr.4: 76011512-76011512 on Build GRCh38
rs1053479315 Chr.4: 76011237-76011237 on Build GRCh38
rs959646033 Chr.4: 76011307-76011307 on Build GRCh38
rs141537373 Chr.4: 76011215-76011215 on Build GRCh38
rs532820019 Chr.4: 76011308-76011308 on Build GRCh38
rs1003426809 Chr.4: 76011511-76011511 on Build GRCh38
rs758645495 Chr.4: 76011359-76011359 on Build GRCh38
rs1020238475 Chr.4: 76011499-76011499 on Build GRCh38
rs190090274 Chr.4: 76011136-76011136 on Build GRCh38
rs987397170 Chr.4: 76011271-76011271 on Build GRCh38
rs959337147 Chr.4: 76011527-76011527 on Build GRCh38
rs535982784 Chr.4: 76011132-76011132 on Build GRCh38
rs1002416770 Chr.4: 76011229-76011229 on Build GRCh38
rs919984833 Chr.4: 76011525-76011525 on Build GRCh38
rs765565668 Chr.4: 76011279-76011279 on Build GRCh38
rs754220697 Chr.4: 76011124-76011124 on Build GRCh38
rs561760506 Chr.4: 76011233-76011233 on Build GRCh38
rs550429086 Chr.4: 76011358-76011358 on Build GRCh38
rs909986373 Chr.4: 76011315-76011315 on Build GRCh38
rs757575752 Chr.4: 76011234-76011234 on Build GRCh38
rs138633833 Chr.4: 76011325-76011325 on Build GRCh38
rs867065188 Chr.4: 76011385-76011385 on Build GRCh38
rs572537350 Chr.4: 76011163-76011163 on Build GRCh38
rs780005984 Chr.4: 76011430-76011430 on Build GRCh38
rs922797378 Chr.4: 76011324-76011324 on Build GRCh38
rs1016977911 Chr.4: 76011561-76011561 on Build GRCh38
rs972885573 Chr.4: 76011321-76011321 on Build GRCh38
rs1005537372 Chr.4: 76011532-76011532 on Build GRCh38
rs535443307 Chr.4: 76011521-76011521 on Build GRCh38
rs569625190 Chr.4: 76011350-76011350 on Build GRCh38
rs982121244 Chr.4: 76011119-76011119 on Build GRCh38
rs967718859 Chr.4: 76011427-76011427 on Build GRCh38
rs150412491 Chr.4: 76011224-76011224 on Build GRCh38
rs979673182 Chr.4: 76011134-76011134 on Build GRCh38
rs1027968985 Chr.4: 76011451-76011451 on Build GRCh38
rs931282068 Chr.4: 76011305-76011305 on Build GRCh38
rs1020666485 Chr.4: 76011235-76011235 on Build GRCh38
rs546222952 Chr.4: 76011191-76011191 on Build GRCh38
rs1051202228 Chr.4: 76011154-76011154 on Build GRCh38

View Full Product Details