Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76400157-76400668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSC4D
Entrez Gene ID
136853
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Gene Aliases
S4D-SRCRB, SRCRB-S4D, SRCRB4D
UniGene
Hs.567684
Gene Chromosome Location
Chr.7: 76389329-76410025 on Build GRCh38
Transcripts
NM_080744.1 XM_017011750.1 NM_007155.5 XM_017012586.1

Gene Symbol
ZP3
Entrez Gene ID
7784
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
Gene Aliases
ZP3A, ZP3B, ZPC, Zp-3
UniGene
Hs.656137
Gene Chromosome Location
Chr.7: 76397524-76442071 on Build GRCh38
Transcripts
NM_080744.1 XM_017011750.1 NM_007155.5 XM_017012586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7273568 c.350G>A p.R117Q Chr.7: 76400411-76400411 on Build GRCh38
6351412 c.405C>T p.S135S Chr.7: 76400356-76400356 on Build GRCh38
2152877 c.417_418delCC p.S139fs*19 Chr.7: 76400343-76400344 on Build GRCh38
3923973 c.421G>A p.G141S Chr.7: 76400340-76400340 on Build GRCh38
7140965 c.289C>A p.R97S Chr.7: 76400472-76400472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773593386 Chr.7: 76400435-76400435 on Build GRCh38
rs764344413 Chr.7: 76400635-76400635 on Build GRCh38
rs756335400 Chr.7: 76400628-76400628 on Build GRCh38
rs754293744 Chr.7: 76400436-76400436 on Build GRCh38
rs752804800 Chr.7: 76400624-76400624 on Build GRCh38
rs369258749 Chr.7: 76400580-76400580 on Build GRCh38
rs115316236 Chr.7: 76400276-76400276 on Build GRCh38
rs894285577 Chr.7: 76400199-76400199 on Build GRCh38
rs1021511259 Chr.7: 76400234-76400234 on Build GRCh38
rs146956663 Chr.7: 76400310-76400310 on Build GRCh38
rs750796775 Chr.7: 76400503-76400503 on Build GRCh38
rs751682350 Chr.7: 76400510-76400510 on Build GRCh38
rs139339963 Chr.7: 76400408-76400408 on Build GRCh38
rs774226297 Chr.7: 76400244-76400244 on Build GRCh38
rs755084642 Chr.7: 76400517-76400517 on Build GRCh38
rs367656779 Chr.7: 76400343-76400343 on Build GRCh38
rs564222991 Chr.7: 76400558-76400558 on Build GRCh38
rs771145890 Chr.7: 76400238-76400238 on Build GRCh38
rs745651709 Chr.7: 76400540-76400540 on Build GRCh38
rs745912872 Chr.7: 76400324-76400324 on Build GRCh38
rs575407993 Chr.7: 76400506-76400506 on Build GRCh38
rs751627418 Chr.7: 76400445-76400445 on Build GRCh38
rs773157641 Chr.7: 76400285-76400285 on Build GRCh38
rs772170908 Chr.7: 76400255-76400255 on Build GRCh38
rs774779955 Chr.7: 76400315-76400315 on Build GRCh38
rs775186548 Chr.7: 76400487-76400487 on Build GRCh38
rs762821758 Chr.7: 76400289-76400289 on Build GRCh38
rs759473168 Chr.7: 76400407-76400407 on Build GRCh38
rs370215401 Chr.7: 76400331-76400331 on Build GRCh38
rs747237989 Chr.7: 76400376-76400376 on Build GRCh38
rs938177086 Chr.7: 76400573-76400573 on Build GRCh38
rs542129562 Chr.7: 76400488-76400488 on Build GRCh38
rs766693149 Chr.7: 76400568-76400568 on Build GRCh38
rs778626772 Chr.7: 76400539-76400539 on Build GRCh38
rs200166313 Chr.7: 76400442-76400442 on Build GRCh38
rs906688545 Chr.7: 76400333-76400333 on Build GRCh38
rs903002424 Chr.7: 76400599-76400599 on Build GRCh38
rs543456048 Chr.7: 76400518-76400518 on Build GRCh38
rs765918525 Chr.7: 76400296-76400296 on Build GRCh38
rs752879864 Chr.7: 76400535-76400535 on Build GRCh38
rs769013776 Chr.7: 76400253-76400253 on Build GRCh38
rs370613189 Chr.7: 76400294-76400297 on Build GRCh38
rs761563199 Chr.7: 76400265-76400265 on Build GRCh38
rs566275931 Chr.7: 76400323-76400323 on Build GRCh38
rs147783985 Chr.7: 76400536-76400536 on Build GRCh38
rs778715074 Chr.7: 76400471-76400471 on Build GRCh38
rs775605306 Chr.7: 76400332-76400332 on Build GRCh38
rs751027721 Chr.7: 76400298-76400298 on Build GRCh38
rs548871030 Chr.7: 76400637-76400637 on Build GRCh38
rs138497833 Chr.7: 76400311-76400311 on Build GRCh38
rs775131403 Chr.7: 76400429-76400429 on Build GRCh38
rs149419007 Chr.7: 76400362-76400362 on Build GRCh38
rs976893549 Chr.7: 76400179-76400179 on Build GRCh38
rs928258142 Chr.7: 76400525-76400525 on Build GRCh38
rs73361552 Chr.7: 76400621-76400621 on Build GRCh38
rs371791239 Chr.7: 76400439-76400439 on Build GRCh38
rs769591656 Chr.7: 76400341-76400341 on Build GRCh38
rs773500648 Chr.7: 76400564-76400564 on Build GRCh38
rs756615057 Chr.7: 76400573-76400573 on Build GRCh38
rs746419002 Chr.7: 76400568-76400569 on Build GRCh38
rs570424679 Chr.7: 76400261-76400261 on Build GRCh38
rs760458748 Chr.7: 76400434-76400434 on Build GRCh38
rs902509208 Chr.7: 76400596-76400596 on Build GRCh38
rs940254897 Chr.7: 76400281-76400281 on Build GRCh38
rs749063727 Chr.7: 76400388-76400388 on Build GRCh38
rs771484204 Chr.7: 76400411-76400411 on Build GRCh38
rs768861095 Chr.7: 76400377-76400377 on Build GRCh38
rs998711354 Chr.7: 76400617-76400618 on Build GRCh38
rs150554690 Chr.7: 76400444-76400444 on Build GRCh38
rs546644248 Chr.7: 76400565-76400565 on Build GRCh38
rs190681385 Chr.7: 76400496-76400496 on Build GRCh38
rs763429612 Chr.7: 76400504-76400504 on Build GRCh38
rs114782962 Chr.7: 76400481-76400481 on Build GRCh38
rs748331950 Chr.7: 76400462-76400462 on Build GRCh38
rs746892123 Chr.7: 76400552-76400552 on Build GRCh38
rs769474587 Chr.7: 76400563-76400563 on Build GRCh38
rs528086616 Chr.7: 76400559-76400559 on Build GRCh38
rs770658473 Chr.7: 76400465-76400465 on Build GRCh38
rs781363674 Chr.7: 76400457-76400457 on Build GRCh38
rs1030742338 Chr.7: 76400303-76400303 on Build GRCh38
rs367686256 Chr.7: 76400308-76400308 on Build GRCh38
rs754721371 Chr.7: 76400301-76400301 on Build GRCh38
rs150028943 Chr.7: 76400202-76400202 on Build GRCh38
rs781500691 Chr.7: 76400417-76400418 on Build GRCh38
rs986164702 Chr.7: 76400329-76400329 on Build GRCh38
rs1043740591 Chr.7: 76400572-76400572 on Build GRCh38
rs747184626 Chr.7: 76400340-76400340 on Build GRCh38
rs146084521 Chr.7: 76400389-76400389 on Build GRCh38
rs774191435 Chr.7: 76400403-76400403 on Build GRCh38
rs144685336 Chr.7: 76400363-76400363 on Build GRCh38
rs1031524507 Chr.7: 76400626-76400626 on Build GRCh38
rs920960543 Chr.7: 76400186-76400186 on Build GRCh38
rs768933328 Chr.7: 76400464-76400464 on Build GRCh38
rs374554032 Chr.7: 76400267-76400267 on Build GRCh38
rs866396771 Chr.7: 76400379-76400379 on Build GRCh38
rs202242854 Chr.7: 76400349-76400349 on Build GRCh38
rs73361550 Chr.7: 76400443-76400443 on Build GRCh38
rs142522233 Chr.7: 76400348-76400348 on Build GRCh38
rs755244124 Chr.7: 76400450-76400450 on Build GRCh38
rs779103440 Chr.7: 76400316-76400316 on Build GRCh38
rs780547967 Chr.7: 76400616-76400617 on Build GRCh38
rs368490025 Chr.7: 76400482-76400482 on Build GRCh38
rs4728712 Chr.7: 76400378-76400378 on Build GRCh38
rs868693716 Chr.7: 76400531-76400531 on Build GRCh38
rs79960636 Chr.7: 76400356-76400356 on Build GRCh38
rs758428000 Chr.7: 76400369-76400369 on Build GRCh38
rs780170759 Chr.7: 76400545-76400545 on Build GRCh38
rs150236892 Chr.7: 76400350-76400350 on Build GRCh38
rs1056666734 Chr.7: 76400579-76400579 on Build GRCh38
rs867255740 Chr.7: 76400412-76400412 on Build GRCh38
rs138202050 Chr.7: 76400374-76400374 on Build GRCh38
rs752423568 Chr.7: 76400352-76400352 on Build GRCh38
rs149742950 Chr.7: 76400494-76400494 on Build GRCh38
rs772926816 Chr.7: 76400342-76400342 on Build GRCh38
rs1011379271 Chr.7: 76400405-76400405 on Build GRCh38

View Full Product Details