Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179102415-179102923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL2
Entrez Gene ID
27
Gene Name
ABL proto-oncogene 2, non-receptor tyrosine kinase
Gene Aliases
ABLL, ARG
UniGene
Hs.159472
Gene Chromosome Location
Chr.1: 179099327-179229693 on Build GRCh38
Transcripts
NM_001136000.2 NM_001168236.1 NM_001168237.1 NM_001168238.1 NM_001168239.1 NM_005158.4 NM_007314.3

SNP Detail

SNP ID Chromosome Location
rs192732952 Chr.1: 179102580-179102580 on Build GRCh38
rs1325194 Chr.1: 179102783-179102783 on Build GRCh38
rs534027820 Chr.1: 179102636-179102636 on Build GRCh38
rs550202884 Chr.1: 179102882-179102882 on Build GRCh38
rs551941773 Chr.1: 179102612-179102612 on Build GRCh38
rs988465407 Chr.1: 179102846-179102846 on Build GRCh38
rs4651012 Chr.1: 179102730-179102730 on Build GRCh38
rs549122723 Chr.1: 179102556-179102556 on Build GRCh38
rs575660498 Chr.1: 179102761-179102761 on Build GRCh38
rs1028068814 Chr.1: 179102807-179102807 on Build GRCh38
rs912044661 Chr.1: 179102563-179102563 on Build GRCh38
rs971138249 Chr.1: 179102883-179102883 on Build GRCh38
rs1043124337 Chr.1: 179102617-179102617 on Build GRCh38
rs890008504 Chr.1: 179102782-179102782 on Build GRCh38
rs529245096 Chr.1: 179102549-179102549 on Build GRCh38
rs945693275 Chr.1: 179102610-179102610 on Build GRCh38
rs981268409 Chr.1: 179102894-179102894 on Build GRCh38
rs187364738 Chr.1: 179102734-179102734 on Build GRCh38
rs957025527 Chr.1: 179102811-179102811 on Build GRCh38
rs957077661 Chr.1: 179102509-179102509 on Build GRCh38
rs149984339 Chr.1: 179102648-179102648 on Build GRCh38
rs981704454 Chr.1: 179102590-179102590 on Build GRCh38
rs115727481 Chr.1: 179102657-179102657 on Build GRCh38
rs1030262989 Chr.1: 179102758-179102758 on Build GRCh38
rs184192725 Chr.1: 179102595-179102595 on Build GRCh38
rs954467653 Chr.1: 179102460-179102460 on Build GRCh38
rs1033303532 Chr.1: 179102799-179102799 on Build GRCh38
rs985121795 Chr.1: 179102487-179102487 on Build GRCh38
rs535394578 Chr.1: 179102458-179102459 on Build GRCh38
rs796847404 Chr.1: 179102672-179102672 on Build GRCh38
rs988516081 Chr.1: 179102545-179102545 on Build GRCh38
rs937269509 Chr.1: 179102596-179102596 on Build GRCh38
rs1325195 Chr.1: 179102621-179102621 on Build GRCh38
rs201369334 Chr.1: 179102792-179102796 on Build GRCh38
rs888448944 Chr.1: 179102618-179102618 on Build GRCh38
rs1055100842 Chr.1: 179102597-179102597 on Build GRCh38
rs560625070 Chr.1: 179102491-179102491 on Build GRCh38
rs949873867 Chr.1: 179102581-179102581 on Build GRCh38
rs192470920 Chr.1: 179102796-179102796 on Build GRCh38
rs1052611659 Chr.1: 179102582-179102582 on Build GRCh38
rs953430583 Chr.1: 179102831-179102831 on Build GRCh38
rs895623196 Chr.1: 179102670-179102670 on Build GRCh38
rs879160734 Chr.1: 179102628-179102628 on Build GRCh38
rs925616146 Chr.1: 179102500-179102500 on Build GRCh38
rs375041123 Chr.1: 179102668-179102668 on Build GRCh38
rs763836609 Chr.1: 179102450-179102450 on Build GRCh38
rs953577893 Chr.1: 179102515-179102515 on Build GRCh38
rs944906311 Chr.1: 179102578-179102578 on Build GRCh38
rs1001672601 Chr.1: 179102640-179102640 on Build GRCh38
rs761720167 Chr.1: 179102468-179102468 on Build GRCh38

View Full Product Details