Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 90844172-90844637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYK
Entrez Gene ID
6850
Gene Name
spleen associated tyrosine kinase
Gene Aliases
p72-Syk
UniGene
Hs.371720
Gene Chromosome Location
Chr.9: 90801680-90898560 on Build GRCh38
Transcripts
NM_001135052.3 NM_001174167.2 NM_001174168.2 NM_003177.6 XM_005252147.3 XM_011518946.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7185261 c.343C>A p.P115T Chr.9: 90844241-90844241 on Build GRCh38
6928079 c.281A>G p.Q94R Chr.9: 90844179-90844179 on Build GRCh38
4880166 c.302G>T p.C101F Chr.9: 90844200-90844200 on Build GRCh38
5727283 c.287C>T p.S96F Chr.9: 90844185-90844185 on Build GRCh38
346152 c.315G>T p.K105N Chr.9: 90844213-90844213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945776814 Chr.9: 90844464-90844464 on Build GRCh38
rs201640567 Chr.9: 90844333-90844333 on Build GRCh38
rs375028044 Chr.9: 90844227-90844227 on Build GRCh38
rs12555502 Chr.9: 90844549-90844549 on Build GRCh38
rs766024718 Chr.9: 90844319-90844319 on Build GRCh38
rs567269671 Chr.9: 90844529-90844529 on Build GRCh38
rs200097669 Chr.9: 90844308-90844308 on Build GRCh38
rs147465567 Chr.9: 90844351-90844351 on Build GRCh38
rs765252567 Chr.9: 90844298-90844298 on Build GRCh38
rs986133570 Chr.9: 90844380-90844380 on Build GRCh38
rs963109544 Chr.9: 90844327-90844327 on Build GRCh38
rs368093787 Chr.9: 90844335-90844335 on Build GRCh38
rs960287440 Chr.9: 90844199-90844199 on Build GRCh38
rs551467167 Chr.9: 90844393-90844393 on Build GRCh38
rs571653395 Chr.9: 90844290-90844290 on Build GRCh38
rs528230417 Chr.9: 90844385-90844385 on Build GRCh38
rs904282596 Chr.9: 90844496-90844496 on Build GRCh38
rs746718066 Chr.9: 90844199-90844200 on Build GRCh38
rs752276138 Chr.9: 90844210-90844210 on Build GRCh38
rs962420195 Chr.9: 90844262-90844262 on Build GRCh38
rs947435354 Chr.9: 90844414-90844414 on Build GRCh38
rs767306766 Chr.9: 90844331-90844331 on Build GRCh38
rs746010260 Chr.9: 90844239-90844239 on Build GRCh38
rs1042849767 Chr.9: 90844470-90844470 on Build GRCh38
rs775510434 Chr.9: 90844287-90844287 on Build GRCh38
rs550638338 Chr.9: 90844460-90844460 on Build GRCh38
rs375940286 Chr.9: 90844363-90844363 on Build GRCh38
rs896048097 Chr.9: 90844600-90844600 on Build GRCh38
rs759213202 Chr.9: 90844323-90844323 on Build GRCh38
rs779088381 Chr.9: 90844355-90844355 on Build GRCh38
rs754686051 Chr.9: 90844200-90844200 on Build GRCh38
rs776902749 Chr.9: 90844365-90844366 on Build GRCh38
rs772293286 Chr.9: 90844246-90844246 on Build GRCh38
rs1005030927 Chr.9: 90844575-90844575 on Build GRCh38
rs751386077 Chr.9: 90844320-90844320 on Build GRCh38
rs142980255 Chr.9: 90844312-90844312 on Build GRCh38
rs913091043 Chr.9: 90844442-90844442 on Build GRCh38
rs756646754 Chr.9: 90844224-90844224 on Build GRCh38
rs780794130 Chr.9: 90844207-90844207 on Build GRCh38
rs747578282 Chr.9: 90844566-90844566 on Build GRCh38
rs933242907 Chr.9: 90844550-90844550 on Build GRCh38
rs746263659 Chr.9: 90844361-90844361 on Build GRCh38
rs886282026 Chr.9: 90844573-90844573 on Build GRCh38
rs200111713 Chr.9: 90844338-90844338 on Build GRCh38
rs900319493 Chr.9: 90844219-90844219 on Build GRCh38
rs1038958031 Chr.9: 90844513-90844513 on Build GRCh38
rs988946678 Chr.9: 90844402-90844402 on Build GRCh38
rs202043759 Chr.9: 90844226-90844226 on Build GRCh38
rs773009850 Chr.9: 90844309-90844309 on Build GRCh38
rs139937468 Chr.9: 90844556-90844556 on Build GRCh38
rs267602311 Chr.9: 90844271-90844271 on Build GRCh38
rs914556980 Chr.9: 90844407-90844407 on Build GRCh38
rs2278275 Chr.9: 90844394-90844394 on Build GRCh38
rs530615396 Chr.9: 90844431-90844431 on Build GRCh38
rs865783333 Chr.9: 90844305-90844305 on Build GRCh38
rs577986649 Chr.9: 90844195-90844195 on Build GRCh38
rs200453337 Chr.9: 90844209-90844209 on Build GRCh38
rs143342183 Chr.9: 90844234-90844234 on Build GRCh38
rs747578597 Chr.9: 90844348-90844348 on Build GRCh38
rs943951250 Chr.9: 90844392-90844392 on Build GRCh38

View Full Product Details