Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101592784-101593292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN3A
Entrez Gene ID
116443
Gene Name
glutamate ionotropic receptor NMDA type subunit 3A
Gene Aliases
GluN3A, NMDAR-L, NR3A
UniGene
Hs.654783
Gene Chromosome Location
Chr.9: 101569352-101738580 on Build GRCh38
Transcripts
NM_133445.2 NM_147180.3

Gene Symbol
PPP3R2
Entrez Gene ID
5535
Gene Name
protein phosphatase 3 regulatory subunit B, beta
Gene Aliases
PPP3RL
UniGene
Hs.151167
Gene Chromosome Location
Chr.9: 101591615-101595001 on Build GRCh38
Transcripts
NM_133445.2 NM_147180.3

SNP Detail

SNP ID Chromosome Location
rs908597623 Chr.9: 101592931-101592931 on Build GRCh38
rs575428378 Chr.9: 101593258-101593258 on Build GRCh38
rs548336737 Chr.9: 101592996-101592996 on Build GRCh38
rs1022106678 Chr.9: 101593086-101593086 on Build GRCh38
rs763080895 Chr.9: 101592975-101592975 on Build GRCh38
rs73659533 Chr.9: 101592859-101592859 on Build GRCh38
rs933991506 Chr.9: 101592914-101592914 on Build GRCh38
rs546929861 Chr.9: 101592809-101592809 on Build GRCh38
rs113160687 Chr.9: 101593186-101593186 on Build GRCh38
rs1053573852 Chr.9: 101593000-101593000 on Build GRCh38
rs868767873 Chr.9: 101593017-101593017 on Build GRCh38
rs1021052879 Chr.9: 101593180-101593180 on Build GRCh38
rs1002362338 Chr.9: 101593090-101593090 on Build GRCh38
rs923212409 Chr.9: 101592870-101592870 on Build GRCh38
rs898666019 Chr.9: 101593029-101593029 on Build GRCh38
rs1030227118 Chr.9: 101593263-101593263 on Build GRCh38
rs879024276 Chr.9: 101593060-101593060 on Build GRCh38
rs908282719 Chr.9: 101593179-101593179 on Build GRCh38
rs745826284 Chr.9: 101593100-101593100 on Build GRCh38
rs568502989 Chr.9: 101593124-101593124 on Build GRCh38
rs1407877 Chr.9: 101593173-101593173 on Build GRCh38
rs552161687 Chr.9: 101593035-101593035 on Build GRCh38
rs1055368835 Chr.9: 101593115-101593115 on Build GRCh38
rs940118421 Chr.9: 101592960-101592960 on Build GRCh38
rs964583402 Chr.9: 101592839-101592839 on Build GRCh38
rs925114645 Chr.9: 101592823-101592823 on Build GRCh38
rs912821914 Chr.9: 101593259-101593259 on Build GRCh38
rs117276735 Chr.9: 101592929-101592929 on Build GRCh38
rs921461526 Chr.9: 101593215-101593215 on Build GRCh38
rs1051082604 Chr.9: 101592889-101592889 on Build GRCh38
rs577040128 Chr.9: 101593188-101593188 on Build GRCh38
rs556346287 Chr.9: 101593225-101593225 on Build GRCh38
rs1000618850 Chr.9: 101593210-101593210 on Build GRCh38
rs974599749 Chr.9: 101593248-101593248 on Build GRCh38
rs1011154232 Chr.9: 101593081-101593081 on Build GRCh38
rs969263763 Chr.9: 101593089-101593089 on Build GRCh38
rs570465880 Chr.9: 101593171-101593171 on Build GRCh38
rs1051325316 Chr.9: 101593226-101593226 on Build GRCh38
rs779915417 Chr.9: 101593241-101593241 on Build GRCh38

View Full Product Details