Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153349543-153350044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF275
Entrez Gene ID
10838
Gene Name
zinc finger protein 275
Gene Aliases
-
UniGene
Hs.348963
Gene Chromosome Location
Chr.X: 153334155-153352926 on Build GRCh38
Transcripts
NM_001080485.3

SNP Detail

SNP ID Chromosome Location
rs569929141 Chr.X: 153349851-153349851 on Build GRCh38
rs908653784 Chr.X: 153349997-153349997 on Build GRCh38
rs531587901 Chr.X: 153349819-153349819 on Build GRCh38
rs782299349 Chr.X: 153349850-153349850 on Build GRCh38
rs782184576 Chr.X: 153349833-153349833 on Build GRCh38
rs782688521 Chr.X: 153349792-153349792 on Build GRCh38
rs782491703 Chr.X: 153349670-153349670 on Build GRCh38
rs191734703 Chr.X: 153349572-153349572 on Build GRCh38
rs782780005 Chr.X: 153349573-153349573 on Build GRCh38
rs904359189 Chr.X: 153349773-153349773 on Build GRCh38
rs899812729 Chr.X: 153349629-153349629 on Build GRCh38
rs944124605 Chr.X: 153350014-153350014 on Build GRCh38
rs931510730 Chr.X: 153349911-153349911 on Build GRCh38
rs1022604605 Chr.X: 153349798-153349798 on Build GRCh38
rs144519696 Chr.X: 153349685-153349685 on Build GRCh38
rs992477119 Chr.X: 153349796-153349796 on Build GRCh38
rs782256066 Chr.X: 153349801-153349801 on Build GRCh38
rs781799037 Chr.X: 153349787-153349787 on Build GRCh38
rs572620335 Chr.X: 153349799-153349799 on Build GRCh38
rs782602541 Chr.X: 153349917-153349917 on Build GRCh38
rs148426956 Chr.X: 153349791-153349791 on Build GRCh38
rs1021509868 Chr.X: 153349764-153349764 on Build GRCh38
rs782031644 Chr.X: 153349933-153349933 on Build GRCh38
rs181816143 Chr.X: 153349589-153349589 on Build GRCh38
rs782276997 Chr.X: 153349947-153349947 on Build GRCh38
rs717303 Chr.X: 153349742-153349742 on Build GRCh38

View Full Product Details