Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66362606-66363144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724064
Entrez Gene ID
102724064
Gene Name
uncharacterized LOC102724064
Gene Aliases
-
UniGene
Hs.614641
Gene Chromosome Location
Chr.11: 66347991-66363764 on Build GRCh38
Transcripts
NR_135761.1 NM_001300868.1 NM_001300869.1 NM_001532.2 NR_125343.1 XM_011544974.1 XM_017017630.1 XM_017017632.1 XM_017017635.1 XM_017017637.1 XM_017017638.1

Gene Symbol
SLC29A2
Entrez Gene ID
3177
Gene Name
solute carrier family 29 member 2
Gene Aliases
DER12, ENT2, HNP36
UniGene
Hs.569017
Gene Chromosome Location
Chr.11: 66362521-66372476 on Build GRCh38
Transcripts
NR_135761.1 NM_001300868.1 NM_001300869.1 NM_001532.2 NR_125343.1 XM_011544974.1 XM_017017630.1 XM_017017632.1 XM_017017635.1 XM_017017637.1 XM_017017638.1

SNP Detail

SNP ID Chromosome Location
rs896598337 Chr.11: 66362982-66362982 on Build GRCh38
rs985019875 Chr.11: 66362810-66362810 on Build GRCh38
rs532108020 Chr.11: 66362642-66362642 on Build GRCh38
rs971808403 Chr.11: 66362853-66362853 on Build GRCh38
rs3177514 Chr.11: 66362887-66362887 on Build GRCh38
rs1014069119 Chr.11: 66362996-66362996 on Build GRCh38
rs906452525 Chr.11: 66363111-66363111 on Build GRCh38
rs961051346 Chr.11: 66362938-66362938 on Build GRCh38
rs536523852 Chr.11: 66363099-66363099 on Build GRCh38
rs753102720 Chr.11: 66362865-66362865 on Build GRCh38
rs1047029 Chr.11: 66363077-66363077 on Build GRCh38
rs936031146 Chr.11: 66362749-66362749 on Build GRCh38
rs913302635 Chr.11: 66363094-66363094 on Build GRCh38
rs116864252 Chr.11: 66363006-66363006 on Build GRCh38
rs188481560 Chr.11: 66362699-66362699 on Build GRCh38
rs149792941 Chr.11: 66362750-66362750 on Build GRCh38
rs145780180 Chr.11: 66362826-66362826 on Build GRCh38
rs1003823041 Chr.11: 66362872-66362872 on Build GRCh38
rs947324633 Chr.11: 66362895-66362895 on Build GRCh38
rs946070470 Chr.11: 66363096-66363096 on Build GRCh38
rs764137872 Chr.11: 66362903-66362903 on Build GRCh38
rs922858941 Chr.11: 66362689-66362689 on Build GRCh38
rs537860035 Chr.11: 66362897-66362897 on Build GRCh38
rs751350402 Chr.11: 66362989-66362989 on Build GRCh38
rs903032186 Chr.11: 66362937-66362937 on Build GRCh38
rs556194094 Chr.11: 66362886-66362886 on Build GRCh38
rs570066211 Chr.11: 66362746-66362746 on Build GRCh38
rs1010229948 Chr.11: 66362793-66362793 on Build GRCh38
rs1042894633 Chr.11: 66362676-66362676 on Build GRCh38
rs961074068 Chr.11: 66362708-66362708 on Build GRCh38
rs879308756 Chr.11: 66363076-66363077 on Build GRCh38
rs1024044695 Chr.11: 66362797-66362797 on Build GRCh38
rs969692434 Chr.11: 66362800-66362801 on Build GRCh38
rs529299355 Chr.11: 66363028-66363028 on Build GRCh38
rs889683314 Chr.11: 66362962-66362963 on Build GRCh38
rs116183086 Chr.11: 66362881-66362881 on Build GRCh38
rs891773471 Chr.11: 66362768-66362768 on Build GRCh38
rs749286699 Chr.11: 66362691-66362691 on Build GRCh38
rs756859035 Chr.11: 66362689-66362692 on Build GRCh38
rs367972776 Chr.11: 66363014-66363014 on Build GRCh38
rs761438578 Chr.11: 66362978-66362978 on Build GRCh38
rs562614637 Chr.11: 66363114-66363116 on Build GRCh38
rs933055067 Chr.11: 66362880-66362880 on Build GRCh38
rs192877986 Chr.11: 66362896-66362896 on Build GRCh38
rs902999760 Chr.11: 66362677-66362677 on Build GRCh38
rs905167514 Chr.11: 66363030-66363030 on Build GRCh38
rs911369160 Chr.11: 66363016-66363016 on Build GRCh38
rs909514737 Chr.11: 66362671-66362671 on Build GRCh38
rs201724518 Chr.11: 66363011-66363011 on Build GRCh38
rs560320262 Chr.11: 66363026-66363026 on Build GRCh38
rs983761635 Chr.11: 66362660-66362660 on Build GRCh38
rs567330411 Chr.11: 66362877-66362877 on Build GRCh38
rs879298207 Chr.11: 66362888-66362888 on Build GRCh38
rs3741366 Chr.11: 66363056-66363056 on Build GRCh38
rs962172163 Chr.11: 66362840-66362840 on Build GRCh38
rs972987223 Chr.11: 66362680-66362680 on Build GRCh38
rs1049915233 Chr.11: 66362703-66362703 on Build GRCh38
rs11227466 Chr.11: 66363009-66363009 on Build GRCh38
rs551853990 Chr.11: 66362679-66362679 on Build GRCh38
rs922877254 Chr.11: 66362987-66362987 on Build GRCh38

View Full Product Details