Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74978441-74979010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPCS2
Entrez Gene ID
9789
Gene Name
signal peptidase complex subunit 2
Gene Aliases
-
UniGene
Hs.282700
Gene Chromosome Location
Chr.11: 74949247-74979031 on Build GRCh38
Transcripts
NM_014752.2

SNP Detail

SNP ID Chromosome Location
rs1001287141 Chr.11: 74978903-74978903 on Build GRCh38
rs781082524 Chr.11: 74978976-74978976 on Build GRCh38
rs940213192 Chr.11: 74978666-74978666 on Build GRCh38
rs868338052 Chr.11: 74978974-74978974 on Build GRCh38
rs977697512 Chr.11: 74978873-74978873 on Build GRCh38
rs1040967186 Chr.11: 74978808-74978808 on Build GRCh38
rs1027346841 Chr.11: 74978587-74978587 on Build GRCh38
rs765739098 Chr.11: 74978582-74978582 on Build GRCh38
rs962138059 Chr.11: 74978530-74978530 on Build GRCh38
rs953013467 Chr.11: 74978604-74978604 on Build GRCh38
rs757916789 Chr.11: 74978626-74978626 on Build GRCh38
rs113229311 Chr.11: 74978971-74978971 on Build GRCh38
rs1010545454 Chr.11: 74978853-74978853 on Build GRCh38
rs754620365 Chr.11: 74978948-74978948 on Build GRCh38
rs574102856 Chr.11: 74978741-74978741 on Build GRCh38
rs371734594 Chr.11: 74978938-74978938 on Build GRCh38
rs891690450 Chr.11: 74978824-74978824 on Build GRCh38
rs968921172 Chr.11: 74978902-74978902 on Build GRCh38
rs1003456015 Chr.11: 74978786-74978786 on Build GRCh38
rs192548097 Chr.11: 74978668-74978668 on Build GRCh38
rs1010055911 Chr.11: 74978510-74978510 on Build GRCh38
rs369427124 Chr.11: 74978678-74978678 on Build GRCh38
rs116386889 Chr.11: 74978580-74978580 on Build GRCh38
rs1014726918 Chr.11: 74978522-74978522 on Build GRCh38
rs992451147 Chr.11: 74978639-74978639 on Build GRCh38
rs906430174 Chr.11: 74978488-74978488 on Build GRCh38
rs200257038 Chr.11: 74978553-74978554 on Build GRCh38
rs931428615 Chr.11: 74978740-74978740 on Build GRCh38
rs1019485813 Chr.11: 74978475-74978475 on Build GRCh38
rs1015212751 Chr.11: 74978795-74978795 on Build GRCh38
rs966727816 Chr.11: 74978873-74978873 on Build GRCh38
rs551593089 Chr.11: 74978807-74978807 on Build GRCh38
rs565121735 Chr.11: 74978933-74978933 on Build GRCh38
rs184227821 Chr.11: 74978755-74978755 on Build GRCh38
rs538015301 Chr.11: 74978485-74978486 on Build GRCh38
rs956098950 Chr.11: 74978479-74978479 on Build GRCh38
rs772978652 Chr.11: 74978492-74978492 on Build GRCh38
rs1008173883 Chr.11: 74978796-74978796 on Build GRCh38
rs1021472038 Chr.11: 74978889-74978889 on Build GRCh38
rs116956418 Chr.11: 74978936-74978936 on Build GRCh38
rs912168501 Chr.11: 74978616-74978617 on Build GRCh38
rs959654662 Chr.11: 74978638-74978638 on Build GRCh38
rs563005935 Chr.11: 74978960-74978960 on Build GRCh38
rs916013850 Chr.11: 74978904-74978904 on Build GRCh38
rs1036269399 Chr.11: 74978968-74978968 on Build GRCh38
rs1027421199 Chr.11: 74978822-74978822 on Build GRCh38
rs75021616 Chr.11: 74978605-74978605 on Build GRCh38
rs547993291 Chr.11: 74978903-74978904 on Build GRCh38
rs918166242 Chr.11: 74978642-74978642 on Build GRCh38
rs1054525080 Chr.11: 74978680-74978680 on Build GRCh38
rs146058631 Chr.11: 74978486-74978487 on Build GRCh38
rs979818949 Chr.11: 74978619-74978619 on Build GRCh38
rs1018590655 Chr.11: 74978872-74978872 on Build GRCh38
rs939288357 Chr.11: 74978951-74978951 on Build GRCh38
rs948735450 Chr.11: 74978723-74978723 on Build GRCh38
rs920067457 Chr.11: 74978712-74978712 on Build GRCh38

View Full Product Details