Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132147519-132148009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOC4L
Entrez Gene ID
79050
Gene Name
nucleolar complex associated 4 homolog
Gene Aliases
NET49, NOC4, UTP19
UniGene
Hs.558536
Gene Chromosome Location
Chr.12: 132144421-132152473 on Build GRCh38
Transcripts
NM_024078.2 XM_017019960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763474 c.657G>T p.L219L Chr.12: 132147933-132147933 on Build GRCh38
5933097 c.476C>T p.S159F Chr.12: 132147655-132147655 on Build GRCh38
6727583 c.532G>A p.D178N Chr.12: 132147711-132147711 on Build GRCh38
6851011 c.513C>T p.F171F Chr.12: 132147692-132147692 on Build GRCh38
5980433 c.514C>T p.R172W Chr.12: 132147693-132147693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054887504 Chr.12: 132147574-132147574 on Build GRCh38
rs751420540 Chr.12: 132147739-132147739 on Build GRCh38
rs772591655 Chr.12: 132147780-132147780 on Build GRCh38
rs539539647 Chr.12: 132147588-132147588 on Build GRCh38
rs1038032636 Chr.12: 132147865-132147865 on Build GRCh38
rs911207342 Chr.12: 132147933-132147933 on Build GRCh38
rs370177948 Chr.12: 132147639-132147639 on Build GRCh38
rs547422920 Chr.12: 132147561-132147561 on Build GRCh38
rs375410621 Chr.12: 132147614-132147614 on Build GRCh38
rs775006630 Chr.12: 132147776-132147776 on Build GRCh38
rs375636244 Chr.12: 132147844-132147844 on Build GRCh38
rs78636093 Chr.12: 132147622-132147622 on Build GRCh38
rs772671540 Chr.12: 132147858-132147858 on Build GRCh38
rs766224169 Chr.12: 132147948-132147948 on Build GRCh38
rs370828767 Chr.12: 132147924-132147924 on Build GRCh38
rs202227049 Chr.12: 132147693-132147693 on Build GRCh38
rs377613547 Chr.12: 132147817-132147817 on Build GRCh38
rs771339553 Chr.12: 132147934-132147934 on Build GRCh38
rs776828332 Chr.12: 132147702-132147702 on Build GRCh38
rs776810673 Chr.12: 132147935-132147935 on Build GRCh38
rs771780811 Chr.12: 132147625-132147625 on Build GRCh38
rs746035373 Chr.12: 132147709-132147709 on Build GRCh38
rs901327119 Chr.12: 132147850-132147850 on Build GRCh38
rs761788217 Chr.12: 132147883-132147883 on Build GRCh38
rs202070598 Chr.12: 132147977-132147977 on Build GRCh38
rs374292708 Chr.12: 132147852-132147852 on Build GRCh38
rs561943999 Chr.12: 132147553-132147553 on Build GRCh38
rs148483243 Chr.12: 132147974-132147974 on Build GRCh38
rs750253485 Chr.12: 132147889-132147889 on Build GRCh38
rs780906260 Chr.12: 132147744-132147744 on Build GRCh38
rs768385174 Chr.12: 132147591-132147591 on Build GRCh38
rs112840884 Chr.12: 132147730-132147730 on Build GRCh38
rs748786699 Chr.12: 132147610-132147610 on Build GRCh38
rs775268114 Chr.12: 132147695-132147695 on Build GRCh38
rs763825435 Chr.12: 132147759-132147759 on Build GRCh38
rs146645255 Chr.12: 132147688-132147688 on Build GRCh38
rs754138548 Chr.12: 132147912-132147912 on Build GRCh38
rs537280336 Chr.12: 132147802-132147802 on Build GRCh38
rs764585370 Chr.12: 132147660-132147660 on Build GRCh38
rs779091480 Chr.12: 132147851-132147851 on Build GRCh38
rs143756467 Chr.12: 132147747-132147747 on Build GRCh38
rs568405731 Chr.12: 132147653-132147653 on Build GRCh38
rs531572514 Chr.12: 132147870-132147870 on Build GRCh38
rs372807012 Chr.12: 132147952-132147952 on Build GRCh38
rs373772458 Chr.12: 132147803-132147803 on Build GRCh38
rs200918523 Chr.12: 132147940-132147940 on Build GRCh38
rs778433035 Chr.12: 132147678-132147678 on Build GRCh38
rs1045588864 Chr.12: 132147841-132147842 on Build GRCh38
rs773810494 Chr.12: 132147876-132147876 on Build GRCh38
rs753458687 Chr.12: 132147673-132147673 on Build GRCh38
rs775210227 Chr.12: 132147874-132147874 on Build GRCh38
rs990570114 Chr.12: 132147903-132147903 on Build GRCh38
rs766118564 Chr.12: 132147670-132147670 on Build GRCh38
rs369215601 Chr.12: 132147720-132147720 on Build GRCh38
rs372575646 Chr.12: 132147642-132147642 on Build GRCh38
rs201466638 Chr.12: 132147925-132147925 on Build GRCh38
rs949896151 Chr.12: 132147826-132147826 on Build GRCh38
rs370139734 Chr.12: 132147843-132147843 on Build GRCh38
rs199530046 Chr.12: 132147848-132147848 on Build GRCh38
rs779753235 Chr.12: 132147606-132147606 on Build GRCh38
rs549575909 Chr.12: 132147592-132147592 on Build GRCh38
rs777672361 Chr.12: 132147736-132147736 on Build GRCh38
rs755214754 Chr.12: 132147913-132147913 on Build GRCh38
rs755304546 Chr.12: 132147951-132147951 on Build GRCh38
rs994452973 Chr.12: 132147753-132147753 on Build GRCh38
rs989456064 Chr.12: 132147932-132147932 on Build GRCh38
rs374084936 Chr.12: 132147589-132147589 on Build GRCh38
rs766872737 Chr.12: 132147944-132147944 on Build GRCh38
rs149306359 Chr.12: 132147967-132147967 on Build GRCh38
rs531462202 Chr.12: 132147797-132147797 on Build GRCh38
rs139503412 Chr.12: 132147954-132147954 on Build GRCh38
rs570414274 Chr.12: 132147587-132147587 on Build GRCh38
rs755943915 Chr.12: 132147896-132147896 on Build GRCh38
rs541301992 Chr.12: 132147866-132147866 on Build GRCh38
rs761309796 Chr.12: 132147941-132147941 on Build GRCh38
rs747406434 Chr.12: 132147930-132147930 on Build GRCh38
rs116596143 Chr.12: 132147600-132147600 on Build GRCh38
rs773494712 Chr.12: 132147861-132147861 on Build GRCh38
rs1015895989 Chr.12: 132147885-132147885 on Build GRCh38
rs866243929 Chr.12: 132147768-132147768 on Build GRCh38
rs752895441 Chr.12: 132147959-132147959 on Build GRCh38
rs765638511 Chr.12: 132147583-132147583 on Build GRCh38
rs769356870 Chr.12: 132147766-132147766 on Build GRCh38
rs777658725 Chr.12: 132147966-132147966 on Build GRCh38
rs755881673 Chr.12: 132147663-132147663 on Build GRCh38
rs201201651 Chr.12: 132147601-132147601 on Build GRCh38
rs758311489 Chr.12: 132147684-132147684 on Build GRCh38
rs780888985 Chr.12: 132147842-132147842 on Build GRCh38
rs751510422 Chr.12: 132147821-132147821 on Build GRCh38
rs193175644 Chr.12: 132147849-132147849 on Build GRCh38
rs199638081 Chr.12: 132147713-132147713 on Build GRCh38
rs760477400 Chr.12: 132147947-132147947 on Build GRCh38
rs889875632 Chr.12: 132147637-132147637 on Build GRCh38
rs935917660 Chr.12: 132147822-132147822 on Build GRCh38
rs764680094 Chr.12: 132147729-132147729 on Build GRCh38
rs11543303 Chr.12: 132147920-132147920 on Build GRCh38
rs773216825 Chr.12: 132147945-132147945 on Build GRCh38
rs751943168 Chr.12: 132147662-132147662 on Build GRCh38
rs749375574 Chr.12: 132147760-132147760 on Build GRCh38
rs767928976 Chr.12: 132147718-132147718 on Build GRCh38
rs762266021 Chr.12: 132147716-132147716 on Build GRCh38
rs527322731 Chr.12: 132147554-132147554 on Build GRCh38
rs747258590 Chr.12: 132147628-132147628 on Build GRCh38
rs775865033 Chr.12: 132147648-132147648 on Build GRCh38
rs112569492 Chr.12: 132147541-132147541 on Build GRCh38
rs911206887 Chr.12: 132147630-132147630 on Build GRCh38
rs771199488 Chr.12: 132147694-132147694 on Build GRCh38
rs148171235 Chr.12: 132147752-132147752 on Build GRCh38
rs775599091 Chr.12: 132147711-132147711 on Build GRCh38
rs375850317 Chr.12: 132147989-132147989 on Build GRCh38
rs748055378 Chr.12: 132147623-132147623 on Build GRCh38
rs141988623 Chr.12: 132147911-132147911 on Build GRCh38
rs748054380 Chr.12: 132147681-132147681 on Build GRCh38
rs763526045 Chr.12: 132147820-132147820 on Build GRCh38
rs754454799 Chr.12: 132147675-132147675 on Build GRCh38
rs771107725 Chr.12: 132147633-132147633 on Build GRCh38
rs185086751 Chr.12: 132147888-132147888 on Build GRCh38
rs994411269 Chr.12: 132147689-132147689 on Build GRCh38
rs1025805787 Chr.12: 132147774-132147774 on Build GRCh38
rs147903181 Chr.12: 132147710-132147710 on Build GRCh38
rs199897574 Chr.12: 132147706-132147706 on Build GRCh38
rs772554226 Chr.12: 132147982-132147982 on Build GRCh38
rs752203541 Chr.12: 132147811-132147811 on Build GRCh38
rs989157740 Chr.12: 132147823-132147823 on Build GRCh38
rs548061561 Chr.12: 132147973-132147973 on Build GRCh38
rs937845474 Chr.12: 132147569-132147569 on Build GRCh38
rs768363543 Chr.12: 132147976-132147976 on Build GRCh38
rs142341531 Chr.12: 132147937-132147937 on Build GRCh38
rs748212526 Chr.12: 132147923-132147923 on Build GRCh38
rs758376800 Chr.12: 132147735-132147735 on Build GRCh38
rs986891130 Chr.12: 132147786-132147786 on Build GRCh38
rs142447795 Chr.12: 132147636-132147636 on Build GRCh38
rs759182697 Chr.12: 132147721-132147721 on Build GRCh38
rs913053056 Chr.12: 132147824-132147824 on Build GRCh38
rs774916512 Chr.12: 132147714-132147714 on Build GRCh38
rs145983450 Chr.12: 132147950-132147950 on Build GRCh38
rs779169383 Chr.12: 132147918-132147918 on Build GRCh38
rs556368580 Chr.12: 132147743-132147743 on Build GRCh38
rs760703595 Chr.12: 132147984-132147984 on Build GRCh38
rs368218530 Chr.12: 132147875-132147875 on Build GRCh38
rs757136625 Chr.12: 132147837-132147837 on Build GRCh38
rs758512094 Chr.12: 132147965-132147965 on Build GRCh38
rs370940839 Chr.12: 132147978-132147978 on Build GRCh38
rs781620822 Chr.12: 132147582-132147583 on Build GRCh38
rs762355865 Chr.12: 132147779-132147779 on Build GRCh38

View Full Product Details