Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26184211-26184715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR1
Entrez Gene ID
10256
Gene Name
connector enhancer of kinase suppressor of Ras 1
Gene Aliases
CNK, CNK1
UniGene
Hs.16232
Gene Chromosome Location
Chr.1: 26177490-26189884 on Build GRCh38
Transcripts
NM_001297647.1 NM_001297648.1 NM_006314.2 XM_011540484.2 XM_011540485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5907860 c.1060C>T p.P354S Chr.1: 26184460-26184460 on Build GRCh38
4030935 c.987C>T p.S329S Chr.1: 26184274-26184274 on Build GRCh38
5903316 c.1039C>T p.P347S Chr.1: 26184439-26184439 on Build GRCh38
1687232 c.1037C>T p.P346L Chr.1: 26184437-26184437 on Build GRCh38
6866634 c.1067G>T p.G356V Chr.1: 26184467-26184467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147047319 Chr.1: 26184283-26184283 on Build GRCh38
rs757826978 Chr.1: 26184237-26184237 on Build GRCh38
rs149576633 Chr.1: 26184440-26184440 on Build GRCh38
rs768985826 Chr.1: 26184414-26184414 on Build GRCh38
rs755110824 Chr.1: 26184595-26184595 on Build GRCh38
rs190132337 Chr.1: 26184233-26184233 on Build GRCh38
rs767002209 Chr.1: 26184587-26184587 on Build GRCh38
rs759026253 Chr.1: 26184485-26184485 on Build GRCh38
rs754282674 Chr.1: 26184350-26184350 on Build GRCh38
rs746008112 Chr.1: 26184265-26184265 on Build GRCh38
rs767991327 Chr.1: 26184537-26184537 on Build GRCh38
rs1031304351 Chr.1: 26184333-26184333 on Build GRCh38
rs537321990 Chr.1: 26184315-26184315 on Build GRCh38
rs767457352 Chr.1: 26184597-26184597 on Build GRCh38
rs1048478305 Chr.1: 26184264-26184264 on Build GRCh38
rs772209035 Chr.1: 26184266-26184266 on Build GRCh38
rs780052438 Chr.1: 26184635-26184635 on Build GRCh38
rs751291867 Chr.1: 26184434-26184435 on Build GRCh38
rs372015710 Chr.1: 26184558-26184558 on Build GRCh38
rs111870124 Chr.1: 26184321-26184321 on Build GRCh38
rs557328646 Chr.1: 26184349-26184349 on Build GRCh38
rs748271342 Chr.1: 26184521-26184521 on Build GRCh38
rs753362199 Chr.1: 26184465-26184465 on Build GRCh38
rs370866944 Chr.1: 26184371-26184371 on Build GRCh38
rs780698052 Chr.1: 26184486-26184486 on Build GRCh38
rs762249869 Chr.1: 26184469-26184469 on Build GRCh38
rs376326724 Chr.1: 26184629-26184629 on Build GRCh38
rs866662054 Chr.1: 26184248-26184248 on Build GRCh38
rs754950342 Chr.1: 26184507-26184507 on Build GRCh38
rs760415083 Chr.1: 26184320-26184320 on Build GRCh38
rs779270949 Chr.1: 26184242-26184242 on Build GRCh38
rs780324234 Chr.1: 26184378-26184378 on Build GRCh38
rs759061601 Chr.1: 26184314-26184314 on Build GRCh38
rs748648581 Chr.1: 26184299-26184299 on Build GRCh38
rs781267315 Chr.1: 26184417-26184417 on Build GRCh38
rs999478093 Chr.1: 26184329-26184329 on Build GRCh38
rs749115087 Chr.1: 26184425-26184425 on Build GRCh38
rs1010814103 Chr.1: 26184589-26184589 on Build GRCh38
rs1003262667 Chr.1: 26184624-26184624 on Build GRCh38
rs370166938 Chr.1: 26184577-26184577 on Build GRCh38
rs999908148 Chr.1: 26184562-26184562 on Build GRCh38
rs369925886 Chr.1: 26184290-26184290 on Build GRCh38
rs766036204 Chr.1: 26184402-26184403 on Build GRCh38
rs867995657 Chr.1: 26184410-26184410 on Build GRCh38
rs368701597 Chr.1: 26184532-26184532 on Build GRCh38
rs370735726 Chr.1: 26184531-26184531 on Build GRCh38
rs148237566 Chr.1: 26184441-26184441 on Build GRCh38
rs1049073322 Chr.1: 26184508-26184508 on Build GRCh38
rs201813702 Chr.1: 26184474-26184474 on Build GRCh38
rs771965978 Chr.1: 26184446-26184446 on Build GRCh38
rs762582924 Chr.1: 26184248-26184248 on Build GRCh38
rs752376171 Chr.1: 26184317-26184317 on Build GRCh38
rs770741751 Chr.1: 26184526-26184526 on Build GRCh38
rs776366879 Chr.1: 26184539-26184539 on Build GRCh38
rs759361876 Chr.1: 26184528-26184529 on Build GRCh38
rs756374440 Chr.1: 26184618-26184618 on Build GRCh38
rs558527518 Chr.1: 26184471-26184471 on Build GRCh38
rs763632608 Chr.1: 26184337-26184337 on Build GRCh38
rs1019664538 Chr.1: 26184303-26184303 on Build GRCh38
rs80305988 Chr.1: 26184529-26184529 on Build GRCh38
rs139381171 Chr.1: 26184274-26184274 on Build GRCh38
rs546385304 Chr.1: 26184524-26184524 on Build GRCh38
rs373382597 Chr.1: 26184326-26184326 on Build GRCh38
rs747067656 Chr.1: 26184384-26184384 on Build GRCh38
rs751956083 Chr.1: 26184591-26184591 on Build GRCh38
rs781493747 Chr.1: 26184516-26184516 on Build GRCh38
rs376434802 Chr.1: 26184560-26184560 on Build GRCh38
rs753667404 Chr.1: 26184530-26184530 on Build GRCh38
rs761414911 Chr.1: 26184556-26184556 on Build GRCh38
rs146265932 Chr.1: 26184598-26184598 on Build GRCh38
rs770969546 Chr.1: 26184304-26184304 on Build GRCh38
rs2297711 Chr.1: 26184648-26184648 on Build GRCh38
rs745730216 Chr.1: 26184628-26184628 on Build GRCh38
rs779564702 Chr.1: 26184361-26184361 on Build GRCh38
rs777954969 Chr.1: 26184619-26184619 on Build GRCh38
rs757554517 Chr.1: 26184356-26184356 on Build GRCh38
rs747196702 Chr.1: 26184276-26184276 on Build GRCh38
rs763869609 Chr.1: 26184454-26184454 on Build GRCh38
rs955141282 Chr.1: 26184592-26184592 on Build GRCh38
rs751732315 Chr.1: 26184503-26184503 on Build GRCh38
rs767394145 Chr.1: 26184572-26184572 on Build GRCh38
rs759427379 Chr.1: 26184444-26184444 on Build GRCh38
rs868393948 Chr.1: 26184355-26184355 on Build GRCh38
rs199807460 Chr.1: 26184230-26184230 on Build GRCh38
rs773112396 Chr.1: 26184285-26184285 on Build GRCh38
rs774208555 Chr.1: 26184312-26184312 on Build GRCh38
rs755720967 Chr.1: 26184393-26184393 on Build GRCh38
rs372878532 Chr.1: 26184652-26184652 on Build GRCh38
rs1023347759 Chr.1: 26184357-26184357 on Build GRCh38
rs57880419 Chr.1: 26184370-26184370 on Build GRCh38
rs752696920 Chr.1: 26184654-26184654 on Build GRCh38
rs2297710 Chr.1: 26184449-26184449 on Build GRCh38

View Full Product Details