Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62601617-62602138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL3
Entrez Gene ID
27329
Gene Name
angiopoietin like 3
Gene Aliases
ANG-5, ANGPT5, ANL3, FHBL2
UniGene
Hs.209153
Gene Chromosome Location
Chr.1: 62597487-62606305 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
UniGene
Hs.744927
Gene Chromosome Location
Chr.1: 62454726-62688368 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5785416 c.755G>A p.R252K Chr.1: 62601802-62601802 on Build GRCh38
6599649 c.744C>A p.T248T Chr.1: 62601791-62601791 on Build GRCh38
7355427 c.776T>C p.M259T Chr.1: 62601823-62601823 on Build GRCh38
7138401 c.755G>C p.R252T Chr.1: 62601802-62601802 on Build GRCh38
3772052 c.783C>T p.A261A Chr.1: 62601830-62601830 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200949988 Chr.1: 62601813-62601813 on Build GRCh38
rs145535735 Chr.1: 62601904-62601904 on Build GRCh38
rs746076650 Chr.1: 62601748-62601748 on Build GRCh38
rs913182728 Chr.1: 62601972-62601973 on Build GRCh38
rs191293319 Chr.1: 62601883-62601883 on Build GRCh38
rs892443494 Chr.1: 62601980-62601980 on Build GRCh38
rs556810307 Chr.1: 62601817-62601817 on Build GRCh38
rs769948598 Chr.1: 62601757-62601757 on Build GRCh38
rs115032502 Chr.1: 62601939-62601939 on Build GRCh38
rs770842763 Chr.1: 62601905-62601905 on Build GRCh38
rs191012841 Chr.1: 62602002-62602002 on Build GRCh38
rs938560345 Chr.1: 62601935-62601935 on Build GRCh38
rs538757784 Chr.1: 62601848-62601848 on Build GRCh38
rs757962265 Chr.1: 62601679-62601679 on Build GRCh38
rs1052425124 Chr.1: 62601948-62601948 on Build GRCh38
rs185472483 Chr.1: 62601923-62601923 on Build GRCh38
rs773165983 Chr.1: 62601784-62601784 on Build GRCh38
rs748026946 Chr.1: 62602056-62602056 on Build GRCh38
rs971756240 Chr.1: 62601811-62601811 on Build GRCh38
rs377050341 Chr.1: 62601907-62601907 on Build GRCh38
rs757750632 Chr.1: 62601835-62601835 on Build GRCh38
rs779553947 Chr.1: 62601827-62601827 on Build GRCh38
rs568204829 Chr.1: 62601836-62601836 on Build GRCh38
rs992806506 Chr.1: 62601829-62601829 on Build GRCh38
rs764880720 Chr.1: 62601927-62601927 on Build GRCh38
rs1040701201 Chr.1: 62602076-62602076 on Build GRCh38
rs879715812 Chr.1: 62602079-62602079 on Build GRCh38
rs891624648 Chr.1: 62601692-62601692 on Build GRCh38
rs774189215 Chr.1: 62601778-62601778 on Build GRCh38
rs763356461 Chr.1: 62601930-62601930 on Build GRCh38
rs1016095138 Chr.1: 62602099-62602099 on Build GRCh38
rs771162376 Chr.1: 62601729-62601729 on Build GRCh38
rs776665464 Chr.1: 62601744-62601744 on Build GRCh38
rs560184338 Chr.1: 62602087-62602087 on Build GRCh38
rs373994255 Chr.1: 62601994-62601994 on Build GRCh38
rs538866116 Chr.1: 62601667-62601667 on Build GRCh38
rs778380462 Chr.1: 62601825-62601825 on Build GRCh38
rs765893431 Chr.1: 62601796-62601796 on Build GRCh38
rs760130286 Chr.1: 62601787-62601787 on Build GRCh38
rs868528057 Chr.1: 62602085-62602085 on Build GRCh38
rs747898335 Chr.1: 62601859-62601859 on Build GRCh38
rs752131105 Chr.1: 62601830-62601830 on Build GRCh38
rs933284403 Chr.1: 62601993-62601993 on Build GRCh38
rs544069771 Chr.1: 62601979-62601979 on Build GRCh38
rs754658074 Chr.1: 62601812-62601812 on Build GRCh38
rs377002243 Chr.1: 62601719-62601719 on Build GRCh38
rs751008551 Chr.1: 62601911-62601911 on Build GRCh38
rs889517621 Chr.1: 62602050-62602050 on Build GRCh38
rs985004442 Chr.1: 62601876-62601876 on Build GRCh38
rs577719740 Chr.1: 62602016-62602016 on Build GRCh38
rs1011448161 Chr.1: 62601713-62601713 on Build GRCh38
rs180801384 Chr.1: 62601895-62601895 on Build GRCh38
rs141266927 Chr.1: 62601783-62601783 on Build GRCh38
rs746000132 Chr.1: 62601842-62601842 on Build GRCh38
rs780152039 Chr.1: 62601855-62601855 on Build GRCh38
rs544824600 Chr.1: 62602019-62602019 on Build GRCh38
rs141967457 Chr.1: 62601804-62601804 on Build GRCh38
rs370958533 Chr.1: 62601752-62601752 on Build GRCh38
rs752008847 Chr.1: 62601934-62601934 on Build GRCh38
rs77871363 Chr.1: 62601823-62601823 on Build GRCh38
rs781299623 Chr.1: 62601837-62601837 on Build GRCh38
rs867243797 Chr.1: 62601767-62601767 on Build GRCh38

View Full Product Details