Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154027218-154027712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECP2
Entrez Gene ID
4204
Gene Name
methyl-CpG binding protein 2
Gene Aliases
AUTSX3, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT
UniGene
Hs.200716
Gene Chromosome Location
Chr.X: 154021800-154097731 on Build GRCh38
Transcripts
NM_004992.3 XM_005274682.4 XM_006724819.3 XM_011531166.2 XM_017029533.1

SNP Detail

SNP ID Chromosome Location
rs1041874910 Chr.X: 154027686-154027686 on Build GRCh38
rs782737377 Chr.X: 154027540-154027540 on Build GRCh38
rs911336239 Chr.X: 154027662-154027662 on Build GRCh38
rs970955025 Chr.X: 154027438-154027438 on Build GRCh38
rs1011943842 Chr.X: 154027553-154027553 on Build GRCh38
rs972503004 Chr.X: 154027609-154027609 on Build GRCh38
rs972554369 Chr.X: 154027452-154027452 on Build GRCh38
rs970689569 Chr.X: 154027570-154027570 on Build GRCh38
rs781857417 Chr.X: 154027379-154027380 on Build GRCh38
rs918649444 Chr.X: 154027454-154027454 on Build GRCh38
rs267608358 Chr.X: 154027411-154027411 on Build GRCh38
rs1000625371 Chr.X: 154027541-154027541 on Build GRCh38
rs1048350389 Chr.X: 154027470-154027470 on Build GRCh38
rs1010790721 Chr.X: 154027499-154027499 on Build GRCh38
rs1000509245 Chr.X: 154027334-154027334 on Build GRCh38
rs952583250 Chr.X: 154027655-154027655 on Build GRCh38
rs959280082 Chr.X: 154027552-154027552 on Build GRCh38
rs917864473 Chr.X: 154027431-154027431 on Build GRCh38
rs902224885 Chr.X: 154027514-154027514 on Build GRCh38
rs1041066242 Chr.X: 154027673-154027673 on Build GRCh38
rs959284319 Chr.X: 154027428-154027428 on Build GRCh38
rs1041673518 Chr.X: 154027486-154027486 on Build GRCh38
rs946323430 Chr.X: 154027678-154027678 on Build GRCh38
rs782369979 Chr.X: 154027466-154027466 on Build GRCh38
rs782070600 Chr.X: 154027634-154027634 on Build GRCh38
rs1020701037 Chr.X: 154027277-154027277 on Build GRCh38
rs918428918 Chr.X: 154027642-154027642 on Build GRCh38
rs782299775 Chr.X: 154027317-154027317 on Build GRCh38
rs782604692 Chr.X: 154027429-154027429 on Build GRCh38
rs942752846 Chr.X: 154027482-154027482 on Build GRCh38
rs782265654 Chr.X: 154027606-154027606 on Build GRCh38
rs782237665 Chr.X: 154027495-154027495 on Build GRCh38
rs781980870 Chr.X: 154027676-154027676 on Build GRCh38
rs782311841 Chr.X: 154027520-154027520 on Build GRCh38
rs782672469 Chr.X: 154027257-154027257 on Build GRCh38
rs911216139 Chr.X: 154027477-154027477 on Build GRCh38
rs782378994 Chr.X: 154027392-154027392 on Build GRCh38
rs782600953 Chr.X: 154027551-154027551 on Build GRCh38
rs781927274 Chr.X: 154027513-154027513 on Build GRCh38
rs942684853 Chr.X: 154027670-154027670 on Build GRCh38
rs1010426181 Chr.X: 154027249-154027249 on Build GRCh38
rs966461691 Chr.X: 154027278-154027278 on Build GRCh38
rs1025284314 Chr.X: 154027567-154027567 on Build GRCh38

View Full Product Details