Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63609860-63610364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REEP3
Entrez Gene ID
221035
Gene Name
receptor accessory protein 3
Gene Aliases
C10orf74, Yip2b
UniGene
Hs.499833
Gene Chromosome Location
Chr.10: 63521353-63625123 on Build GRCh38
Transcripts
NM_001001330.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4989578 c.559T>A p.S187T Chr.10: 63610328-63610328 on Build GRCh38
4994147 c.419G>T p.S140I Chr.10: 63610188-63610188 on Build GRCh38
6042461 c.424G>A p.G142R Chr.10: 63610193-63610193 on Build GRCh38
5592841 c.521C>T p.P174L Chr.10: 63610290-63610290 on Build GRCh38
2158667 c.513A>G p.Q171Q Chr.10: 63610282-63610282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753619150 Chr.10: 63610250-63610250 on Build GRCh38
rs748442346 Chr.10: 63610317-63610317 on Build GRCh38
rs372963754 Chr.10: 63610208-63610208 on Build GRCh38
rs959467387 Chr.10: 63609964-63609964 on Build GRCh38
rs763966753 Chr.10: 63610245-63610245 on Build GRCh38
rs915755823 Chr.10: 63610056-63610056 on Build GRCh38
rs971706407 Chr.10: 63609910-63609910 on Build GRCh38
rs747603852 Chr.10: 63610160-63610160 on Build GRCh38
rs763012349 Chr.10: 63610229-63610229 on Build GRCh38
rs770208373 Chr.10: 63610328-63610328 on Build GRCh38
rs556616907 Chr.10: 63610022-63610022 on Build GRCh38
rs775266362 Chr.10: 63610232-63610232 on Build GRCh38
rs777016521 Chr.10: 63610190-63610190 on Build GRCh38
rs757960718 Chr.10: 63610292-63610292 on Build GRCh38
rs572495959 Chr.10: 63610301-63610301 on Build GRCh38
rs946105967 Chr.10: 63610046-63610046 on Build GRCh38
rs908483458 Chr.10: 63610209-63610209 on Build GRCh38
rs530091848 Chr.10: 63610240-63610240 on Build GRCh38
rs1040749870 Chr.10: 63610277-63610277 on Build GRCh38
rs990207894 Chr.10: 63609968-63609968 on Build GRCh38
rs1049636534 Chr.10: 63610138-63610138 on Build GRCh38
rs764979700 Chr.10: 63610284-63610284 on Build GRCh38
rs768876050 Chr.10: 63610161-63610161 on Build GRCh38
rs928023890 Chr.10: 63609906-63609906 on Build GRCh38
rs10995569 Chr.10: 63610281-63610281 on Build GRCh38
rs1034316054 Chr.10: 63609957-63609957 on Build GRCh38
rs746536032 Chr.10: 63610303-63610303 on Build GRCh38
rs779659595 Chr.10: 63610302-63610302 on Build GRCh38
rs931177282 Chr.10: 63610186-63610186 on Build GRCh38
rs980320167 Chr.10: 63609935-63609935 on Build GRCh38
rs762191127 Chr.10: 63610203-63610203 on Build GRCh38
rs922422601 Chr.10: 63610074-63610074 on Build GRCh38
rs750158390 Chr.10: 63610291-63610291 on Build GRCh38
rs755466573 Chr.10: 63610304-63610304 on Build GRCh38
rs766534851 Chr.10: 63610230-63610230 on Build GRCh38
rs185029281 Chr.10: 63610071-63610071 on Build GRCh38
rs933896774 Chr.10: 63610085-63610085 on Build GRCh38
rs991243963 Chr.10: 63610001-63610001 on Build GRCh38
rs539017172 Chr.10: 63610152-63610152 on Build GRCh38
rs773424285 Chr.10: 63610218-63610218 on Build GRCh38
rs113298040 Chr.10: 63610280-63610280 on Build GRCh38
rs781566505 Chr.10: 63610315-63610315 on Build GRCh38
rs753282196 Chr.10: 63610194-63610194 on Build GRCh38
rs939674172 Chr.10: 63610222-63610222 on Build GRCh38
rs760705999 Chr.10: 63610241-63610241 on Build GRCh38
rs913293944 Chr.10: 63610031-63610031 on Build GRCh38

View Full Product Details